Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101355966-101356472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
UniGene
Hs.159494
Gene Chromosome Location
Chr.X: 101349447-101390796 on Build GRCh38
Transcripts
NM_000061.2 NM_001287344.1 NM_001287345.1

SNP Detail

SNP ID Chromosome Location
rs372089648 Chr.X: 101356216-101356216 on Build GRCh38
rs1053430211 Chr.X: 101356027-101356027 on Build GRCh38
rs128621202 Chr.X: 101356059-101356059 on Build GRCh38
rs374696747 Chr.X: 101356019-101356019 on Build GRCh38
rs561853537 Chr.X: 101356133-101356133 on Build GRCh38
rs782173857 Chr.X: 101356023-101356023 on Build GRCh38
rs193922125 Chr.X: 101356107-101356107 on Build GRCh38
rs781869026 Chr.X: 101356081-101356081 on Build GRCh38
rs528859775 Chr.X: 101356309-101356309 on Build GRCh38
rs41310709 Chr.X: 101356112-101356112 on Build GRCh38
rs782604457 Chr.X: 101356277-101356277 on Build GRCh38
rs782342481 Chr.X: 101356001-101356001 on Build GRCh38
rs368331876 Chr.X: 101356301-101356301 on Build GRCh38
rs563329397 Chr.X: 101356285-101356285 on Build GRCh38
rs128621200 Chr.X: 101356102-101356102 on Build GRCh38
rs782338603 Chr.X: 101356143-101356143 on Build GRCh38
rs782194510 Chr.X: 101355995-101355995 on Build GRCh38
rs782238208 Chr.X: 101356393-101356393 on Build GRCh38
rs192689502 Chr.X: 101356079-101356079 on Build GRCh38
rs781792640 Chr.X: 101356231-101356231 on Build GRCh38
rs373846453 Chr.X: 101356226-101356226 on Build GRCh38
rs371298091 Chr.X: 101356011-101356011 on Build GRCh38
rs184722523 Chr.X: 101356384-101356384 on Build GRCh38
rs782571484 Chr.X: 101355994-101355994 on Build GRCh38
rs3027643 Chr.X: 101356444-101356444 on Build GRCh38
rs1026961201 Chr.X: 101356337-101356337 on Build GRCh38
rs782093826 Chr.X: 101356076-101356076 on Build GRCh38
rs128621201 Chr.X: 101356060-101356060 on Build GRCh38
rs781986167 Chr.X: 101356190-101356190 on Build GRCh38
rs782457670 Chr.X: 101356260-101356260 on Build GRCh38
rs782391685 Chr.X: 101356407-101356407 on Build GRCh38
rs782459493 Chr.X: 101356299-101356299 on Build GRCh38
rs781946466 Chr.X: 101356045-101356045 on Build GRCh38
rs3747288 Chr.X: 101356297-101356297 on Build GRCh38
rs961449100 Chr.X: 101356306-101356306 on Build GRCh38
rs367698178 Chr.X: 101356034-101356034 on Build GRCh38
rs193922124 Chr.X: 101356163-101356163 on Build GRCh38

View Full Product Details