Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 81268965-81269483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STON2
Entrez Gene ID
85439
Gene Name
stonin 2
Gene Aliases
STN2, STNB, STNB2
UniGene
Hs.14248
Gene Chromosome Location
Chr.14: 81260650-81427404 on Build GRCh38
Transcripts
NM_001256430.1 NM_033104.3 XM_006720279.1 XM_011537240.1 XM_011537242.1

SNP Detail

SNP ID Chromosome Location
rs964317262 Chr.14: 81269137-81269137 on Build GRCh38
rs188230565 Chr.14: 81269163-81269163 on Build GRCh38
rs771174698 Chr.14: 81269076-81269076 on Build GRCh38
rs78064742 Chr.14: 81269458-81269458 on Build GRCh38
rs1029567639 Chr.14: 81269393-81269393 on Build GRCh38
rs191491369 Chr.14: 81269460-81269460 on Build GRCh38
rs554994023 Chr.14: 81269278-81269278 on Build GRCh38
rs892279750 Chr.14: 81269041-81269041 on Build GRCh38
rs962842928 Chr.14: 81269115-81269115 on Build GRCh38
rs975265493 Chr.14: 81269153-81269153 on Build GRCh38
rs899945987 Chr.14: 81269047-81269047 on Build GRCh38
rs995642135 Chr.14: 81269439-81269439 on Build GRCh38
rs770040918 Chr.14: 81269083-81269083 on Build GRCh38
rs939256915 Chr.14: 81269020-81269020 on Build GRCh38
rs536794859 Chr.14: 81269367-81269367 on Build GRCh38
rs373589105 Chr.14: 81269335-81269335 on Build GRCh38
rs757284373 Chr.14: 81269435-81269435 on Build GRCh38
rs774996139 Chr.14: 81269334-81269334 on Build GRCh38
rs8007595 Chr.14: 81269221-81269221 on Build GRCh38
rs939052974 Chr.14: 81269261-81269261 on Build GRCh38
rs548581626 Chr.14: 81269044-81269044 on Build GRCh38
rs753629176 Chr.14: 81269427-81269427 on Build GRCh38
rs575049014 Chr.14: 81269098-81269098 on Build GRCh38
rs1045133968 Chr.14: 81268995-81268995 on Build GRCh38
rs17111638 Chr.14: 81269371-81269371 on Build GRCh38
rs1052401096 Chr.14: 81269021-81269021 on Build GRCh38
rs536858397 Chr.14: 81269380-81269380 on Build GRCh38
rs533812820 Chr.14: 81269269-81269269 on Build GRCh38
rs8007436 Chr.14: 81269160-81269160 on Build GRCh38
rs1004231141 Chr.14: 81269383-81269383 on Build GRCh38
rs570878646 Chr.14: 81269452-81269452 on Build GRCh38
rs938253060 Chr.14: 81269271-81269271 on Build GRCh38
rs185607490 Chr.14: 81269338-81269338 on Build GRCh38
rs765227227 Chr.14: 81269451-81269451 on Build GRCh38
rs529927245 Chr.14: 81269339-81269339 on Build GRCh38
rs960831946 Chr.14: 81269027-81269027 on Build GRCh38
rs552206621 Chr.14: 81269450-81269450 on Build GRCh38
rs563849720 Chr.14: 81269126-81269126 on Build GRCh38
rs927615358 Chr.14: 81269257-81269257 on Build GRCh38
rs542543331 Chr.14: 81269119-81269119 on Build GRCh38
rs532082647 Chr.14: 81269247-81269247 on Build GRCh38
rs573309492 Chr.14: 81269433-81269433 on Build GRCh38
rs955441615 Chr.14: 81269086-81269086 on Build GRCh38
rs557302883 Chr.14: 81269268-81269268 on Build GRCh38
rs913669729 Chr.14: 81269216-81269216 on Build GRCh38
rs773832453 Chr.14: 81269156-81269156 on Build GRCh38
rs759018666 Chr.14: 81269299-81269299 on Build GRCh38
rs896738883 Chr.14: 81269292-81269292 on Build GRCh38
rs117853183 Chr.14: 81269046-81269046 on Build GRCh38
rs955277982 Chr.14: 81269406-81269406 on Build GRCh38
rs996819155 Chr.14: 81269057-81269057 on Build GRCh38
rs1052766782 Chr.14: 81269296-81269296 on Build GRCh38
rs868382138 Chr.14: 81269011-81269011 on Build GRCh38
rs927871220 Chr.14: 81269001-81269001 on Build GRCh38
rs1056693167 Chr.14: 81269291-81269291 on Build GRCh38
rs1035200051 Chr.14: 81269022-81269022 on Build GRCh38
rs138920649 Chr.14: 81269091-81269091 on Build GRCh38
rs946529067 Chr.14: 81269302-81269302 on Build GRCh38
rs181064045 Chr.14: 81269310-81269310 on Build GRCh38

View Full Product Details