Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19502773-19503267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDE1
Entrez Gene ID
51573
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
363E6.2, MIR16
UniGene
Hs.512607
Gene Chromosome Location
Chr.16: 19501693-19522145 on Build GRCh38
Transcripts
NM_001324066.1 NM_001324067.1 NM_016641.3 NR_136689.1

SNP Detail

SNP ID Chromosome Location
rs760470244 Chr.16: 19502934-19502934 on Build GRCh38
rs943778174 Chr.16: 19503063-19503063 on Build GRCh38
rs971463332 Chr.16: 19502894-19502894 on Build GRCh38
rs564469686 Chr.16: 19503238-19503238 on Build GRCh38
rs570056516 Chr.16: 19503112-19503112 on Build GRCh38
rs183920599 Chr.16: 19503042-19503042 on Build GRCh38
rs981468136 Chr.16: 19502899-19502899 on Build GRCh38
rs1037130376 Chr.16: 19503081-19503081 on Build GRCh38
rs573647601 Chr.16: 19502985-19502985 on Build GRCh38
rs1013124594 Chr.16: 19503173-19503173 on Build GRCh38
rs1002975674 Chr.16: 19503228-19503228 on Build GRCh38
rs2521795 Chr.16: 19502819-19502819 on Build GRCh38
rs561967642 Chr.16: 19502982-19502982 on Build GRCh38
rs1020088710 Chr.16: 19503091-19503091 on Build GRCh38
rs186701458 Chr.16: 19503239-19503239 on Build GRCh38
rs976009112 Chr.16: 19503130-19503130 on Build GRCh38
rs146678235 Chr.16: 19503146-19503146 on Build GRCh38
rs920077977 Chr.16: 19503136-19503136 on Build GRCh38
rs967313338 Chr.16: 19503061-19503061 on Build GRCh38
rs896837329 Chr.16: 19503126-19503126 on Build GRCh38
rs543974197 Chr.16: 19502986-19502986 on Build GRCh38
rs1053138 Chr.16: 19503003-19503003 on Build GRCh38
rs756817784 Chr.16: 19502795-19502795 on Build GRCh38
rs1048787490 Chr.16: 19502855-19502855 on Build GRCh38
rs1023125522 Chr.16: 19503188-19503188 on Build GRCh38
rs774783879 Chr.16: 19503169-19503169 on Build GRCh38
rs1028299613 Chr.16: 19502801-19502801 on Build GRCh38
rs887987465 Chr.16: 19503172-19503172 on Build GRCh38
rs549164343 Chr.16: 19503210-19503210 on Build GRCh38
rs956209393 Chr.16: 19502935-19502935 on Build GRCh38
rs745819260 Chr.16: 19502864-19502864 on Build GRCh38
rs76065554 Chr.16: 19503029-19503029 on Build GRCh38
rs1011442784 Chr.16: 19503018-19503018 on Build GRCh38
rs540454479 Chr.16: 19502976-19502976 on Build GRCh38
rs779919317 Chr.16: 19503062-19503062 on Build GRCh38
rs7135 Chr.16: 19503161-19503161 on Build GRCh38
rs924616404 Chr.16: 19502916-19502916 on Build GRCh38
rs374313286 Chr.16: 19503089-19503090 on Build GRCh38
rs1008320913 Chr.16: 19503078-19503078 on Build GRCh38
rs1023058493 Chr.16: 19502876-19502876 on Build GRCh38
rs893085977 Chr.16: 19502987-19502987 on Build GRCh38
rs1045850684 Chr.16: 19502877-19502877 on Build GRCh38
rs914436684 Chr.16: 19502974-19502974 on Build GRCh38
rs990095650 Chr.16: 19502958-19502958 on Build GRCh38
rs758729768 Chr.16: 19503115-19503116 on Build GRCh38

View Full Product Details