Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61800443-61800885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 XM_011545022.2

SNP Detail

SNP ID Chromosome Location
rs770697344 Chr.11: 61800727-61800727 on Build GRCh38
rs527659403 Chr.11: 61800689-61800689 on Build GRCh38
rs527762120 Chr.11: 61800598-61800598 on Build GRCh38
rs939111460 Chr.11: 61800770-61800770 on Build GRCh38
rs964524706 Chr.11: 61800522-61800522 on Build GRCh38
rs149991368 Chr.11: 61800623-61800623 on Build GRCh38
rs968936752 Chr.11: 61800494-61800494 on Build GRCh38
rs545419144 Chr.11: 61800533-61800533 on Build GRCh38
rs1035733740 Chr.11: 61800475-61800475 on Build GRCh38
rs986956417 Chr.11: 61800477-61800477 on Build GRCh38
rs990400430 Chr.11: 61800495-61800495 on Build GRCh38
rs776678196 Chr.11: 61800773-61800773 on Build GRCh38
rs552883766 Chr.11: 61800520-61800520 on Build GRCh38
rs911364741 Chr.11: 61800478-61800478 on Build GRCh38
rs369065043 Chr.11: 61800532-61800532 on Build GRCh38
rs181067085 Chr.11: 61800483-61800483 on Build GRCh38
rs8448 Chr.11: 61800465-61800465 on Build GRCh38
rs923005906 Chr.11: 61800577-61800578 on Build GRCh38
rs946332151 Chr.11: 61800599-61800599 on Build GRCh38
rs746804609 Chr.11: 61800592-61800592 on Build GRCh38
rs909150354 Chr.11: 61800640-61800640 on Build GRCh38
rs745850376 Chr.11: 61800817-61800817 on Build GRCh38
rs976232688 Chr.11: 61800562-61800562 on Build GRCh38
rs562688177 Chr.11: 61800480-61800480 on Build GRCh38

View Full Product Details