Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 18324725-18325299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2G
Entrez Gene ID
5288
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma
Gene Aliases
PI3K-C2-gamma, PI3K-C2GAMMA
UniGene
Hs.22500
Gene Chromosome Location
Chr.12: 18242946-18716861 on Build GRCh38
Transcripts
NM_001288772.1 NM_001288774.1 NM_004570.5 XM_011520696.2 XM_011520697.2 XM_011520700.2 XM_011520701.2 XM_017019470.1 XM_017019471.1 XM_017019472.1 XM_017019473.1 XM_017019474.1 XM_017019475.1 XM_017019476.1 XM_017019477.1 XM_017019478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6957511 c.1216C>A p.L406I Chr.12: 18325042-18325042 on Build GRCh38
4438444 c.1209-4C>T p.? Chr.12: 18325031-18325031 on Build GRCh38
5733411 c.1238A>G p.Y413C Chr.12: 18325064-18325064 on Build GRCh38
3458754 c.1245C>T p.F415F Chr.12: 18325071-18325071 on Build GRCh38
6962771 c.1268C>T p.T423I Chr.12: 18325094-18325094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7970755 Chr.12: 18324831-18324831 on Build GRCh38
rs537562060 Chr.12: 18324875-18324875 on Build GRCh38
rs776566334 Chr.12: 18325010-18325010 on Build GRCh38
rs193099085 Chr.12: 18325014-18325014 on Build GRCh38
rs949916834 Chr.12: 18325065-18325065 on Build GRCh38
rs775359894 Chr.12: 18325132-18325132 on Build GRCh38
rs767018919 Chr.12: 18325254-18325254 on Build GRCh38
rs764280933 Chr.12: 18325032-18325035 on Build GRCh38
rs929925017 Chr.12: 18325269-18325269 on Build GRCh38
rs943429545 Chr.12: 18324762-18324762 on Build GRCh38
rs986585687 Chr.12: 18325175-18325175 on Build GRCh38
rs769672508 Chr.12: 18325123-18325123 on Build GRCh38
rs1035493932 Chr.12: 18325038-18325038 on Build GRCh38
rs776762080 Chr.12: 18325091-18325091 on Build GRCh38
rs780494846 Chr.12: 18325071-18325071 on Build GRCh38
rs757530460 Chr.12: 18325098-18325098 on Build GRCh38
rs988494419 Chr.12: 18324887-18324887 on Build GRCh38
rs576444794 Chr.12: 18325209-18325209 on Build GRCh38
rs752652519 Chr.12: 18325010-18325011 on Build GRCh38
rs758906166 Chr.12: 18324874-18324874 on Build GRCh38
rs1032679988 Chr.12: 18324880-18324880 on Build GRCh38
rs553435157 Chr.12: 18324937-18324937 on Build GRCh38
rs994044585 Chr.12: 18324928-18324928 on Build GRCh38
rs551172484 Chr.12: 18324878-18324878 on Build GRCh38
rs536941726 Chr.12: 18324932-18324932 on Build GRCh38
rs534282498 Chr.12: 18325039-18325039 on Build GRCh38
rs765538409 Chr.12: 18325026-18325026 on Build GRCh38
rs957299305 Chr.12: 18324884-18324884 on Build GRCh38
rs759447999 Chr.12: 18325025-18325025 on Build GRCh38
rs143331698 Chr.12: 18325053-18325053 on Build GRCh38
rs898327573 Chr.12: 18325225-18325225 on Build GRCh38
rs978329825 Chr.12: 18325153-18325153 on Build GRCh38
rs756421600 Chr.12: 18325054-18325054 on Build GRCh38
rs753793459 Chr.12: 18325070-18325080 on Build GRCh38
rs772780048 Chr.12: 18324987-18324987 on Build GRCh38
rs777131759 Chr.12: 18325082-18325082 on Build GRCh38
rs889649919 Chr.12: 18324862-18324862 on Build GRCh38
rs370377807 Chr.12: 18325076-18325076 on Build GRCh38
rs760893815 Chr.12: 18325019-18325020 on Build GRCh38
rs372577937 Chr.12: 18325110-18325110 on Build GRCh38
rs566064147 Chr.12: 18324809-18324809 on Build GRCh38
rs1014120056 Chr.12: 18324846-18324846 on Build GRCh38
rs771047800 Chr.12: 18325002-18325002 on Build GRCh38
rs17475920 Chr.12: 18325192-18325192 on Build GRCh38
rs920682228 Chr.12: 18324984-18324984 on Build GRCh38
rs375613945 Chr.12: 18325147-18325147 on Build GRCh38
rs968328238 Chr.12: 18325142-18325142 on Build GRCh38
rs974878175 Chr.12: 18325196-18325196 on Build GRCh38
rs1026308330 Chr.12: 18324961-18324961 on Build GRCh38
rs751675110 Chr.12: 18325045-18325045 on Build GRCh38
rs1051274663 Chr.12: 18324853-18324853 on Build GRCh38
rs77216941 Chr.12: 18325138-18325138 on Build GRCh38
rs369941931 Chr.12: 18325031-18325031 on Build GRCh38
rs866967779 Chr.12: 18324914-18324914 on Build GRCh38
rs543148159 Chr.12: 18325169-18325169 on Build GRCh38
rs778949550 Chr.12: 18325107-18325107 on Build GRCh38
rs558643322 Chr.12: 18324837-18324837 on Build GRCh38
rs190529173 Chr.12: 18324847-18324847 on Build GRCh38
rs892869584 Chr.12: 18325094-18325094 on Build GRCh38
rs746599962 Chr.12: 18325084-18325084 on Build GRCh38
rs933632230 Chr.12: 18325170-18325170 on Build GRCh38
rs747684196 Chr.12: 18325073-18325073 on Build GRCh38
rs571738840 Chr.12: 18324856-18324856 on Build GRCh38
rs943460727 Chr.12: 18324946-18324946 on Build GRCh38
rs373124655 Chr.12: 18325051-18325051 on Build GRCh38
rs781692740 Chr.12: 18325052-18325052 on Build GRCh38
rs577297603 Chr.12: 18325146-18325146 on Build GRCh38
rs912037988 Chr.12: 18324944-18324944 on Build GRCh38
rs759572739 Chr.12: 18325102-18325102 on Build GRCh38
rs910928534 Chr.12: 18325177-18325177 on Build GRCh38
rs770963929 Chr.12: 18325086-18325086 on Build GRCh38
rs991107020 Chr.12: 18324776-18324776 on Build GRCh38
rs567892023 Chr.12: 18324879-18324879 on Build GRCh38
rs534107628 Chr.12: 18324748-18324748 on Build GRCh38
rs1039029348 Chr.12: 18324788-18324788 on Build GRCh38
rs763538150 Chr.12: 18324791-18324791 on Build GRCh38
rs946371117 Chr.12: 18324808-18324808 on Build GRCh38
rs752816686 Chr.12: 18325030-18325030 on Build GRCh38

View Full Product Details