Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58279531-58280121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_017023176.1 XM_017023177.1 NM_001080492.1 NM_001305173.1 NM_001305174.1

Gene Symbol
PRSS54
Entrez Gene ID
221191
Gene Name
protease, serine 54
Gene Aliases
CT67, KLKBL4
UniGene
Hs.411239
Gene Chromosome Location
Chr.16: 58279997-58295047 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_017023176.1 XM_017023177.1 NM_001080492.1 NM_001305173.1 NM_001305174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2992719 c.1110G>T p.Q370H Chr.16: 58279753-58279753 on Build GRCh38
1236392 c.1107G>T p.L369F Chr.16: 58279750-58279750 on Build GRCh38
3510608 c.1066C>T p.R356C Chr.16: 58279709-58279709 on Build GRCh38
6621322 c.1067G>A p.R356H Chr.16: 58279710-58279710 on Build GRCh38
4433357 c.1048-4delG p.? Chr.16: 58279687-58279687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551147249 Chr.16: 58280085-58280085 on Build GRCh38
rs775715167 Chr.16: 58279795-58279795 on Build GRCh38
rs969187625 Chr.16: 58279568-58279568 on Build GRCh38
rs912164622 Chr.16: 58279931-58279931 on Build GRCh38
rs1022820785 Chr.16: 58279889-58279889 on Build GRCh38
rs939759309 Chr.16: 58280065-58280065 on Build GRCh38
rs921927688 Chr.16: 58279619-58279619 on Build GRCh38
rs931037460 Chr.16: 58279732-58279732 on Build GRCh38
rs544800178 Chr.16: 58279961-58279961 on Build GRCh38
rs932273428 Chr.16: 58279644-58279644 on Build GRCh38
rs911872576 Chr.16: 58279973-58279973 on Build GRCh38
rs757549278 Chr.16: 58279676-58279676 on Build GRCh38
rs770661545 Chr.16: 58279754-58279754 on Build GRCh38
rs763584066 Chr.16: 58279724-58279724 on Build GRCh38
rs746152284 Chr.16: 58279792-58279792 on Build GRCh38
rs904366215 Chr.16: 58280062-58280062 on Build GRCh38
rs1041588631 Chr.16: 58280059-58280059 on Build GRCh38
rs6499945 Chr.16: 58279662-58279662 on Build GRCh38
rs201008433 Chr.16: 58279710-58279710 on Build GRCh38
rs371518457 Chr.16: 58279752-58279752 on Build GRCh38
rs968862901 Chr.16: 58279846-58279846 on Build GRCh38
rs71155257 Chr.16: 58279675-58279675 on Build GRCh38
rs937005205 Chr.16: 58280016-58280016 on Build GRCh38
rs16960012 Chr.16: 58279845-58279845 on Build GRCh38
rs372219833 Chr.16: 58279709-58279709 on Build GRCh38
rs774267633 Chr.16: 58279658-58279658 on Build GRCh38
rs754545544 Chr.16: 58279682-58279682 on Build GRCh38
rs763097550 Chr.16: 58279710-58279711 on Build GRCh38
rs543143055 Chr.16: 58279676-58279676 on Build GRCh38
rs545274065 Chr.16: 58279675-58279676 on Build GRCh38
rs765183426 Chr.16: 58279656-58279656 on Build GRCh38
rs978560683 Chr.16: 58279903-58279903 on Build GRCh38
rs978758629 Chr.16: 58279573-58279573 on Build GRCh38
rs139414325 Chr.16: 58279705-58279705 on Build GRCh38
rs759382277 Chr.16: 58279659-58279666 on Build GRCh38
rs759677793 Chr.16: 58279654-58279654 on Build GRCh38
rs991374779 Chr.16: 58279657-58279657 on Build GRCh38
rs956113883 Chr.16: 58279963-58279963 on Build GRCh38
rs766708919 Chr.16: 58279640-58279640 on Build GRCh38
rs112404453 Chr.16: 58279972-58279972 on Build GRCh38
rs762798110 Chr.16: 58279937-58279937 on Build GRCh38
rs753380533 Chr.16: 58279679-58279679 on Build GRCh38
rs563620368 Chr.16: 58279956-58279956 on Build GRCh38
rs369177906 Chr.16: 58280086-58280086 on Build GRCh38
rs141160927 Chr.16: 58279690-58279690 on Build GRCh38
rs562373794 Chr.16: 58279674-58279674 on Build GRCh38
rs182316342 Chr.16: 58279788-58279788 on Build GRCh38
rs562782532 Chr.16: 58279676-58279677 on Build GRCh38
rs931089949 Chr.16: 58279964-58279964 on Build GRCh38
rs367810849 Chr.16: 58279715-58279715 on Build GRCh38
rs146122026 Chr.16: 58279725-58279725 on Build GRCh38
rs770882418 Chr.16: 58279775-58279775 on Build GRCh38
rs750583501 Chr.16: 58279675-58279677 on Build GRCh38
rs776929956 Chr.16: 58279645-58279645 on Build GRCh38
rs770794619 Chr.16: 58279707-58279707 on Build GRCh38
rs767063429 Chr.16: 58279803-58279804 on Build GRCh38
rs999949976 Chr.16: 58280063-58280063 on Build GRCh38
rs895591618 Chr.16: 58280081-58280081 on Build GRCh38
rs763034728 Chr.16: 58279822-58279822 on Build GRCh38
rs753565927 Chr.16: 58279743-58279743 on Build GRCh38
rs1035330324 Chr.16: 58279848-58279848 on Build GRCh38
rs111617606 Chr.16: 58279939-58279939 on Build GRCh38
rs571831778 Chr.16: 58279988-58279988 on Build GRCh38
rs974769621 Chr.16: 58280018-58280018 on Build GRCh38
rs780656305 Chr.16: 58279577-58279577 on Build GRCh38
rs150895046 Chr.16: 58279701-58279701 on Build GRCh38
rs375397864 Chr.16: 58279674-58279675 on Build GRCh38
rs533790150 Chr.16: 58279738-58279738 on Build GRCh38
rs558243329 Chr.16: 58279886-58279886 on Build GRCh38
rs866834481 Chr.16: 58279885-58279885 on Build GRCh38
rs895759461 Chr.16: 58280029-58280029 on Build GRCh38
rs772125273 Chr.16: 58279699-58279699 on Build GRCh38
rs148468373 Chr.16: 58279891-58279891 on Build GRCh38
rs1057303997 Chr.16: 58280066-58280066 on Build GRCh38
rs959945299 Chr.16: 58279562-58279562 on Build GRCh38
rs751917421 Chr.16: 58279672-58279672 on Build GRCh38
rs1012525854 Chr.16: 58280091-58280091 on Build GRCh38
rs1029046504 Chr.16: 58280070-58280070 on Build GRCh38
rs968542389 Chr.16: 58279658-58279658 on Build GRCh38
rs79413375 Chr.16: 58279684-58279686 on Build GRCh38
rs17241091 Chr.16: 58280044-58280044 on Build GRCh38
rs764499155 Chr.16: 58279666-58279666 on Build GRCh38
rs4784915 Chr.16: 58279902-58279902 on Build GRCh38
rs199813526 Chr.16: 58279675-58279675 on Build GRCh38
rs895696136 Chr.16: 58279812-58279812 on Build GRCh38
rs924688258 Chr.16: 58279667-58279667 on Build GRCh38
rs886615223 Chr.16: 58280060-58280060 on Build GRCh38
rs762187012 Chr.16: 58279727-58279727 on Build GRCh38
rs779693561 Chr.16: 58279677-58279677 on Build GRCh38
rs539588982 Chr.16: 58279882-58279882 on Build GRCh38
rs149602136 Chr.16: 58279708-58279708 on Build GRCh38
rs746897992 Chr.16: 58279769-58279769 on Build GRCh38
rs199864869 Chr.16: 58279744-58279744 on Build GRCh38
rs188622492 Chr.16: 58279824-58279824 on Build GRCh38
rs746677093 Chr.16: 58279747-58279747 on Build GRCh38
rs758801282 Chr.16: 58279663-58279663 on Build GRCh38
rs201119159 Chr.16: 58279793-58279793 on Build GRCh38
rs34834166 Chr.16: 58279931-58279932 on Build GRCh38
rs757919729 Chr.16: 58279760-58279760 on Build GRCh38
rs1057180222 Chr.16: 58280028-58280028 on Build GRCh38

View Full Product Details