Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65825968-65826564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
UniGene
Hs.408676
Gene Chromosome Location
Chr.17: 65635540-66192131 on Build GRCh38
Transcripts
NM_001037325.2 NM_001199165.2 NM_001302891.1 NR_126542.1 XM_005257119.4 XM_005257125.3 XM_005257126.4 XM_006721740.2 XM_011524461.2 XM_011524462.2 XM_011524463.2 XM_011524464.2 XM_011524465.2 XM_011524466.2 XM_011524467.2 XM_017024302.1 XM_017024303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231971 c.92C>T p.P31L Chr.17: 65826208-65826208 on Build GRCh38
4546895 c.40G>A p.E14K Chr.17: 65826260-65826260 on Build GRCh38
6626750 c.60G>C p.L20L Chr.17: 65826240-65826240 on Build GRCh38
3755755 c.61A>G p.I21V Chr.17: 65826239-65826239 on Build GRCh38
2796824 c.23A>G p.D8G Chr.17: 65826277-65826277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911214598 Chr.17: 65826107-65826107 on Build GRCh38
rs930081059 Chr.17: 65826426-65826426 on Build GRCh38
rs533465133 Chr.17: 65826362-65826362 on Build GRCh38
rs1034633666 Chr.17: 65826248-65826248 on Build GRCh38
rs757364012 Chr.17: 65826194-65826194 on Build GRCh38
rs573727987 Chr.17: 65826266-65826266 on Build GRCh38
rs930658175 Chr.17: 65826122-65826122 on Build GRCh38
rs753600068 Chr.17: 65825999-65825999 on Build GRCh38
rs199938505 Chr.17: 65826260-65826260 on Build GRCh38
rs1003080415 Chr.17: 65826489-65826490 on Build GRCh38
rs769039753 Chr.17: 65826344-65826344 on Build GRCh38
rs754239419 Chr.17: 65826174-65826174 on Build GRCh38
rs143472634 Chr.17: 65826018-65826018 on Build GRCh38
rs747734506 Chr.17: 65826069-65826069 on Build GRCh38
rs759102227 Chr.17: 65826145-65826145 on Build GRCh38
rs540077908 Chr.17: 65826088-65826088 on Build GRCh38
rs777364871 Chr.17: 65826213-65826213 on Build GRCh38
rs181051275 Chr.17: 65826112-65826112 on Build GRCh38
rs780240239 Chr.17: 65826291-65826291 on Build GRCh38
rs769800714 Chr.17: 65826212-65826212 on Build GRCh38
rs141709389 Chr.17: 65826247-65826247 on Build GRCh38
rs577878792 Chr.17: 65826125-65826125 on Build GRCh38
rs770472687 Chr.17: 65826227-65826227 on Build GRCh38
rs781490391 Chr.17: 65826453-65826453 on Build GRCh38
rs751662706 Chr.17: 65826301-65826301 on Build GRCh38
rs942690790 Chr.17: 65826396-65826396 on Build GRCh38
rs1022707514 Chr.17: 65826342-65826342 on Build GRCh38
rs564669312 Chr.17: 65826357-65826357 on Build GRCh38
rs746784523 Chr.17: 65826213-65826214 on Build GRCh38
rs778705550 Chr.17: 65826310-65826310 on Build GRCh38
rs114616764 Chr.17: 65826200-65826200 on Build GRCh38
rs2191966 Chr.17: 65826126-65826126 on Build GRCh38
rs548948115 Chr.17: 65826436-65826436 on Build GRCh38
rs367546613 Chr.17: 65826407-65826407 on Build GRCh38
rs374112500 Chr.17: 65826205-65826205 on Build GRCh38
rs188833110 Chr.17: 65826305-65826305 on Build GRCh38
rs777236967 Chr.17: 65826349-65826349 on Build GRCh38
rs749117513 Chr.17: 65826223-65826223 on Build GRCh38
rs994717452 Chr.17: 65826179-65826179 on Build GRCh38
rs750438666 Chr.17: 65826543-65826543 on Build GRCh38
rs774053308 Chr.17: 65826229-65826229 on Build GRCh38
rs535087980 Chr.17: 65826533-65826533 on Build GRCh38
rs919230866 Chr.17: 65826044-65826044 on Build GRCh38
rs761269680 Chr.17: 65826162-65826162 on Build GRCh38
rs367967415 Chr.17: 65826254-65826254 on Build GRCh38
rs755946515 Chr.17: 65826303-65826303 on Build GRCh38
rs78053636 Chr.17: 65826336-65826336 on Build GRCh38
rs557909782 Chr.17: 65826123-65826123 on Build GRCh38
rs151095214 Chr.17: 65826304-65826304 on Build GRCh38
rs759012523 Chr.17: 65826238-65826238 on Build GRCh38
rs377650019 Chr.17: 65826104-65826104 on Build GRCh38
rs954593981 Chr.17: 65826343-65826343 on Build GRCh38
rs1047642010 Chr.17: 65826463-65826463 on Build GRCh38
rs75168639 Chr.17: 65826000-65826000 on Build GRCh38
rs56237024 Chr.17: 65826133-65826133 on Build GRCh38
rs773079877 Chr.17: 65826113-65826113 on Build GRCh38
rs780006304 Chr.17: 65826138-65826140 on Build GRCh38
rs765526217 Chr.17: 65826271-65826271 on Build GRCh38
rs747985159 Chr.17: 65826211-65826211 on Build GRCh38
rs62065084 Chr.17: 65826239-65826239 on Build GRCh38
rs764678953 Chr.17: 65826249-65826249 on Build GRCh38
rs914361617 Chr.17: 65826001-65826001 on Build GRCh38
rs565862728 Chr.17: 65826532-65826532 on Build GRCh38
rs986009419 Chr.17: 65826360-65826360 on Build GRCh38
rs972265510 Chr.17: 65826534-65826534 on Build GRCh38
rs750528224 Chr.17: 65826201-65826201 on Build GRCh38
rs1038349586 Chr.17: 65826403-65826403 on Build GRCh38
rs1038398829 Chr.17: 65826142-65826142 on Build GRCh38
rs753254925 Chr.17: 65826157-65826157 on Build GRCh38
rs765058484 Chr.17: 65826414-65826414 on Build GRCh38
rs1002630232 Chr.17: 65826424-65826424 on Build GRCh38
rs933269848 Chr.17: 65826081-65826081 on Build GRCh38
rs570010313 Chr.17: 65826371-65826371 on Build GRCh38
rs371182498 Chr.17: 65826105-65826105 on Build GRCh38
rs773789895 Chr.17: 65826132-65826132 on Build GRCh38
rs977310702 Chr.17: 65826051-65826051 on Build GRCh38
rs892781488 Chr.17: 65826319-65826319 on Build GRCh38
rs923152032 Chr.17: 65826070-65826070 on Build GRCh38
rs1035118395 Chr.17: 65826464-65826464 on Build GRCh38
rs758714456 Chr.17: 65826278-65826278 on Build GRCh38
rs768005776 Chr.17: 65826151-65826151 on Build GRCh38
rs542638941 Chr.17: 65826276-65826276 on Build GRCh38
rs768496288 Chr.17: 65826308-65826309 on Build GRCh38
rs868506202 Chr.17: 65826272-65826272 on Build GRCh38
rs764329039 Chr.17: 65826172-65826172 on Build GRCh38
rs919984175 Chr.17: 65826416-65826416 on Build GRCh38
rs550402513 Chr.17: 65826363-65826363 on Build GRCh38
rs907359161 Chr.17: 65826480-65826480 on Build GRCh38
rs774809746 Chr.17: 65826339-65826339 on Build GRCh38

View Full Product Details