Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113354059-113354476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSPRY
Entrez Gene ID
54836
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
UniGene
Hs.614517
Gene Chromosome Location
Chr.9: 113349532-113371233 on Build GRCh38
Transcripts
NM_001317943.1 NM_001317944.1 NM_017688.2 XM_006717149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350859 c.300+8C>G p.? Chr.9: 113354346-113354346 on Build GRCh38
3320095 c.218A>T p.Q73L Chr.9: 113354256-113354256 on Build GRCh38
4834809 c.202-1G>A p.? Chr.9: 113354239-113354239 on Build GRCh38
3375038 c.208A>G p.I70V Chr.9: 113354246-113354246 on Build GRCh38
3952234 c.202-2A>G p.? Chr.9: 113354238-113354238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992095827 Chr.9: 113354395-113354395 on Build GRCh38
rs553041594 Chr.9: 113354188-113354188 on Build GRCh38
rs779107286 Chr.9: 113354214-113354214 on Build GRCh38
rs557164626 Chr.9: 113354308-113354308 on Build GRCh38
rs970141638 Chr.9: 113354340-113354340 on Build GRCh38
rs10981762 Chr.9: 113354203-113354203 on Build GRCh38
rs774157751 Chr.9: 113354291-113354291 on Build GRCh38
rs748770893 Chr.9: 113354358-113354358 on Build GRCh38
rs774139609 Chr.9: 113354374-113354374 on Build GRCh38
rs374063532 Chr.9: 113354372-113354372 on Build GRCh38
rs764636207 Chr.9: 113354204-113354204 on Build GRCh38
rs368306479 Chr.9: 113354316-113354316 on Build GRCh38
rs772730098 Chr.9: 113354192-113354192 on Build GRCh38
rs762404330 Chr.9: 113354288-113354288 on Build GRCh38
rs139872649 Chr.9: 113354154-113354155 on Build GRCh38
rs573475250 Chr.9: 113354347-113354347 on Build GRCh38
rs758686100 Chr.9: 113354230-113354230 on Build GRCh38
rs940623912 Chr.9: 113354414-113354414 on Build GRCh38
rs10981763 Chr.9: 113354346-113354346 on Build GRCh38
rs772823188 Chr.9: 113354285-113354285 on Build GRCh38
rs764616023 Chr.9: 113354303-113354303 on Build GRCh38
rs201512287 Chr.9: 113354292-113354292 on Build GRCh38
rs1037511315 Chr.9: 113354159-113354159 on Build GRCh38
rs939314621 Chr.9: 113354158-113354158 on Build GRCh38
rs907787371 Chr.9: 113354156-113354156 on Build GRCh38
rs202215549 Chr.9: 113354243-113354243 on Build GRCh38
rs377571038 Chr.9: 113354270-113354270 on Build GRCh38
rs753561751 Chr.9: 113354331-113354331 on Build GRCh38
rs769168556 Chr.9: 113354283-113354283 on Build GRCh38
rs780284789 Chr.9: 113354236-113354236 on Build GRCh38
rs373291577 Chr.9: 113354305-113354305 on Build GRCh38
rs762490754 Chr.9: 113354309-113354309 on Build GRCh38
rs565262878 Chr.9: 113354438-113354438 on Build GRCh38
rs545120786 Chr.9: 113354425-113354425 on Build GRCh38
rs759317375 Chr.9: 113354384-113354384 on Build GRCh38
rs189648342 Chr.9: 113354194-113354194 on Build GRCh38
rs919208300 Chr.9: 113354404-113354404 on Build GRCh38
rs780185557 Chr.9: 113354317-113354317 on Build GRCh38
rs28607343 Chr.9: 113354145-113354145 on Build GRCh38
rs757276879 Chr.9: 113354212-113354212 on Build GRCh38
rs926517449 Chr.9: 113354154-113354154 on Build GRCh38
rs929004561 Chr.9: 113354220-113354220 on Build GRCh38
rs761448076 Chr.9: 113354115-113354115 on Build GRCh38
rs887563192 Chr.9: 113354240-113354240 on Build GRCh38
rs372538396 Chr.9: 113354224-113354224 on Build GRCh38
rs891806843 Chr.9: 113354311-113354311 on Build GRCh38
rs943145601 Chr.9: 113354444-113354444 on Build GRCh38
rs974382061 Chr.9: 113354135-113354135 on Build GRCh38
rs1011567154 Chr.9: 113354312-113354312 on Build GRCh38
rs768445660 Chr.9: 113354253-113354253 on Build GRCh38
rs987551579 Chr.9: 113354360-113354360 on Build GRCh38
rs984477297 Chr.9: 113354430-113354430 on Build GRCh38
rs967847190 Chr.9: 113354120-113354120 on Build GRCh38
rs182334591 Chr.9: 113354345-113354345 on Build GRCh38
rs753073711 Chr.9: 113354202-113354202 on Build GRCh38
rs907757866 Chr.9: 113354393-113354393 on Build GRCh38
rs575842489 Chr.9: 113354442-113354442 on Build GRCh38
rs767965451 Chr.9: 113354201-113354201 on Build GRCh38
rs747299077 Chr.9: 113354319-113354319 on Build GRCh38
rs766065412 Chr.9: 113354224-113354226 on Build GRCh38
rs754370322 Chr.9: 113354211-113354211 on Build GRCh38
rs754285338 Chr.9: 113354304-113354304 on Build GRCh38
rs776546213 Chr.9: 113354258-113354258 on Build GRCh38
rs754754848 Chr.9: 113354343-113354343 on Build GRCh38

View Full Product Details