Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39204765-39205253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR19
Entrez Gene ID
57728
Gene Name
WD repeat domain 19
Gene Aliases
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
UniGene
Hs.438482
Gene Chromosome Location
Chr.4: 39182404-39285810 on Build GRCh38
Transcripts
NM_001317924.1 NM_025132.3 XM_011513724.1 XM_011513725.2 XM_011513726.2 XM_011513727.1 XM_017008501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1429603 c.633_634insT p.L214fs*5 Chr.4: 39205183-39205184 on Build GRCh38
6469544 c.624G>A p.K208K Chr.4: 39205174-39205174 on Build GRCh38
6390256 c.690C>A p.D230E Chr.4: 39205240-39205240 on Build GRCh38
1664986 c.703G>A p.V235I Chr.4: 39205253-39205253 on Build GRCh38
481241 c.637T>A p.F213I Chr.4: 39205187-39205187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777279563 Chr.4: 39204913-39204913 on Build GRCh38
rs981829389 Chr.4: 39205089-39205089 on Build GRCh38
rs78053439 Chr.4: 39205080-39205080 on Build GRCh38
rs760702815 Chr.4: 39205110-39205110 on Build GRCh38
rs766591103 Chr.4: 39205144-39205144 on Build GRCh38
rs1007346114 Chr.4: 39204864-39204864 on Build GRCh38
rs1025484046 Chr.4: 39204951-39204951 on Build GRCh38
rs148423098 Chr.4: 39204972-39204972 on Build GRCh38
rs190472192 Chr.4: 39204803-39204803 on Build GRCh38
rs979320154 Chr.4: 39205032-39205032 on Build GRCh38
rs1046047581 Chr.4: 39204788-39204788 on Build GRCh38
rs764016456 Chr.4: 39205162-39205162 on Build GRCh38
rs913477341 Chr.4: 39205178-39205178 on Build GRCh38
rs1018991834 Chr.4: 39204873-39204873 on Build GRCh38
rs937414802 Chr.4: 39205081-39205081 on Build GRCh38
rs889873251 Chr.4: 39204892-39204892 on Build GRCh38
rs79399994 Chr.4: 39204802-39204802 on Build GRCh38
rs1027514837 Chr.4: 39204854-39204854 on Build GRCh38
rs965328733 Chr.4: 39204945-39204945 on Build GRCh38
rs1011711308 Chr.4: 39204878-39204878 on Build GRCh38
rs375847794 Chr.4: 39204955-39204955 on Build GRCh38
rs758413433 Chr.4: 39205163-39205163 on Build GRCh38
rs917207293 Chr.4: 39205095-39205095 on Build GRCh38
rs192592383 Chr.4: 39205094-39205094 on Build GRCh38
rs1007407250 Chr.4: 39204895-39204895 on Build GRCh38
rs953247647 Chr.4: 39204859-39204859 on Build GRCh38
rs1043650529 Chr.4: 39204889-39204889 on Build GRCh38
rs747438345 Chr.4: 39205149-39205150 on Build GRCh38
rs892054763 Chr.4: 39204838-39204838 on Build GRCh38
rs1470774 Chr.4: 39205137-39205137 on Build GRCh38
rs755179058 Chr.4: 39205159-39205159 on Build GRCh38
rs550185984 Chr.4: 39205091-39205091 on Build GRCh38
rs994133144 Chr.4: 39204949-39204949 on Build GRCh38
rs753999931 Chr.4: 39205150-39205150 on Build GRCh38
rs757241775 Chr.4: 39205214-39205214 on Build GRCh38
rs587777348 Chr.4: 39205191-39205192 on Build GRCh38
rs751290509 Chr.4: 39205191-39205191 on Build GRCh38
rs573386296 Chr.4: 39204961-39204961 on Build GRCh38
rs866346887 Chr.4: 39205072-39205072 on Build GRCh38
rs770303764 Chr.4: 39205016-39205016 on Build GRCh38
rs562263550 Chr.4: 39205069-39205069 on Build GRCh38
rs987812073 Chr.4: 39205135-39205135 on Build GRCh38
rs774983663 Chr.4: 39205218-39205218 on Build GRCh38
rs927643512 Chr.4: 39204791-39204791 on Build GRCh38
rs967650431 Chr.4: 39205004-39205004 on Build GRCh38

View Full Product Details