Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174309694-174310231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
UniGene
Hs.555989
Gene Chromosome Location
Chr.4: 174283677-174334355 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6430576 c.686A>G p.N229S Chr.4: 174309857-174309857 on Build GRCh38
168679 c.758C>T p.S253L Chr.4: 174309929-174309929 on Build GRCh38
6430577 c.686A>G p.N229S Chr.4: 174309857-174309857 on Build GRCh38
5834796 c.863C>T p.T288I Chr.4: 174310034-174310034 on Build GRCh38
6398001 c.787C>A p.Q263K Chr.4: 174309958-174309958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920079618 Chr.4: 174310187-174310187 on Build GRCh38
rs77511056 Chr.4: 174309959-174309959 on Build GRCh38
rs747457036 Chr.4: 174309924-174309924 on Build GRCh38
rs554691137 Chr.4: 174309744-174309744 on Build GRCh38
rs144338962 Chr.4: 174310024-174310024 on Build GRCh38
rs753999371 Chr.4: 174309883-174309883 on Build GRCh38
rs1009523898 Chr.4: 174310067-174310067 on Build GRCh38
rs754326890 Chr.4: 174309899-174309899 on Build GRCh38
rs767580733 Chr.4: 174309998-174309998 on Build GRCh38
rs367613908 Chr.4: 174309957-174309957 on Build GRCh38
rs968791692 Chr.4: 174310121-174310121 on Build GRCh38
rs751584812 Chr.4: 174309866-174309866 on Build GRCh38
rs947402476 Chr.4: 174309749-174309749 on Build GRCh38
rs374542523 Chr.4: 174309942-174309942 on Build GRCh38
rs762020752 Chr.4: 174310013-174310013 on Build GRCh38
rs973016196 Chr.4: 174310175-174310175 on Build GRCh38
rs116383874 Chr.4: 174310206-174310206 on Build GRCh38
rs758862327 Chr.4: 174309922-174309922 on Build GRCh38
rs760828480 Chr.4: 174309879-174309879 on Build GRCh38
rs779217465 Chr.4: 174310005-174310007 on Build GRCh38
rs777280933 Chr.4: 174310077-174310077 on Build GRCh38
rs932563341 Chr.4: 174309834-174309834 on Build GRCh38
rs921250491 Chr.4: 174309770-174309770 on Build GRCh38
rs968100458 Chr.4: 174309757-174309757 on Build GRCh38
rs74954653 Chr.4: 174310100-174310100 on Build GRCh38
rs371383514 Chr.4: 174309901-174309901 on Build GRCh38
rs761389445 Chr.4: 174309832-174309832 on Build GRCh38
rs1002116541 Chr.4: 174309724-174309724 on Build GRCh38
rs200591484 Chr.4: 174309851-174309851 on Build GRCh38
rs1036810416 Chr.4: 174309897-174309897 on Build GRCh38
rs371274791 Chr.4: 174309962-174309962 on Build GRCh38
rs1035374894 Chr.4: 174309734-174309734 on Build GRCh38
rs772102409 Chr.4: 174310105-174310105 on Build GRCh38
rs890978233 Chr.4: 174309853-174309853 on Build GRCh38
rs769517541 Chr.4: 174309842-174309842 on Build GRCh38
rs770820786 Chr.4: 174309970-174309970 on Build GRCh38
rs561778304 Chr.4: 174310143-174310143 on Build GRCh38
rs57747776 Chr.4: 174309769-174309769 on Build GRCh38
rs368621470 Chr.4: 174310173-174310173 on Build GRCh38
rs759503328 Chr.4: 174309984-174309984 on Build GRCh38
rs17060455 Chr.4: 174309745-174309745 on Build GRCh38
rs780410407 Chr.4: 174309923-174309923 on Build GRCh38
rs774236010 Chr.4: 174310104-174310104 on Build GRCh38
rs926365818 Chr.4: 174310208-174310208 on Build GRCh38
rs1028382314 Chr.4: 174310124-174310124 on Build GRCh38
rs780963842 Chr.4: 174310015-174310015 on Build GRCh38
rs912819762 Chr.4: 174309880-174309880 on Build GRCh38
rs758740148 Chr.4: 174309987-174309987 on Build GRCh38
rs753944047 Chr.4: 174310009-174310009 on Build GRCh38
rs764215452 Chr.4: 174309882-174309882 on Build GRCh38
rs541941816 Chr.4: 174310129-174310129 on Build GRCh38
rs148765177 Chr.4: 174310103-174310104 on Build GRCh38
rs931950534 Chr.4: 174310163-174310163 on Build GRCh38
rs904843542 Chr.4: 174310004-174310004 on Build GRCh38
rs757502117 Chr.4: 174309889-174309889 on Build GRCh38
rs766198712 Chr.4: 174309857-174309857 on Build GRCh38
rs772508184 Chr.4: 174310104-174310104 on Build GRCh38
rs954042475 Chr.4: 174310142-174310142 on Build GRCh38
rs750788616 Chr.4: 174309939-174309939 on Build GRCh38
rs747681903 Chr.4: 174309950-174309950 on Build GRCh38
rs202230528 Chr.4: 174309952-174309952 on Build GRCh38
rs755430063 Chr.4: 174310046-174310046 on Build GRCh38
rs751952559 Chr.4: 174310033-174310033 on Build GRCh38
rs780764568 Chr.4: 174309843-174309843 on Build GRCh38
rs766509482 Chr.4: 174310166-174310166 on Build GRCh38
rs928833265 Chr.4: 174309723-174309723 on Build GRCh38
rs781701907 Chr.4: 174310064-174310064 on Build GRCh38
rs777364140 Chr.4: 174310151-174310151 on Build GRCh38
rs79548708 Chr.4: 174310196-174310196 on Build GRCh38
rs557197307 Chr.4: 174309981-174309981 on Build GRCh38
rs3215171 Chr.4: 174310112-174310113 on Build GRCh38
rs769157129 Chr.4: 174309929-174309929 on Build GRCh38
rs572788167 Chr.4: 174310113-174310113 on Build GRCh38
rs757823438 Chr.4: 174309907-174309908 on Build GRCh38
rs530691766 Chr.4: 174310155-174310155 on Build GRCh38
rs755720331 Chr.4: 174310075-174310075 on Build GRCh38
rs762857468 Chr.4: 174309852-174309852 on Build GRCh38
rs149603762 Chr.4: 174309894-174309894 on Build GRCh38
rs750713311 Chr.4: 174310016-174310016 on Build GRCh38
rs904895605 Chr.4: 174309719-174309719 on Build GRCh38
rs149601321 Chr.4: 174309735-174309735 on Build GRCh38
rs546205563 Chr.4: 174310074-174310074 on Build GRCh38
rs758562577 Chr.4: 174310017-174310017 on Build GRCh38
rs377618784 Chr.4: 174310082-174310082 on Build GRCh38

View Full Product Details