Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28769330-28769859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF2
Entrez Gene ID
51441
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Gene Aliases
CAHL, HGRG8, NY-REN-2
UniGene
Hs.532286
Gene Chromosome Location
Chr.1: 28736621-28769775 on Build GRCh38
Transcripts
NM_001172828.1 NM_001173128.1 NM_016258.2

SNP Detail

SNP ID Chromosome Location
rs895777362 Chr.1: 28769738-28769738 on Build GRCh38
rs866258710 Chr.1: 28769378-28769378 on Build GRCh38
rs983514301 Chr.1: 28769581-28769581 on Build GRCh38
rs765667566 Chr.1: 28769711-28769711 on Build GRCh38
rs566988910 Chr.1: 28769413-28769413 on Build GRCh38
rs951168885 Chr.1: 28769757-28769757 on Build GRCh38
rs746057489 Chr.1: 28769754-28769754 on Build GRCh38
rs756700868 Chr.1: 28769517-28769522 on Build GRCh38
rs574371397 Chr.1: 28769650-28769650 on Build GRCh38
rs180728450 Chr.1: 28769438-28769438 on Build GRCh38
rs933570794 Chr.1: 28769656-28769656 on Build GRCh38
rs904200471 Chr.1: 28769678-28769678 on Build GRCh38
rs891932324 Chr.1: 28769667-28769667 on Build GRCh38
rs1014174143 Chr.1: 28769745-28769745 on Build GRCh38
rs1051862122 Chr.1: 28769380-28769380 on Build GRCh38
rs968375341 Chr.1: 28769416-28769416 on Build GRCh38
rs945790154 Chr.1: 28769671-28769671 on Build GRCh38
rs189369572 Chr.1: 28769604-28769604 on Build GRCh38
rs973205142 Chr.1: 28769795-28769798 on Build GRCh38
rs1007008308 Chr.1: 28769714-28769714 on Build GRCh38
rs772280384 Chr.1: 28769521-28769525 on Build GRCh38
rs1034242734 Chr.1: 28769449-28769449 on Build GRCh38
rs950654001 Chr.1: 28769808-28769808 on Build GRCh38
rs985559081 Chr.1: 28769394-28769394 on Build GRCh38
rs929002495 Chr.1: 28769466-28769466 on Build GRCh38
rs919152648 Chr.1: 28769466-28769466 on Build GRCh38
rs374032666 Chr.1: 28769703-28769703 on Build GRCh38
rs1025483017 Chr.1: 28769746-28769746 on Build GRCh38
rs182370004 Chr.1: 28769795-28769795 on Build GRCh38
rs1042788960 Chr.1: 28769672-28769672 on Build GRCh38
rs371452625 Chr.1: 28769651-28769651 on Build GRCh38
rs918422961 Chr.1: 28769545-28769545 on Build GRCh38
rs950813705 Chr.1: 28769550-28769550 on Build GRCh38
rs184502359 Chr.1: 28769561-28769561 on Build GRCh38
rs539227456 Chr.1: 28769425-28769425 on Build GRCh38
rs946861504 Chr.1: 28769568-28769568 on Build GRCh38
rs1042629414 Chr.1: 28769589-28769589 on Build GRCh38
rs538546863 Chr.1: 28769595-28769595 on Build GRCh38
rs942010805 Chr.1: 28769625-28769625 on Build GRCh38
rs992686467 Chr.1: 28769461-28769461 on Build GRCh38
rs1009980472 Chr.1: 28769412-28769412 on Build GRCh38
rs533811724 Chr.1: 28769659-28769659 on Build GRCh38
rs891584628 Chr.1: 28769400-28769400 on Build GRCh38
rs527591643 Chr.1: 28769782-28769782 on Build GRCh38
rs979245356 Chr.1: 28769826-28769827 on Build GRCh38
rs774455849 Chr.1: 28769666-28769666 on Build GRCh38
rs959934550 Chr.1: 28769450-28769450 on Build GRCh38
rs998197720 Chr.1: 28769632-28769632 on Build GRCh38
rs35926776 Chr.1: 28769759-28769760 on Build GRCh38
rs975134788 Chr.1: 28769636-28769636 on Build GRCh38
rs1001239354 Chr.1: 28769698-28769698 on Build GRCh38
rs1016376789 Chr.1: 28769828-28769828 on Build GRCh38
rs982152840 Chr.1: 28769512-28769512 on Build GRCh38
rs775677324 Chr.1: 28769824-28769824 on Build GRCh38
rs922211111 Chr.1: 28769640-28769640 on Build GRCh38

View Full Product Details