Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168280704-168281297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX19
Entrez Gene ID
9095
Gene Name
T-box 19
Gene Aliases
TBS19, TPIT, dJ747L4.1
UniGene
Hs.716656
Gene Chromosome Location
Chr.1: 168281040-168314426 on Build GRCh38
Transcripts
NM_005149.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5643092 c.203+1G>A p.? Chr.1: 168281294-168281294 on Build GRCh38
1228680 c.175A>G p.N59D Chr.1: 168281265-168281265 on Build GRCh38
368439 c.189G>T p.V63V Chr.1: 168281279-168281279 on Build GRCh38
7135035 c.92G>C p.R31T Chr.1: 168281182-168281182 on Build GRCh38
4732522 c.95delA p.G34fs*28 Chr.1: 168281185-168281185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181723245 Chr.1: 168280741-168280741 on Build GRCh38
rs886045508 Chr.1: 168281129-168281129 on Build GRCh38
rs746455583 Chr.1: 168281072-168281072 on Build GRCh38
rs751238563 Chr.1: 168281244-168281244 on Build GRCh38
rs780760574 Chr.1: 168281259-168281259 on Build GRCh38
rs528606805 Chr.1: 168281168-168281168 on Build GRCh38
rs202179787 Chr.1: 168281172-168281172 on Build GRCh38
rs771279737 Chr.1: 168281180-168281180 on Build GRCh38
rs1051313746 Chr.1: 168280792-168280792 on Build GRCh38
rs1017377053 Chr.1: 168281223-168281223 on Build GRCh38
rs766630651 Chr.1: 168280836-168280836 on Build GRCh38
rs753791024 Chr.1: 168281145-168281145 on Build GRCh38
rs771353874 Chr.1: 168281097-168281097 on Build GRCh38
rs773760897 Chr.1: 168281085-168281085 on Build GRCh38
rs772489505 Chr.1: 168281193-168281193 on Build GRCh38
rs764256599 Chr.1: 168281200-168281200 on Build GRCh38
rs138336580 Chr.1: 168281236-168281236 on Build GRCh38
rs1017241094 Chr.1: 168280879-168280879 on Build GRCh38
rs767791473 Chr.1: 168281116-168281116 on Build GRCh38
rs144173772 Chr.1: 168281126-168281126 on Build GRCh38
rs994950334 Chr.1: 168280891-168280891 on Build GRCh38
rs760885658 Chr.1: 168281197-168281197 on Build GRCh38
rs143086625 Chr.1: 168280981-168280981 on Build GRCh38
rs1027280708 Chr.1: 168280951-168280951 on Build GRCh38
rs999458323 Chr.1: 168280838-168280838 on Build GRCh38
rs923207588 Chr.1: 168280919-168280919 on Build GRCh38
rs894490454 Chr.1: 168280761-168280761 on Build GRCh38
rs534144943 Chr.1: 168280887-168280887 on Build GRCh38
rs750287749 Chr.1: 168281235-168281235 on Build GRCh38
rs146259816 Chr.1: 168281115-168281115 on Build GRCh38
rs535442563 Chr.1: 168281157-168281157 on Build GRCh38
rs891300633 Chr.1: 168280872-168280872 on Build GRCh38
rs753819978 Chr.1: 168281205-168281205 on Build GRCh38
rs935571603 Chr.1: 168280787-168280787 on Build GRCh38
rs760520604 Chr.1: 168281162-168281165 on Build GRCh38
rs1013083829 Chr.1: 168280795-168280795 on Build GRCh38
rs187423054 Chr.1: 168281070-168281070 on Build GRCh38
rs55867415 Chr.1: 168280980-168280980 on Build GRCh38
rs372107667 Chr.1: 168281151-168281151 on Build GRCh38
rs149094298 Chr.1: 168281152-168281152 on Build GRCh38
rs950558668 Chr.1: 168280948-168280948 on Build GRCh38
rs563196275 Chr.1: 168280881-168280881 on Build GRCh38
rs761813219 Chr.1: 168281222-168281222 on Build GRCh38
rs758056532 Chr.1: 168281165-168281165 on Build GRCh38
rs552045315 Chr.1: 168280936-168280936 on Build GRCh38
rs774542130 Chr.1: 168281108-168281108 on Build GRCh38
rs749105937 Chr.1: 168281050-168281050 on Build GRCh38
rs567056722 Chr.1: 168281023-168281023 on Build GRCh38
rs779718152 Chr.1: 168281239-168281239 on Build GRCh38
rs774982649 Chr.1: 168281071-168281071 on Build GRCh38
rs776025094 Chr.1: 168281062-168281062 on Build GRCh38
rs770339238 Chr.1: 168281079-168281079 on Build GRCh38
rs760832699 Chr.1: 168281127-168281127 on Build GRCh38
rs999025307 Chr.1: 168281143-168281143 on Build GRCh38
rs544236100 Chr.1: 168280878-168280878 on Build GRCh38
rs754589236 Chr.1: 168281246-168281246 on Build GRCh38
rs34284181 Chr.1: 168281195-168281195 on Build GRCh38
rs367613004 Chr.1: 168281088-168281088 on Build GRCh38
rs1045982044 Chr.1: 168280807-168280807 on Build GRCh38
rs778479278 Chr.1: 168281054-168281054 on Build GRCh38
rs200517364 Chr.1: 168281069-168281069 on Build GRCh38
rs907341018 Chr.1: 168280818-168280818 on Build GRCh38
rs745555504 Chr.1: 168281058-168281058 on Build GRCh38
rs759095375 Chr.1: 168281170-168281170 on Build GRCh38
rs960844295 Chr.1: 168281061-168281061 on Build GRCh38
rs1043736118 Chr.1: 168280733-168280733 on Build GRCh38
rs768136578 Chr.1: 168281177-168281177 on Build GRCh38
rs774793160 Chr.1: 168281183-168281183 on Build GRCh38
rs764168276 Chr.1: 168281131-168281131 on Build GRCh38
rs757045023 Chr.1: 168281053-168281053 on Build GRCh38
rs765023781 Chr.1: 168281229-168281229 on Build GRCh38
rs190633524 Chr.1: 168281100-168281100 on Build GRCh38
rs763818059 Chr.1: 168281245-168281246 on Build GRCh38
rs1038420569 Chr.1: 168281046-168281046 on Build GRCh38
rs759967925 Chr.1: 168281192-168281192 on Build GRCh38
rs1031417698 Chr.1: 168280873-168280873 on Build GRCh38
rs780523079 Chr.1: 168281179-168281179 on Build GRCh38
rs866428753 Chr.1: 168281238-168281238 on Build GRCh38

View Full Product Details