Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50081047-50081644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM189-UBE2V1
Entrez Gene ID
387522
Gene Name
TMEM189-UBE2V1 readthrough
Gene Aliases
CROC-1B, CROC1B, KUA-UEV
UniGene
Hs.744839
Gene Chromosome Location
Chr.20: 50081124-50153798 on Build GRCh38
Transcripts
NM_199203.2 NM_001032288.2 NM_001257393.1 NM_001257394.1 NM_001257395.1 NM_001257396.1 NM_001257397.1 NM_001257398.1 NM_001257399.1 NM_001282575.2 NM_001282576.1 NM_001282577.2 NM_001282578.2 NM_001282579.1 NM_021988.5 NM_022442.5 NM_199144.2 NR_047554.1

Gene Symbol
UBE2V1
Entrez Gene ID
7335
Gene Name
ubiquitin conjugating enzyme E2 V1
Gene Aliases
CIR1, CROC-1, CROC1, UBE2V, UEV-1, UEV1, UEV1A
UniGene
Hs.420529
Gene Chromosome Location
Chr.20: 50081124-50115959 on Build GRCh38
Transcripts
NM_199203.2 NM_001032288.2 NM_001257393.1 NM_001257394.1 NM_001257395.1 NM_001257396.1 NM_001257397.1 NM_001257398.1 NM_001257399.1 NM_001282575.2 NM_001282576.1 NM_001282577.2 NM_001282578.2 NM_001282579.1 NM_021988.5 NM_022442.5 NM_199144.2 NR_047554.1

SNP Detail

SNP ID Chromosome Location
rs2733 Chr.20: 50081535-50081535 on Build GRCh38
rs368384878 Chr.20: 50081460-50081462 on Build GRCh38
rs754458946 Chr.20: 50081123-50081123 on Build GRCh38
rs878876092 Chr.20: 50081347-50081347 on Build GRCh38
rs201015788 Chr.20: 50081136-50081136 on Build GRCh38
rs752330982 Chr.20: 50081341-50081342 on Build GRCh38
rs1049871 Chr.20: 50081254-50081254 on Build GRCh38
rs116141128 Chr.20: 50081113-50081113 on Build GRCh38
rs942949266 Chr.20: 50081445-50081445 on Build GRCh38
rs765686922 Chr.20: 50081502-50081502 on Build GRCh38
rs1011001131 Chr.20: 50081503-50081503 on Build GRCh38
rs989456149 Chr.20: 50081152-50081152 on Build GRCh38
rs74435910 Chr.20: 50081360-50081362 on Build GRCh38
rs553923039 Chr.20: 50081076-50081076 on Build GRCh38
rs980407866 Chr.20: 50081548-50081549 on Build GRCh38
rs1007098894 Chr.20: 50081071-50081071 on Build GRCh38
rs969926367 Chr.20: 50081174-50081174 on Build GRCh38
rs535813598 Chr.20: 50081617-50081617 on Build GRCh38
rs931083930 Chr.20: 50081356-50081357 on Build GRCh38
rs1048011735 Chr.20: 50081357-50081357 on Build GRCh38
rs567922169 Chr.20: 50081155-50081155 on Build GRCh38
rs938551200 Chr.20: 50081308-50081308 on Build GRCh38
rs568498428 Chr.20: 50081571-50081571 on Build GRCh38
rs15218 Chr.20: 50081486-50081486 on Build GRCh38
rs1044975480 Chr.20: 50081135-50081135 on Build GRCh38
rs201099481 Chr.20: 50081347-50081347 on Build GRCh38
rs1031603733 Chr.20: 50081501-50081501 on Build GRCh38
rs889457438 Chr.20: 50081393-50081393 on Build GRCh38
rs956697022 Chr.20: 50081097-50081097 on Build GRCh38
rs2664563 Chr.20: 50081324-50081324 on Build GRCh38
rs374324590 Chr.20: 50081431-50081431 on Build GRCh38
rs556570713 Chr.20: 50081563-50081563 on Build GRCh38
rs901321066 Chr.20: 50081475-50081475 on Build GRCh38
rs10567504 Chr.20: 50081142-50081145 on Build GRCh38
rs115164526 Chr.20: 50081483-50081483 on Build GRCh38
rs762056905 Chr.20: 50081451-50081451 on Build GRCh38
rs960241899 Chr.20: 50081604-50081604 on Build GRCh38
rs73611751 Chr.20: 50081473-50081473 on Build GRCh38
rs972879830 Chr.20: 50081344-50081345 on Build GRCh38
rs2664532 Chr.20: 50081244-50081244 on Build GRCh38
rs535738819 Chr.20: 50081524-50081524 on Build GRCh38
rs777176288 Chr.20: 50081446-50081446 on Build GRCh38
rs35338409 Chr.20: 50081362-50081363 on Build GRCh38
rs775575773 Chr.20: 50081129-50081129 on Build GRCh38
rs566510389 Chr.20: 50081347-50081348 on Build GRCh38
rs987765722 Chr.20: 50081108-50081108 on Build GRCh38
rs980050190 Chr.20: 50081199-50081199 on Build GRCh38
rs186621934 Chr.20: 50081455-50081455 on Build GRCh38
rs761101085 Chr.20: 50081178-50081178 on Build GRCh38

View Full Product Details