Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43528031-43528630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
UniGene
Hs.434059
Gene Chromosome Location
Chr.17: 43527843-43546432 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
94256 c.1358C>G p.T453R Chr.17: 43528616-43528616 on Build GRCh38
3190340 c.1439G>A p.G480D Chr.17: 43528535-43528535 on Build GRCh38
6656745 c.1390G>A p.A464T Chr.17: 43528584-43528584 on Build GRCh38
1383558 c.1351G>A p.E451K Chr.17: 43528623-43528623 on Build GRCh38
5991265 c.1423C>G p.P475A Chr.17: 43528551-43528551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997279278 Chr.17: 43528310-43528310 on Build GRCh38
rs892227491 Chr.17: 43528275-43528275 on Build GRCh38
rs770702451 Chr.17: 43528587-43528587 on Build GRCh38
rs143919906 Chr.17: 43528065-43528065 on Build GRCh38
rs778891947 Chr.17: 43528500-43528500 on Build GRCh38
rs749607350 Chr.17: 43528592-43528592 on Build GRCh38
rs538818050 Chr.17: 43528351-43528351 on Build GRCh38
rs749134020 Chr.17: 43528495-43528495 on Build GRCh38
rs560866512 Chr.17: 43528358-43528358 on Build GRCh38
rs747487116 Chr.17: 43528114-43528114 on Build GRCh38
rs896774071 Chr.17: 43528421-43528421 on Build GRCh38
rs1045769531 Chr.17: 43528548-43528548 on Build GRCh38
rs562857882 Chr.17: 43528086-43528086 on Build GRCh38
rs933736272 Chr.17: 43528082-43528082 on Build GRCh38
rs948309876 Chr.17: 43528230-43528230 on Build GRCh38
rs561366431 Chr.17: 43528581-43528581 on Build GRCh38
rs748064563 Chr.17: 43528583-43528583 on Build GRCh38
rs764630172 Chr.17: 43528527-43528527 on Build GRCh38
rs538570019 Chr.17: 43528511-43528511 on Build GRCh38
rs978800312 Chr.17: 43528251-43528251 on Build GRCh38
rs775984025 Chr.17: 43528521-43528521 on Build GRCh38
rs769166723 Chr.17: 43528594-43528594 on Build GRCh38
rs531425765 Chr.17: 43528391-43528391 on Build GRCh38
rs967859380 Chr.17: 43528150-43528150 on Build GRCh38
rs776427258 Chr.17: 43528591-43528591 on Build GRCh38
rs761905265 Chr.17: 43528547-43528547 on Build GRCh38
rs781663777 Chr.17: 43528178-43528178 on Build GRCh38
rs948325065 Chr.17: 43528531-43528531 on Build GRCh38
rs1038056008 Chr.17: 43528405-43528405 on Build GRCh38
rs757289344 Chr.17: 43528471-43528471 on Build GRCh38
rs375332940 Chr.17: 43528569-43528569 on Build GRCh38
rs1018271907 Chr.17: 43528453-43528453 on Build GRCh38
rs992289835 Chr.17: 43528524-43528524 on Build GRCh38
rs746585925 Chr.17: 43528586-43528586 on Build GRCh38
rs867116796 Chr.17: 43528182-43528182 on Build GRCh38
rs183855313 Chr.17: 43528247-43528247 on Build GRCh38
rs180992684 Chr.17: 43528239-43528239 on Build GRCh38
rs140975276 Chr.17: 43528558-43528558 on Build GRCh38
rs1012023602 Chr.17: 43528470-43528470 on Build GRCh38
rs754900082 Chr.17: 43528498-43528498 on Build GRCh38
rs766650217 Chr.17: 43528565-43528565 on Build GRCh38
rs911221357 Chr.17: 43528113-43528113 on Build GRCh38
rs941077799 Chr.17: 43528401-43528401 on Build GRCh38
rs545693403 Chr.17: 43528142-43528142 on Build GRCh38
rs746231923 Chr.17: 43528494-43528494 on Build GRCh38
rs570890035 Chr.17: 43528510-43528510 on Build GRCh38
rs918089102 Chr.17: 43528526-43528526 on Build GRCh38
rs774331707 Chr.17: 43528196-43528196 on Build GRCh38
rs953288044 Chr.17: 43528386-43528386 on Build GRCh38
rs1034900105 Chr.17: 43528429-43528429 on Build GRCh38
rs763477409 Chr.17: 43528522-43528522 on Build GRCh38
rs985438177 Chr.17: 43528101-43528101 on Build GRCh38
rs578258023 Chr.17: 43528115-43528115 on Build GRCh38
rs994082197 Chr.17: 43528428-43528428 on Build GRCh38
rs934143979 Chr.17: 43528359-43528359 on Build GRCh38
rs892174986 Chr.17: 43528606-43528606 on Build GRCh38
rs1005198630 Chr.17: 43528110-43528110 on Build GRCh38
rs549401602 Chr.17: 43528504-43528504 on Build GRCh38
rs960522762 Chr.17: 43528191-43528191 on Build GRCh38
rs533283267 Chr.17: 43528170-43528170 on Build GRCh38
rs934932302 Chr.17: 43528309-43528309 on Build GRCh38
rs573984220 Chr.17: 43528062-43528062 on Build GRCh38
rs1034816643 Chr.17: 43528179-43528179 on Build GRCh38
rs992914901 Chr.17: 43528238-43528239 on Build GRCh38
rs755525910 Chr.17: 43528474-43528487 on Build GRCh38
rs762197744 Chr.17: 43528604-43528604 on Build GRCh38
rs549071785 Chr.17: 43528264-43528264 on Build GRCh38
rs889528792 Chr.17: 43528398-43528399 on Build GRCh38
rs771214543 Chr.17: 43528134-43528134 on Build GRCh38
rs896348937 Chr.17: 43528465-43528465 on Build GRCh38
rs774883014 Chr.17: 43528600-43528600 on Build GRCh38
rs766424584 Chr.17: 43528096-43528096 on Build GRCh38
rs771500754 Chr.17: 43528505-43528505 on Build GRCh38
rs896712813 Chr.17: 43528222-43528222 on Build GRCh38
rs775012630 Chr.17: 43528532-43528532 on Build GRCh38
rs755899580 Chr.17: 43528485-43528485 on Build GRCh38
rs750558611 Chr.17: 43528557-43528557 on Build GRCh38
rs754777184 Chr.17: 43528580-43528580 on Build GRCh38
rs953434638 Chr.17: 43528083-43528083 on Build GRCh38
rs750635016 Chr.17: 43528477-43528477 on Build GRCh38
rs777123700 Chr.17: 43528585-43528585 on Build GRCh38
rs975957389 Chr.17: 43528063-43528063 on Build GRCh38
rs781418149 Chr.17: 43528476-43528476 on Build GRCh38
rs767526603 Chr.17: 43528549-43528549 on Build GRCh38
rs796159098 Chr.17: 43528246-43528246 on Build GRCh38
rs772878183 Chr.17: 43528506-43528506 on Build GRCh38
rs758397885 Chr.17: 43528584-43528584 on Build GRCh38
rs986486865 Chr.17: 43528441-43528441 on Build GRCh38

View Full Product Details