Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 617685-618079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUL1
Entrez Gene ID
27098
Gene Name
clusterin like 1
Gene Aliases
RA337M
UniGene
Hs.732341
Gene Chromosome Location
Chr.18: 596998-650298 on Build GRCh38
Transcripts
NM_001289036.1 NM_001318522.1 NM_014410.4 NM_199167.1 XM_005258103.3 XM_006722322.2 XM_011525648.2 XM_011525649.1 XM_011525651.2 XM_011525652.1 XM_011525653.2 XM_017025707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854381 c.42G>A p.W14* Chr.18: 618042-618042 on Build GRCh38
3144759 c.4A>G p.K2E Chr.18: 618004-618004 on Build GRCh38
279946 c.37C>T p.L13L Chr.18: 618037-618037 on Build GRCh38
438320 c.71G>C p.W24S Chr.18: 618071-618071 on Build GRCh38
3527168 c.75G>A p.K25K Chr.18: 618075-618075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146337166 Chr.18: 617817-617817 on Build GRCh38
rs962816630 Chr.18: 617802-617802 on Build GRCh38
rs966462990 Chr.18: 617934-617934 on Build GRCh38
rs759953054 Chr.18: 617950-617950 on Build GRCh38
rs201337289 Chr.18: 618021-618021 on Build GRCh38
rs35437309 Chr.18: 618036-618036 on Build GRCh38
rs549976296 Chr.18: 617730-617730 on Build GRCh38
rs752597508 Chr.18: 618056-618056 on Build GRCh38
rs765102691 Chr.18: 618051-618051 on Build GRCh38
rs9960753 Chr.18: 617928-617928 on Build GRCh38
rs187911981 Chr.18: 617806-617806 on Build GRCh38
rs758743019 Chr.18: 617987-617987 on Build GRCh38
rs555564906 Chr.18: 617942-617942 on Build GRCh38
rs185097213 Chr.18: 617737-617737 on Build GRCh38
rs1026344027 Chr.18: 617818-617818 on Build GRCh38
rs766405796 Chr.18: 617978-617978 on Build GRCh38
rs201505273 Chr.18: 617775-617776 on Build GRCh38
rs116985056 Chr.18: 618015-618015 on Build GRCh38
rs779228301 Chr.18: 617985-617985 on Build GRCh38
rs766168896 Chr.18: 618032-618032 on Build GRCh38
rs558235819 Chr.18: 617856-617856 on Build GRCh38
rs771278123 Chr.18: 617998-617998 on Build GRCh38
rs749995478 Chr.18: 617964-617964 on Build GRCh38
rs760235295 Chr.18: 618029-618029 on Build GRCh38
rs1057509408 Chr.18: 617713-617713 on Build GRCh38
rs545215024 Chr.18: 617740-617740 on Build GRCh38
rs754815472 Chr.18: 617983-617983 on Build GRCh38
rs1013511117 Chr.18: 617757-617757 on Build GRCh38
rs576627534 Chr.18: 617924-617924 on Build GRCh38
rs777988778 Chr.18: 617990-617990 on Build GRCh38
rs10550964 Chr.18: 617776-617777 on Build GRCh38
rs774018504 Chr.18: 617958-617958 on Build GRCh38
rs1048147099 Chr.18: 618037-618037 on Build GRCh38
rs1008791487 Chr.18: 617841-617841 on Build GRCh38
rs766611664 Chr.18: 617923-617923 on Build GRCh38
rs1057354536 Chr.18: 618008-618008 on Build GRCh38
rs977823817 Chr.18: 617937-617937 on Build GRCh38
rs539935482 Chr.18: 617842-617842 on Build GRCh38
rs975562978 Chr.18: 617805-617805 on Build GRCh38
rs763915515 Chr.18: 617996-617996 on Build GRCh38
rs531753041 Chr.18: 617726-617726 on Build GRCh38
rs879031267 Chr.18: 617873-617873 on Build GRCh38
rs761269027 Chr.18: 617961-617961 on Build GRCh38
rs771767873 Chr.18: 618024-618024 on Build GRCh38
rs573838333 Chr.18: 618012-618012 on Build GRCh38
rs398058970 Chr.18: 617778-617779 on Build GRCh38
rs1015968381 Chr.18: 617796-617796 on Build GRCh38
rs924629503 Chr.18: 617925-617925 on Build GRCh38
rs770179574 Chr.18: 618013-618013 on Build GRCh38
rs899285550 Chr.18: 617783-617783 on Build GRCh38
rs772589301 Chr.18: 618026-618026 on Build GRCh38
rs1025967366 Chr.18: 617771-617771 on Build GRCh38
rs193214837 Chr.18: 617821-617821 on Build GRCh38
rs73942581 Chr.18: 617772-617772 on Build GRCh38
rs753507594 Chr.18: 617979-617979 on Build GRCh38
rs1003226604 Chr.18: 617762-617762 on Build GRCh38
rs781730115 Chr.18: 618000-618000 on Build GRCh38
rs914979699 Chr.18: 617930-617930 on Build GRCh38
rs755989171 Chr.18: 617969-617969 on Build GRCh38
rs1044671279 Chr.18: 617744-617744 on Build GRCh38
rs748349535 Chr.18: 617986-617986 on Build GRCh38
rs372467847 Chr.18: 617957-617957 on Build GRCh38

View Full Product Details