Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186231816-186232412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLKB1
Entrez Gene ID
3818
Gene Name
kallikrein B1
Gene Aliases
KLK3, PKK, PKKD, PPK
UniGene
Hs.237642
Gene Chromosome Location
Chr.4: 186215714-186258477 on Build GRCh38
Transcripts
NM_000892.4 NM_001318394.1 NM_001318396.1 XM_011531930.2 XM_017008181.1 XM_017008182.1 XM_017008183.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232644 c.89T>G p.F30C Chr.4: 186232157-186232157 on Build GRCh38
6261227 c.101G>T p.G34V Chr.4: 186232169-186232169 on Build GRCh38
120027 c.167G>A p.R56K Chr.4: 186232235-186232235 on Build GRCh38
1618679 c.152G>T p.C51F Chr.4: 186232220-186232220 on Build GRCh38
5937154 c.187C>T p.L63F Chr.4: 186232255-186232255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750128151 Chr.4: 186232142-186232142 on Build GRCh38
rs759506261 Chr.4: 186232115-186232115 on Build GRCh38
rs779565094 Chr.4: 186232152-186232152 on Build GRCh38
rs544290141 Chr.4: 186231871-186231871 on Build GRCh38
rs187970629 Chr.4: 186232350-186232350 on Build GRCh38
rs983928348 Chr.4: 186232051-186232051 on Build GRCh38
rs750325714 Chr.4: 186232229-186232244 on Build GRCh38
rs142462112 Chr.4: 186232278-186232278 on Build GRCh38
rs138408657 Chr.4: 186232173-186232173 on Build GRCh38
rs754196064 Chr.4: 186232135-186232135 on Build GRCh38
rs779821874 Chr.4: 186232078-186232078 on Build GRCh38
rs1035341072 Chr.4: 186232380-186232380 on Build GRCh38
rs201749287 Chr.4: 186232307-186232307 on Build GRCh38
rs902620270 Chr.4: 186232259-186232259 on Build GRCh38
rs369855039 Chr.4: 186232309-186232309 on Build GRCh38
rs114817536 Chr.4: 186232299-186232299 on Build GRCh38
rs1014368211 Chr.4: 186232231-186232231 on Build GRCh38
rs774765326 Chr.4: 186232112-186232112 on Build GRCh38
rs528518519 Chr.4: 186232203-186232203 on Build GRCh38
rs752644194 Chr.4: 186232306-186232306 on Build GRCh38
rs764588974 Chr.4: 186232134-186232134 on Build GRCh38
rs781339817 Chr.4: 186232172-186232172 on Build GRCh38
rs767139832 Chr.4: 186231953-186231954 on Build GRCh38
rs762131874 Chr.4: 186232243-186232243 on Build GRCh38
rs751071483 Chr.4: 186232285-186232285 on Build GRCh38
rs573848787 Chr.4: 186231945-186231945 on Build GRCh38
rs369438768 Chr.4: 186232098-186232098 on Build GRCh38
rs775456108 Chr.4: 186232122-186232122 on Build GRCh38
rs754586302 Chr.4: 186232287-186232287 on Build GRCh38
rs1023894061 Chr.4: 186232335-186232335 on Build GRCh38
rs764844498 Chr.4: 186232137-186232137 on Build GRCh38
rs562611827 Chr.4: 186232050-186232050 on Build GRCh38
rs751039368 Chr.4: 186232153-186232153 on Build GRCh38
rs766233613 Chr.4: 186232284-186232285 on Build GRCh38
rs4253243 Chr.4: 186232357-186232357 on Build GRCh38
rs148277206 Chr.4: 186232136-186232136 on Build GRCh38
rs777462435 Chr.4: 186232189-186232189 on Build GRCh38
rs761185677 Chr.4: 186232126-186232126 on Build GRCh38
rs1024330759 Chr.4: 186231894-186231894 on Build GRCh38
rs563548109 Chr.4: 186231868-186231868 on Build GRCh38
rs1005035816 Chr.4: 186232387-186232387 on Build GRCh38
rs549102630 Chr.4: 186232373-186232373 on Build GRCh38
rs765264091 Chr.4: 186232077-186232077 on Build GRCh38
rs894088265 Chr.4: 186231902-186231902 on Build GRCh38
rs760733760 Chr.4: 186232217-186232217 on Build GRCh38
rs1027580862 Chr.4: 186232313-186232313 on Build GRCh38
rs777170227 Chr.4: 186232242-186232242 on Build GRCh38
rs749039930 Chr.4: 186232311-186232311 on Build GRCh38
rs972063187 Chr.4: 186231982-186231982 on Build GRCh38
rs780197667 Chr.4: 186232320-186232320 on Build GRCh38
rs941996231 Chr.4: 186232130-186232130 on Build GRCh38
rs796467244 Chr.4: 186232167-186232168 on Build GRCh38
rs750771073 Chr.4: 186232277-186232277 on Build GRCh38
rs35389013 Chr.4: 186231914-186231915 on Build GRCh38
rs565625980 Chr.4: 186232386-186232386 on Build GRCh38
rs974624857 Chr.4: 186232132-186232132 on Build GRCh38
rs1041364548 Chr.4: 186232140-186232140 on Build GRCh38
rs576466131 Chr.4: 186232174-186232174 on Build GRCh38
rs964208143 Chr.4: 186232374-186232374 on Build GRCh38
rs758014329 Chr.4: 186232146-186232146 on Build GRCh38
rs767595392 Chr.4: 186232121-186232121 on Build GRCh38
rs371834857 Chr.4: 186232337-186232338 on Build GRCh38
rs748026544 Chr.4: 186232099-186232099 on Build GRCh38
rs762984757 Chr.4: 186232276-186232276 on Build GRCh38
rs1035455796 Chr.4: 186232059-186232059 on Build GRCh38
rs1022670455 Chr.4: 186232004-186232004 on Build GRCh38
rs746053973 Chr.4: 186232190-186232190 on Build GRCh38
rs772197825 Chr.4: 186232199-186232199 on Build GRCh38
rs935403391 Chr.4: 186232273-186232273 on Build GRCh38
rs922345776 Chr.4: 186232389-186232389 on Build GRCh38
rs754965474 Chr.4: 186232171-186232171 on Build GRCh38
rs542357918 Chr.4: 186232335-186232336 on Build GRCh38
rs772333301 Chr.4: 186232314-186232314 on Build GRCh38
rs765543142 Chr.4: 186232250-186232250 on Build GRCh38
rs374181790 Chr.4: 186232239-186232239 on Build GRCh38
rs1032898348 Chr.4: 186231876-186231876 on Build GRCh38
rs370086908 Chr.4: 186232085-186232085 on Build GRCh38
rs778768806 Chr.4: 186232014-186232014 on Build GRCh38
rs747068124 Chr.4: 186232326-186232326 on Build GRCh38
rs865850009 Chr.4: 186232141-186232141 on Build GRCh38
rs867959786 Chr.4: 186232289-186232289 on Build GRCh38
rs962342237 Chr.4: 186231958-186231958 on Build GRCh38
rs771268600 Chr.4: 186232106-186232106 on Build GRCh38
rs1012896681 Chr.4: 186231956-186231956 on Build GRCh38
rs542759899 Chr.4: 186232097-186232097 on Build GRCh38
rs1012380472 Chr.4: 186232312-186232312 on Build GRCh38
rs911047957 Chr.4: 186232067-186232067 on Build GRCh38
rs769752981 Chr.4: 186232186-186232186 on Build GRCh38
rs999718918 Chr.4: 186231845-186231845 on Build GRCh38
rs950489839 Chr.4: 186232062-186232062 on Build GRCh38
rs535642106 Chr.4: 186232371-186232371 on Build GRCh38
rs533148132 Chr.4: 186232088-186232088 on Build GRCh38

View Full Product Details