Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 93242167-93242761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA7
Entrez Gene ID
2045
Gene Name
EPH receptor A7
Gene Aliases
EHK-3, EHK3, EK11, HEK11
UniGene
Hs.73962
Gene Chromosome Location
Chr.6: 93240020-93419687 on Build GRCh38
Transcripts
NM_001288629.1 NM_004440.3 XM_005248669.2 XM_017010365.1

SNP Detail

SNP ID Chromosome Location
rs537210040 Chr.6: 93242356-93242356 on Build GRCh38
rs917452571 Chr.6: 93242649-93242649 on Build GRCh38
rs551642366 Chr.6: 93242607-93242607 on Build GRCh38
rs902908816 Chr.6: 93242394-93242394 on Build GRCh38
rs867136929 Chr.6: 93242448-93242448 on Build GRCh38
rs533451975 Chr.6: 93242724-93242724 on Build GRCh38
rs771341239 Chr.6: 93242616-93242616 on Build GRCh38
rs774940584 Chr.6: 93242349-93242349 on Build GRCh38
rs933524089 Chr.6: 93242508-93242508 on Build GRCh38
rs35956234 Chr.6: 93242339-93242340 on Build GRCh38
rs564465919 Chr.6: 93242716-93242716 on Build GRCh38
rs1026748259 Chr.6: 93242640-93242640 on Build GRCh38
rs954716968 Chr.6: 93242529-93242529 on Build GRCh38
rs148343742 Chr.6: 93242365-93242365 on Build GRCh38
rs745370929 Chr.6: 93242387-93242387 on Build GRCh38
rs1035569618 Chr.6: 93242270-93242270 on Build GRCh38
rs949715382 Chr.6: 93242650-93242650 on Build GRCh38
rs576862153 Chr.6: 93242404-93242404 on Build GRCh38
rs1041374136 Chr.6: 93242660-93242660 on Build GRCh38
rs1042798867 Chr.6: 93242544-93242544 on Build GRCh38
rs960728569 Chr.6: 93242303-93242303 on Build GRCh38
rs950883523 Chr.6: 93242380-93242380 on Build GRCh38
rs891436083 Chr.6: 93242532-93242532 on Build GRCh38
rs575755807 Chr.6: 93242634-93242634 on Build GRCh38
rs189259446 Chr.6: 93242549-93242549 on Build GRCh38
rs991766043 Chr.6: 93242622-93242622 on Build GRCh38
rs923507219 Chr.6: 93242498-93242498 on Build GRCh38
rs950736471 Chr.6: 93242644-93242644 on Build GRCh38
rs568179102 Chr.6: 93242352-93242352 on Build GRCh38
rs560137620 Chr.6: 93242729-93242729 on Build GRCh38
rs529215774 Chr.6: 93242739-93242739 on Build GRCh38
rs920874790 Chr.6: 93242249-93242249 on Build GRCh38
rs973514732 Chr.6: 93242196-93242196 on Build GRCh38
rs141511769 Chr.6: 93242699-93242699 on Build GRCh38
rs768112576 Chr.6: 93242517-93242517 on Build GRCh38
rs546619166 Chr.6: 93242726-93242726 on Build GRCh38
rs1000041780 Chr.6: 93242412-93242412 on Build GRCh38
rs920772642 Chr.6: 93242595-93242595 on Build GRCh38
rs935717059 Chr.6: 93242670-93242670 on Build GRCh38
rs899656790 Chr.6: 93242732-93242732 on Build GRCh38
rs766641001 Chr.6: 93242669-93242669 on Build GRCh38
rs199733455 Chr.6: 93242270-93242274 on Build GRCh38
rs899769145 Chr.6: 93242476-93242476 on Build GRCh38
rs867794653 Chr.6: 93242684-93242684 on Build GRCh38
rs542553998 Chr.6: 93242591-93242591 on Build GRCh38
rs866045845 Chr.6: 93242255-93242259 on Build GRCh38
rs909402856 Chr.6: 93242438-93242438 on Build GRCh38
rs539473321 Chr.6: 93242541-93242541 on Build GRCh38
rs9342356 Chr.6: 93242193-93242193 on Build GRCh38
rs997049994 Chr.6: 93242736-93242736 on Build GRCh38
rs1011284914 Chr.6: 93242253-93242253 on Build GRCh38
rs973567142 Chr.6: 93242594-93242594 on Build GRCh38
rs768851772 Chr.6: 93242458-93242458 on Build GRCh38
rs762449697 Chr.6: 93242505-93242505 on Build GRCh38
rs984865553 Chr.6: 93242418-93242418 on Build GRCh38
rs895476918 Chr.6: 93242332-93242332 on Build GRCh38
rs548782900 Chr.6: 93242265-93242265 on Build GRCh38
rs775029170 Chr.6: 93242503-93242503 on Build GRCh38
rs937591253 Chr.6: 93242260-93242260 on Build GRCh38
rs919226682 Chr.6: 93242366-93242366 on Build GRCh38
rs1054280906 Chr.6: 93242449-93242449 on Build GRCh38
rs1008829745 Chr.6: 93242536-93242536 on Build GRCh38
rs41273545 Chr.6: 93242563-93242563 on Build GRCh38

View Full Product Details