Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76328911-76329473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAG
Entrez Gene ID
7532
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Gene Aliases
14-3-3GAMMA, PPP1R170
UniGene
Hs.744840
Gene Chromosome Location
Chr.7: 76326791-76359025 on Build GRCh38
Transcripts
NM_012479.3

SNP Detail

SNP ID Chromosome Location
rs760535528 Chr.7: 76328988-76328988 on Build GRCh38
rs761870996 Chr.7: 76329290-76329290 on Build GRCh38
rs553097822 Chr.7: 76329123-76329123 on Build GRCh38
rs947581910 Chr.7: 76328989-76328989 on Build GRCh38
rs11541901 Chr.7: 76329033-76329033 on Build GRCh38
rs978075675 Chr.7: 76329302-76329302 on Build GRCh38
rs944190086 Chr.7: 76329350-76329350 on Build GRCh38
rs1044439739 Chr.7: 76329229-76329229 on Build GRCh38
rs958423498 Chr.7: 76329245-76329245 on Build GRCh38
rs1027216602 Chr.7: 76329392-76329392 on Build GRCh38
rs571501867 Chr.7: 76328946-76328946 on Build GRCh38
rs566056923 Chr.7: 76329125-76329125 on Build GRCh38
rs776743778 Chr.7: 76329049-76329049 on Build GRCh38
rs566614979 Chr.7: 76329128-76329128 on Build GRCh38
rs962180745 Chr.7: 76329185-76329185 on Build GRCh38
rs191351438 Chr.7: 76329341-76329341 on Build GRCh38
rs73357710 Chr.7: 76329237-76329237 on Build GRCh38
rs981319191 Chr.7: 76329188-76329188 on Build GRCh38
rs1040659534 Chr.7: 76329132-76329132 on Build GRCh38
rs996899173 Chr.7: 76329091-76329091 on Build GRCh38
rs771897820 Chr.7: 76328947-76328947 on Build GRCh38
rs932698131 Chr.7: 76329386-76329386 on Build GRCh38
rs991241343 Chr.7: 76329273-76329273 on Build GRCh38
rs1053668885 Chr.7: 76329174-76329174 on Build GRCh38
rs1026918029 Chr.7: 76329093-76329093 on Build GRCh38
rs140844951 Chr.7: 76329434-76329434 on Build GRCh38
rs200070734 Chr.7: 76329180-76329181 on Build GRCh38
rs924912638 Chr.7: 76329343-76329343 on Build GRCh38
rs1035573080 Chr.7: 76329336-76329336 on Build GRCh38
rs11541900 Chr.7: 76329080-76329080 on Build GRCh38
rs80074591 Chr.7: 76329284-76329284 on Build GRCh38
rs900027929 Chr.7: 76329384-76329384 on Build GRCh38
rs899955816 Chr.7: 76329058-76329058 on Build GRCh38
rs985167735 Chr.7: 76329088-76329088 on Build GRCh38
rs1002742231 Chr.7: 76329012-76329012 on Build GRCh38
rs764323565 Chr.7: 76329270-76329270 on Build GRCh38
rs879085474 Chr.7: 76329180-76329180 on Build GRCh38
rs73703132 Chr.7: 76329435-76329435 on Build GRCh38
rs770729884 Chr.7: 76329032-76329032 on Build GRCh38
rs547444143 Chr.7: 76329070-76329070 on Build GRCh38
rs934865377 Chr.7: 76329164-76329164 on Build GRCh38
rs772819920 Chr.7: 76328965-76328965 on Build GRCh38
rs893187833 Chr.7: 76328973-76328973 on Build GRCh38
rs777215284 Chr.7: 76329040-76329040 on Build GRCh38
rs969971072 Chr.7: 76328991-76328991 on Build GRCh38
rs888449760 Chr.7: 76329114-76329114 on Build GRCh38
rs952557834 Chr.7: 76329084-76329084 on Build GRCh38
rs532569269 Chr.7: 76328977-76328977 on Build GRCh38
rs1004254107 Chr.7: 76329447-76329447 on Build GRCh38
rs1011588824 Chr.7: 76328979-76328979 on Build GRCh38
rs768203272 Chr.7: 76329013-76329013 on Build GRCh38
rs923479681 Chr.7: 76329150-76329150 on Build GRCh38
rs1041167254 Chr.7: 76329381-76329381 on Build GRCh38
rs571008155 Chr.7: 76328964-76328964 on Build GRCh38
rs68159547 Chr.7: 76329068-76329069 on Build GRCh38
rs878957226 Chr.7: 76329181-76329181 on Build GRCh38

View Full Product Details