Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73420988-73421559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAU2
Entrez Gene ID
27067
Gene Name
staufen double-stranded RNA binding protein 2
Gene Aliases
39K2, 39K3
UniGene
Hs.561815
Gene Chromosome Location
Chr.8: 73420369-73747708 on Build GRCh38
Transcripts
NM_001164380.1 NM_001164381.1 NM_001164382.1 NM_001164383.1 NR_038406.1

Gene Symbol
STAU2-AS1
Entrez Gene ID
100128126
Gene Name
STAU2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.679921
Gene Chromosome Location
Chr.8: 73420074-73441518 on Build GRCh38
Transcripts
NM_001164380.1 NM_001164381.1 NM_001164382.1 NM_001164383.1 NR_038406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825486 c.1494G>T p.K498N Chr.8: 73421393-73421393 on Build GRCh38
6347006 c.1509C>A p.A503A Chr.8: 73421378-73421378 on Build GRCh38
6347005 c.1707C>A p.A569A Chr.8: 73421378-73421378 on Build GRCh38
4825485 c.1692G>T p.K564N Chr.8: 73421393-73421393 on Build GRCh38
4836418 c.1509C>G p.A503A Chr.8: 73421378-73421378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996638734 Chr.8: 73421526-73421526 on Build GRCh38
rs979845551 Chr.8: 73421480-73421480 on Build GRCh38
rs931928885 Chr.8: 73421063-73421063 on Build GRCh38
rs896476344 Chr.8: 73421197-73421197 on Build GRCh38
rs750936424 Chr.8: 73421353-73421353 on Build GRCh38
rs978584462 Chr.8: 73421234-73421234 on Build GRCh38
rs964213512 Chr.8: 73421506-73421506 on Build GRCh38
rs921809606 Chr.8: 73421053-73421053 on Build GRCh38
rs903070575 Chr.8: 73421436-73421436 on Build GRCh38
rs559969547 Chr.8: 73421324-73421324 on Build GRCh38
rs757383181 Chr.8: 73421257-73421257 on Build GRCh38
rs867437327 Chr.8: 73421366-73421366 on Build GRCh38
rs535388266 Chr.8: 73421016-73421016 on Build GRCh38
rs748286998 Chr.8: 73421364-73421364 on Build GRCh38
rs1038871434 Chr.8: 73421114-73421114 on Build GRCh38
rs1009483490 Chr.8: 73421417-73421417 on Build GRCh38
rs773967008 Chr.8: 73421124-73421124 on Build GRCh38
rs368365328 Chr.8: 73421438-73421438 on Build GRCh38
rs72659432 Chr.8: 73421078-73421078 on Build GRCh38
rs36069340 Chr.8: 73421032-73421033 on Build GRCh38
rs576160250 Chr.8: 73421166-73421166 on Build GRCh38
rs925107577 Chr.8: 73421326-73421326 on Build GRCh38
rs146501256 Chr.8: 73421381-73421381 on Build GRCh38
rs754084239 Chr.8: 73421245-73421245 on Build GRCh38
rs1032874360 Chr.8: 73421449-73421449 on Build GRCh38
rs1035006337 Chr.8: 73421190-73421190 on Build GRCh38
rs905338942 Chr.8: 73421367-73421367 on Build GRCh38
rs184498114 Chr.8: 73421328-73421328 on Build GRCh38
rs949156255 Chr.8: 73421341-73421341 on Build GRCh38
rs774883873 Chr.8: 73421422-73421422 on Build GRCh38
rs145759808 Chr.8: 73421441-73421441 on Build GRCh38
rs925169259 Chr.8: 73421502-73421502 on Build GRCh38
rs192055318 Chr.8: 73421046-73421046 on Build GRCh38
rs758709829 Chr.8: 73421017-73421017 on Build GRCh38
rs887314730 Chr.8: 73421092-73421092 on Build GRCh38
rs1041625120 Chr.8: 73421418-73421418 on Build GRCh38
rs1013174332 Chr.8: 73421161-73421161 on Build GRCh38
rs952225331 Chr.8: 73421227-73421227 on Build GRCh38
rs922297554 Chr.8: 73421311-73421311 on Build GRCh38
rs767288569 Chr.8: 73421451-73421451 on Build GRCh38
rs375647181 Chr.8: 73421355-73421355 on Build GRCh38
rs557706364 Chr.8: 73421151-73421151 on Build GRCh38
rs921972247 Chr.8: 73421289-73421289 on Build GRCh38
rs759350429 Chr.8: 73421140-73421140 on Build GRCh38
rs941501414 Chr.8: 73421113-73421113 on Build GRCh38
rs775365164 Chr.8: 73421502-73421502 on Build GRCh38
rs572846209 Chr.8: 73421238-73421238 on Build GRCh38
rs966125108 Chr.8: 73421239-73421239 on Build GRCh38
rs967618804 Chr.8: 73421226-73421226 on Build GRCh38
rs564062038 Chr.8: 73421379-73421379 on Build GRCh38
rs143999038 Chr.8: 73421320-73421320 on Build GRCh38
rs975148097 Chr.8: 73421263-73421263 on Build GRCh38
rs896530336 Chr.8: 73421397-73421397 on Build GRCh38
rs1020154180 Chr.8: 73421223-73421223 on Build GRCh38
rs555663650 Chr.8: 73421337-73421337 on Build GRCh38
rs564251232 Chr.8: 73421167-73421167 on Build GRCh38
rs541378678 Chr.8: 73421321-73421321 on Build GRCh38
rs543510017 Chr.8: 73421180-73421180 on Build GRCh38
rs1014158229 Chr.8: 73421200-73421200 on Build GRCh38
rs549853579 Chr.8: 73421408-73421408 on Build GRCh38
rs369462384 Chr.8: 73421378-73421378 on Build GRCh38
rs1052229155 Chr.8: 73421332-73421332 on Build GRCh38

View Full Product Details