Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109245331-109245892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDCD1
Entrez Gene ID
84955
Gene Name
NudC domain containing 1
Gene Aliases
CML66, OVA66
UniGene
Hs.380291
Gene Chromosome Location
Chr.8: 109240919-109334205 on Build GRCh38
Transcripts
NM_001128211.1 NM_032869.3 XM_017013910.1 XM_017013911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6731103 c.1344G>A p.W448* Chr.8: 109245350-109245350 on Build GRCh38
6033811 c.1324C>A p.L442I Chr.8: 109245457-109245457 on Build GRCh38
5976586 c.1301T>G p.V434G Chr.8: 109245480-109245480 on Build GRCh38
6111992 c.1263A>T p.K421N Chr.8: 109245431-109245431 on Build GRCh38
4441282 c.1442C>T p.A481V Chr.8: 109245339-109245339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958326651 Chr.8: 109245621-109245621 on Build GRCh38
rs191292225 Chr.8: 109245627-109245627 on Build GRCh38
rs553378846 Chr.8: 109245651-109245651 on Build GRCh38
rs76107104 Chr.8: 109245784-109245784 on Build GRCh38
rs950844171 Chr.8: 109245652-109245652 on Build GRCh38
rs992529705 Chr.8: 109245653-109245653 on Build GRCh38
rs545338782 Chr.8: 109245410-109245410 on Build GRCh38
rs998554212 Chr.8: 109245463-109245463 on Build GRCh38
rs183979237 Chr.8: 109245788-109245788 on Build GRCh38
rs551061486 Chr.8: 109245658-109245658 on Build GRCh38
rs1035424232 Chr.8: 109245700-109245700 on Build GRCh38
rs868162659 Chr.8: 109245808-109245808 on Build GRCh38
rs751277523 Chr.8: 109245354-109245355 on Build GRCh38
rs915424686 Chr.8: 109245720-109245722 on Build GRCh38
rs1026376114 Chr.8: 109245633-109245633 on Build GRCh38
rs1007430834 Chr.8: 109245539-109245539 on Build GRCh38
rs867733315 Chr.8: 109245416-109245416 on Build GRCh38
rs771665042 Chr.8: 109245495-109245495 on Build GRCh38
rs892700335 Chr.8: 109245846-109245846 on Build GRCh38
rs917170527 Chr.8: 109245668-109245668 on Build GRCh38
rs144957968 Chr.8: 109245831-109245831 on Build GRCh38
rs573928354 Chr.8: 109245534-109245534 on Build GRCh38
rs749609758 Chr.8: 109245553-109245553 on Build GRCh38
rs777836000 Chr.8: 109245441-109245441 on Build GRCh38
rs1017396253 Chr.8: 109245544-109245544 on Build GRCh38
rs920391736 Chr.8: 109245705-109245705 on Build GRCh38
rs780365437 Chr.8: 109245473-109245473 on Build GRCh38
rs899231828 Chr.8: 109245443-109245443 on Build GRCh38
rs188709383 Chr.8: 109245832-109245832 on Build GRCh38
rs1009688200 Chr.8: 109245536-109245536 on Build GRCh38
rs567510225 Chr.8: 109245761-109245761 on Build GRCh38
rs770182102 Chr.8: 109245355-109245355 on Build GRCh38
rs867927267 Chr.8: 109245752-109245752 on Build GRCh38
rs770673946 Chr.8: 109245530-109245530 on Build GRCh38
rs770968996 Chr.8: 109245683-109245684 on Build GRCh38
rs760309532 Chr.8: 109245529-109245529 on Build GRCh38
rs141306275 Chr.8: 109245780-109245780 on Build GRCh38
rs374965981 Chr.8: 109245467-109245467 on Build GRCh38
rs558523715 Chr.8: 109245593-109245593 on Build GRCh38
rs146565102 Chr.8: 109245772-109245772 on Build GRCh38
rs770248396 Chr.8: 109245472-109245472 on Build GRCh38
rs1009746908 Chr.8: 109245847-109245847 on Build GRCh38
rs781445087 Chr.8: 109245450-109245450 on Build GRCh38
rs67022633 Chr.8: 109245773-109245773 on Build GRCh38
rs559721113 Chr.8: 109245600-109245600 on Build GRCh38
rs879049902 Chr.8: 109245546-109245546 on Build GRCh38
rs530482530 Chr.8: 109245721-109245721 on Build GRCh38
rs766227678 Chr.8: 109245400-109245400 on Build GRCh38
rs757497616 Chr.8: 109245447-109245447 on Build GRCh38
rs765083084 Chr.8: 109245408-109245408 on Build GRCh38
rs930749750 Chr.8: 109245471-109245471 on Build GRCh38
rs201312641 Chr.8: 109245476-109245476 on Build GRCh38
rs761581682 Chr.8: 109245415-109245417 on Build GRCh38
rs774596345 Chr.8: 109245686-109245686 on Build GRCh38
rs971912291 Chr.8: 109245704-109245704 on Build GRCh38
rs776107866 Chr.8: 109245362-109245362 on Build GRCh38
rs940615190 Chr.8: 109245671-109245671 on Build GRCh38
rs767241167 Chr.8: 109245496-109245496 on Build GRCh38
rs772929517 Chr.8: 109245380-109245380 on Build GRCh38
rs1034107029 Chr.8: 109245594-109245594 on Build GRCh38
rs751744029 Chr.8: 109245444-109245444 on Build GRCh38
rs959613185 Chr.8: 109245713-109245713 on Build GRCh38
rs550654301 Chr.8: 109245756-109245756 on Build GRCh38
rs1044001335 Chr.8: 109245848-109245848 on Build GRCh38
rs760434103 Chr.8: 109245395-109245395 on Build GRCh38
rs541308898 Chr.8: 109245783-109245783 on Build GRCh38
rs1042782466 Chr.8: 109245420-109245420 on Build GRCh38
rs753552492 Chr.8: 109245405-109245405 on Build GRCh38
rs758416339 Chr.8: 109245409-109245409 on Build GRCh38
rs903981753 Chr.8: 109245868-109245868 on Build GRCh38
rs772869821 Chr.8: 109245496-109245496 on Build GRCh38
rs761212633 Chr.8: 109245363-109245363 on Build GRCh38
rs940499270 Chr.8: 109245383-109245383 on Build GRCh38
rs1051477532 Chr.8: 109245779-109245779 on Build GRCh38
rs780431890 Chr.8: 109245533-109245533 on Build GRCh38
rs187458831 Chr.8: 109245517-109245517 on Build GRCh38
rs917180593 Chr.8: 109245364-109245364 on Build GRCh38
rs1022908370 Chr.8: 109245543-109245543 on Build GRCh38
rs759414155 Chr.8: 109245406-109245406 on Build GRCh38
rs370651891 Chr.8: 109245375-109245375 on Build GRCh38
rs564911190 Chr.8: 109245716-109245716 on Build GRCh38

View Full Product Details