Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 91085635-91085994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD6B
Entrez Gene ID
51633
Gene Name
OTU domain containing 6B
Gene Aliases
CGI-77, DUBA-5, DUBA5
UniGene
Hs.30532
Gene Chromosome Location
Chr.8: 91070196-91087095 on Build GRCh38
Transcripts
NM_001286745.1 NM_016023.3 XM_011517129.2

SNP Detail

SNP ID Chromosome Location
rs758034950 Chr.8: 91085926-91085926 on Build GRCh38
rs190300400 Chr.8: 91085858-91085858 on Build GRCh38
rs180792439 Chr.8: 91085724-91085724 on Build GRCh38
rs911301558 Chr.8: 91085952-91085952 on Build GRCh38
rs986877683 Chr.8: 91085918-91085918 on Build GRCh38
rs185195505 Chr.8: 91085763-91085763 on Build GRCh38
rs1022972190 Chr.8: 91085734-91085734 on Build GRCh38
rs1021542885 Chr.8: 91085771-91085772 on Build GRCh38
rs942746517 Chr.8: 91085961-91085961 on Build GRCh38
rs1034081350 Chr.8: 91085673-91085673 on Build GRCh38
rs572517188 Chr.8: 91085684-91085684 on Build GRCh38
rs985479919 Chr.8: 91085912-91085912 on Build GRCh38
rs538912128 Chr.8: 91085933-91085933 on Build GRCh38
rs968295243 Chr.8: 91085746-91085746 on Build GRCh38
rs924111292 Chr.8: 91085838-91085838 on Build GRCh38
rs999750997 Chr.8: 91085687-91085687 on Build GRCh38
rs878927230 Chr.8: 91085893-91085897 on Build GRCh38
rs954135280 Chr.8: 91085870-91085870 on Build GRCh38
rs1019758504 Chr.8: 91085936-91085936 on Build GRCh38
rs531436202 Chr.8: 91085707-91085707 on Build GRCh38
rs551920191 Chr.8: 91085758-91085758 on Build GRCh38
rs754828891 Chr.8: 91085677-91085677 on Build GRCh38

View Full Product Details