Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203870166-203870723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPE
Entrez Gene ID
6635
Gene Name
small nuclear ribonucleoprotein polypeptide E
Gene Aliases
HYPT11, SME, Sm-E, snRNP-E
UniGene
Hs.334612
Gene Chromosome Location
Chr.1: 203861585-203871152 on Build GRCh38
Transcripts
NM_001304464.1 NM_003094.3 NR_130746.1

SNP Detail

SNP ID Chromosome Location
rs775731903 Chr.1: 203870353-203870353 on Build GRCh38
rs776979299 Chr.1: 203870196-203870196 on Build GRCh38
rs907780384 Chr.1: 203870284-203870284 on Build GRCh38
rs4341420 Chr.1: 203870490-203870490 on Build GRCh38
rs987573390 Chr.1: 203870610-203870610 on Build GRCh38
rs918049771 Chr.1: 203870228-203870228 on Build GRCh38
rs759731096 Chr.1: 203870282-203870282 on Build GRCh38
rs912277782 Chr.1: 203870383-203870383 on Build GRCh38
rs113918394 Chr.1: 203870195-203870195 on Build GRCh38
rs75487868 Chr.1: 203870341-203870341 on Build GRCh38
rs921057092 Chr.1: 203870675-203870675 on Build GRCh38
rs111624280 Chr.1: 203870390-203870390 on Build GRCh38
rs759385010 Chr.1: 203870498-203870498 on Build GRCh38
rs535550741 Chr.1: 203870521-203870521 on Build GRCh38
rs183934532 Chr.1: 203870598-203870598 on Build GRCh38
rs533542527 Chr.1: 203870460-203870460 on Build GRCh38
rs549005560 Chr.1: 203870632-203870632 on Build GRCh38
rs1054104786 Chr.1: 203870484-203870484 on Build GRCh38
rs538627140 Chr.1: 203870689-203870689 on Build GRCh38
rs994884542 Chr.1: 203870569-203870569 on Build GRCh38
rs752357329 Chr.1: 203870527-203870527 on Build GRCh38
rs77576156 Chr.1: 203870491-203870491 on Build GRCh38
rs963434658 Chr.1: 203870308-203870308 on Build GRCh38
rs760754803 Chr.1: 203870455-203870455 on Build GRCh38
rs890311380 Chr.1: 203870677-203870677 on Build GRCh38
rs75335430 Chr.1: 203870387-203870387 on Build GRCh38
rs757856767 Chr.1: 203870528-203870528 on Build GRCh38
rs970805043 Chr.1: 203870247-203870247 on Build GRCh38
rs1047750865 Chr.1: 203870243-203870243 on Build GRCh38
rs1046272287 Chr.1: 203870492-203870492 on Build GRCh38
rs549834490 Chr.1: 203870344-203870344 on Build GRCh38
rs2298871 Chr.1: 203870373-203870373 on Build GRCh38
rs5003874 Chr.1: 203870377-203870377 on Build GRCh38
rs949381197 Chr.1: 203870233-203870233 on Build GRCh38
rs773295919 Chr.1: 203870400-203870400 on Build GRCh38
rs931871598 Chr.1: 203870369-203870369 on Build GRCh38
rs752777099 Chr.1: 203870414-203870414 on Build GRCh38
rs141785435 Chr.1: 203870653-203870653 on Build GRCh38
rs764824747 Chr.1: 203870518-203870518 on Build GRCh38
rs564527825 Chr.1: 203870454-203870454 on Build GRCh38
rs7415947 Chr.1: 203870231-203870231 on Build GRCh38
rs950974752 Chr.1: 203870607-203870607 on Build GRCh38
rs531274218 Chr.1: 203870397-203870397 on Build GRCh38
rs964875047 Chr.1: 203870641-203870641 on Build GRCh38
rs963698945 Chr.1: 203870519-203870519 on Build GRCh38
rs1037629524 Chr.1: 203870332-203870332 on Build GRCh38
rs942037446 Chr.1: 203870536-203870536 on Build GRCh38
rs4512716 Chr.1: 203870416-203870416 on Build GRCh38
rs367872182 Chr.1: 203870274-203870274 on Build GRCh38
rs71281152 Chr.1: 203870299-203870300 on Build GRCh38
rs769956993 Chr.1: 203870330-203870330 on Build GRCh38
rs1002269985 Chr.1: 203870506-203870506 on Build GRCh38

View Full Product Details