Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42495083-42495612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
UniGene
Hs.511149
Gene Chromosome Location
Chr.15: 42491244-42533061 on Build GRCh38
Transcripts
NM_003825.3 NM_130798.2 XM_006720725.2 XM_017022692.1 XM_017022693.1 XM_017022694.1

SNP Detail

SNP ID Chromosome Location
rs528329241 Chr.15: 42495447-42495447 on Build GRCh38
rs779065591 Chr.15: 42495325-42495325 on Build GRCh38
rs753158536 Chr.15: 42495108-42495108 on Build GRCh38
rs977210797 Chr.15: 42495256-42495256 on Build GRCh38
rs570678977 Chr.15: 42495202-42495202 on Build GRCh38
rs547521034 Chr.15: 42495179-42495179 on Build GRCh38
rs994432517 Chr.15: 42495124-42495124 on Build GRCh38
rs1024761338 Chr.15: 42495545-42495545 on Build GRCh38
rs528845595 Chr.15: 42495268-42495268 on Build GRCh38
rs548965009 Chr.15: 42495512-42495512 on Build GRCh38
rs746093448 Chr.15: 42495448-42495448 on Build GRCh38
rs549037075 Chr.15: 42495522-42495522 on Build GRCh38
rs564023769 Chr.15: 42495223-42495223 on Build GRCh38
rs528362891 Chr.15: 42495278-42495278 on Build GRCh38
rs1016721310 Chr.15: 42495292-42495292 on Build GRCh38
rs936300301 Chr.15: 42495386-42495386 on Build GRCh38
rs187235019 Chr.15: 42495309-42495309 on Build GRCh38
rs780019896 Chr.15: 42495118-42495118 on Build GRCh38
rs961920669 Chr.15: 42495312-42495312 on Build GRCh38
rs541110537 Chr.15: 42495401-42495401 on Build GRCh38
rs926519565 Chr.15: 42495582-42495582 on Build GRCh38
rs182314520 Chr.15: 42495488-42495488 on Build GRCh38
rs6493048 Chr.15: 42495261-42495261 on Build GRCh38
rs980443157 Chr.15: 42495344-42495344 on Build GRCh38
rs1055065995 Chr.15: 42495130-42495130 on Build GRCh38
rs1016360505 Chr.15: 42495511-42495511 on Build GRCh38
rs748016540 Chr.15: 42495572-42495572 on Build GRCh38
rs561731224 Chr.15: 42495535-42495535 on Build GRCh38
rs568287631 Chr.15: 42495519-42495519 on Build GRCh38
rs949285460 Chr.15: 42495326-42495326 on Build GRCh38
rs546830416 Chr.15: 42495453-42495453 on Build GRCh38
rs1018409897 Chr.15: 42495526-42495526 on Build GRCh38
rs926282895 Chr.15: 42495380-42495380 on Build GRCh38
rs149605784 Chr.15: 42495164-42495164 on Build GRCh38
rs1018336220 Chr.15: 42495205-42495205 on Build GRCh38
rs191267190 Chr.15: 42495411-42495411 on Build GRCh38
rs556535039 Chr.15: 42495230-42495230 on Build GRCh38
rs1026282588 Chr.15: 42495138-42495138 on Build GRCh38
rs927083685 Chr.15: 42495366-42495366 on Build GRCh38
rs1010798770 Chr.15: 42495181-42495181 on Build GRCh38
rs535816127 Chr.15: 42495302-42495302 on Build GRCh38
rs964084017 Chr.15: 42495247-42495247 on Build GRCh38
rs776286543 Chr.15: 42495557-42495557 on Build GRCh38
rs781427497 Chr.15: 42495105-42495105 on Build GRCh38
rs543604514 Chr.15: 42495476-42495476 on Build GRCh38
rs895449766 Chr.15: 42495112-42495112 on Build GRCh38
rs577734506 Chr.15: 42495467-42495467 on Build GRCh38
rs147305138 Chr.15: 42495314-42495314 on Build GRCh38
rs902138347 Chr.15: 42495457-42495457 on Build GRCh38
rs186976654 Chr.15: 42495591-42495591 on Build GRCh38
rs970652120 Chr.15: 42495563-42495563 on Build GRCh38
rs778381707 Chr.15: 42495239-42495239 on Build GRCh38
rs373761385 Chr.15: 42495490-42495490 on Build GRCh38
rs972277658 Chr.15: 42495319-42495319 on Build GRCh38

View Full Product Details