Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65598157-65598543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
UniGene
Hs.6686
Gene Chromosome Location
Chr.15: 65578757-65611289 on Build GRCh38
Transcripts
NM_001136043.2 NM_001207058.1 NM_001207059.1 NR_045104.1 NR_045105.2 NR_045106.2 NR_045901.1 XM_006720708.3 XM_006720709.3 XM_006720711.2 XM_011522101.2 XM_017022644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503232 c.558C>T p.I186I Chr.15: 65598340-65598340 on Build GRCh38
6821052 c.737C>T p.T246M Chr.15: 65598440-65598440 on Build GRCh38
6209436 c.657C>T p.G219G Chr.15: 65598412-65598412 on Build GRCh38
4868625 c.829G>T p.D277Y Chr.15: 65598348-65598348 on Build GRCh38
1301347 c.606-1G>A p.? Chr.15: 65598464-65598464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369534890 Chr.15: 65598338-65598338 on Build GRCh38
rs527608898 Chr.15: 65598517-65598517 on Build GRCh38
rs149850636 Chr.15: 65598230-65598230 on Build GRCh38
rs747746982 Chr.15: 65598279-65598279 on Build GRCh38
rs773415586 Chr.15: 65598499-65598499 on Build GRCh38
rs759649959 Chr.15: 65598515-65598515 on Build GRCh38
rs77371623 Chr.15: 65598371-65598371 on Build GRCh38
rs150997635 Chr.15: 65598350-65598350 on Build GRCh38
rs150662544 Chr.15: 65598207-65598207 on Build GRCh38
rs1015656884 Chr.15: 65598482-65598485 on Build GRCh38
rs1053930203 Chr.15: 65598248-65598248 on Build GRCh38
rs748828387 Chr.15: 65598302-65598302 on Build GRCh38
rs770295767 Chr.15: 65598498-65598498 on Build GRCh38
rs761092071 Chr.15: 65598323-65598323 on Build GRCh38
rs766474361 Chr.15: 65598293-65598299 on Build GRCh38
rs766372521 Chr.15: 65598362-65598362 on Build GRCh38
rs892846847 Chr.15: 65598257-65598257 on Build GRCh38
rs763031365 Chr.15: 65598357-65598357 on Build GRCh38
rs193047786 Chr.15: 65598233-65598233 on Build GRCh38
rs755274622 Chr.15: 65598325-65598326 on Build GRCh38
rs545926726 Chr.15: 65598243-65598243 on Build GRCh38
rs770407914 Chr.15: 65598439-65598439 on Build GRCh38
rs765550602 Chr.15: 65598342-65598342 on Build GRCh38
rs922541977 Chr.15: 65598209-65598209 on Build GRCh38
rs769103361 Chr.15: 65598324-65598324 on Build GRCh38
rs780405954 Chr.15: 65598310-65598310 on Build GRCh38
rs763111996 Chr.15: 65598502-65598502 on Build GRCh38
rs777473133 Chr.15: 65598411-65598411 on Build GRCh38
rs971425473 Chr.15: 65598348-65598348 on Build GRCh38
rs1019922846 Chr.15: 65598315-65598315 on Build GRCh38
rs1025216693 Chr.15: 65598336-65598336 on Build GRCh38
rs999955405 Chr.15: 65598237-65598237 on Build GRCh38
rs201799669 Chr.15: 65598440-65598440 on Build GRCh38
rs751729503 Chr.15: 65598303-65598304 on Build GRCh38
rs748910708 Chr.15: 65598412-65598412 on Build GRCh38
rs755972081 Chr.15: 65598396-65598396 on Build GRCh38
rs752461680 Chr.15: 65598372-65598372 on Build GRCh38
rs751875750 Chr.15: 65598505-65598505 on Build GRCh38
rs751470132 Chr.15: 65598370-65598370 on Build GRCh38
rs141108853 Chr.15: 65598443-65598443 on Build GRCh38
rs769410854 Chr.15: 65598280-65598280 on Build GRCh38
rs139910286 Chr.15: 65598215-65598215 on Build GRCh38
rs777159240 Chr.15: 65598330-65598330 on Build GRCh38
rs547953158 Chr.15: 65598446-65598446 on Build GRCh38
rs1010337603 Chr.15: 65598284-65598284 on Build GRCh38
rs777342925 Chr.15: 65598471-65598471 on Build GRCh38
rs1040950569 Chr.15: 65598190-65598190 on Build GRCh38
rs568030934 Chr.15: 65598503-65598503 on Build GRCh38
rs770409345 Chr.15: 65598303-65598303 on Build GRCh38
rs762253453 Chr.15: 65598476-65598476 on Build GRCh38
rs562404641 Chr.15: 65598298-65598298 on Build GRCh38
rs977026155 Chr.15: 65598519-65598519 on Build GRCh38
rs777445601 Chr.15: 65598301-65598301 on Build GRCh38

View Full Product Details