Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35419964-35420428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFN12
Entrez Gene ID
55106
Gene Name
schlafen family member 12
Gene Aliases
SLFN3
UniGene
Hs.663548
Gene Chromosome Location
Chr.17: 35406684-35433176 on Build GRCh38
Transcripts
NM_001289009.1 NM_018042.4 XM_005257995.4 XM_011524967.2 XM_017024809.1 XM_017024810.1 XM_017024811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
977918 c.1126C>T p.R376W Chr.17: 35420295-35420295 on Build GRCh38
6311311 c.1140C>A p.H380Q Chr.17: 35420281-35420281 on Build GRCh38
5447212 c.1084T>G p.S362A Chr.17: 35420337-35420337 on Build GRCh38
3516254 c.1082C>G p.T361R Chr.17: 35420339-35420339 on Build GRCh38
6271178 c.1101C>A p.H367Q Chr.17: 35420320-35420320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770648023 Chr.17: 35420352-35420352 on Build GRCh38
rs764674461 Chr.17: 35420252-35420252 on Build GRCh38
rs780123794 Chr.17: 35420176-35420176 on Build GRCh38
rs776494856 Chr.17: 35420246-35420246 on Build GRCh38
rs751925498 Chr.17: 35420281-35420281 on Build GRCh38
rs367674776 Chr.17: 35420240-35420240 on Build GRCh38
rs769683803 Chr.17: 35420358-35420358 on Build GRCh38
rs759075722 Chr.17: 35420251-35420251 on Build GRCh38
rs749873640 Chr.17: 35420079-35420079 on Build GRCh38
rs371196916 Chr.17: 35420296-35420296 on Build GRCh38
rs147654371 Chr.17: 35420339-35420339 on Build GRCh38
rs554936005 Chr.17: 35420115-35420115 on Build GRCh38
rs762511546 Chr.17: 35420376-35420376 on Build GRCh38
rs773454037 Chr.17: 35420379-35420380 on Build GRCh38
rs762549453 Chr.17: 35420275-35420275 on Build GRCh38
rs750922602 Chr.17: 35420329-35420329 on Build GRCh38
rs1047397684 Chr.17: 35420204-35420204 on Build GRCh38
rs577737999 Chr.17: 35420174-35420174 on Build GRCh38
rs772921548 Chr.17: 35420228-35420228 on Build GRCh38
rs775040167 Chr.17: 35420259-35420259 on Build GRCh38
rs557249288 Chr.17: 35420304-35420304 on Build GRCh38
rs191160447 Chr.17: 35420083-35420083 on Build GRCh38
rs778451166 Chr.17: 35420049-35420049 on Build GRCh38
rs565485710 Chr.17: 35420133-35420133 on Build GRCh38
rs200355125 Chr.17: 35420343-35420343 on Build GRCh38
rs111326215 Chr.17: 35420132-35420132 on Build GRCh38
rs756285406 Chr.17: 35420335-35420335 on Build GRCh38
rs970108737 Chr.17: 35420066-35420066 on Build GRCh38
rs912308030 Chr.17: 35420188-35420188 on Build GRCh38
rs573971221 Chr.17: 35420338-35420338 on Build GRCh38
rs747006472 Chr.17: 35420242-35420242 on Build GRCh38
rs928609574 Chr.17: 35420091-35420091 on Build GRCh38
rs372367340 Chr.17: 35420205-35420205 on Build GRCh38
rs745851173 Chr.17: 35420356-35420356 on Build GRCh38
rs367977896 Chr.17: 35420355-35420355 on Build GRCh38
rs1050891933 Chr.17: 35420365-35420365 on Build GRCh38
rs774926196 Chr.17: 35420371-35420371 on Build GRCh38
rs762704378 Chr.17: 35420382-35420383 on Build GRCh38
rs200992312 Chr.17: 35420295-35420295 on Build GRCh38
rs920950397 Chr.17: 35420073-35420073 on Build GRCh38
rs527818674 Chr.17: 35420399-35420399 on Build GRCh38
rs780046876 Chr.17: 35420336-35420336 on Build GRCh38
rs534699569 Chr.17: 35420134-35420134 on Build GRCh38
rs757978083 Chr.17: 35420141-35420141 on Build GRCh38
rs990283090 Chr.17: 35419988-35419988 on Build GRCh38
rs962961251 Chr.17: 35420019-35420019 on Build GRCh38
rs557682614 Chr.17: 35420154-35420154 on Build GRCh38
rs768904470 Chr.17: 35420225-35420225 on Build GRCh38
rs763665729 Chr.17: 35420384-35420384 on Build GRCh38

View Full Product Details