Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43607090-43607551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF428
Entrez Gene ID
126299
Gene Name
zinc finger protein 428
Gene Aliases
C19orf37, Zfp428
UniGene
Hs.99093
Gene Chromosome Location
Chr.19: 43607224-43619862 on Build GRCh38
Transcripts
NM_182498.3

SNP Detail

SNP ID Chromosome Location
rs896442250 Chr.19: 43607345-43607345 on Build GRCh38
rs556007843 Chr.19: 43607196-43607196 on Build GRCh38
rs978524775 Chr.19: 43607410-43607410 on Build GRCh38
rs554385677 Chr.19: 43607340-43607340 on Build GRCh38
rs1026378653 Chr.19: 43607397-43607398 on Build GRCh38
rs796636636 Chr.19: 43607370-43607370 on Build GRCh38
rs62114065 Chr.19: 43607397-43607397 on Build GRCh38
rs954382245 Chr.19: 43607397-43607400 on Build GRCh38
rs994058512 Chr.19: 43607299-43607299 on Build GRCh38
rs183768748 Chr.19: 43607260-43607260 on Build GRCh38
rs576765471 Chr.19: 43607444-43607444 on Build GRCh38
rs971771703 Chr.19: 43607395-43607396 on Build GRCh38
rs536010412 Chr.19: 43607326-43607326 on Build GRCh38
rs935928012 Chr.19: 43607397-43607426 on Build GRCh38
rs747273468 Chr.19: 43607323-43607323 on Build GRCh38
rs146070175 Chr.19: 43607373-43607374 on Build GRCh38
rs910292936 Chr.19: 43607400-43607400 on Build GRCh38
rs941453294 Chr.19: 43607253-43607253 on Build GRCh38
rs904445130 Chr.19: 43607256-43607256 on Build GRCh38
rs892490749 Chr.19: 43607436-43607437 on Build GRCh38
rs922841653 Chr.19: 43607443-43607443 on Build GRCh38
rs985444048 Chr.19: 43607506-43607506 on Build GRCh38
rs757673799 Chr.19: 43607333-43607333 on Build GRCh38
rs866707300 Chr.19: 43607428-43607428 on Build GRCh38
rs989486457 Chr.19: 43607132-43607132 on Build GRCh38
rs765887082 Chr.19: 43607530-43607530 on Build GRCh38
rs568121262 Chr.19: 43607341-43607341 on Build GRCh38
rs571682501 Chr.19: 43607394-43607395 on Build GRCh38
rs987342148 Chr.19: 43607139-43607139 on Build GRCh38
rs774038137 Chr.19: 43607374-43607375 on Build GRCh38
rs1016529586 Chr.19: 43607385-43607396 on Build GRCh38
rs536673309 Chr.19: 43607407-43607407 on Build GRCh38
rs949406058 Chr.19: 43607437-43607437 on Build GRCh38
rs943243361 Chr.19: 43607406-43607406 on Build GRCh38
rs1002371532 Chr.19: 43607383-43607396 on Build GRCh38
rs398034727 Chr.19: 43607435-43607436 on Build GRCh38
rs35296355 Chr.19: 43607417-43607418 on Build GRCh38
rs3052821 Chr.19: 43607395-43607396 on Build GRCh38
rs139385711 Chr.19: 43607373-43607378 on Build GRCh38
rs910407278 Chr.19: 43607146-43607146 on Build GRCh38
rs878860665 Chr.19: 43607372-43607373 on Build GRCh38
rs370373478 Chr.19: 43607438-43607438 on Build GRCh38
rs967758651 Chr.19: 43607394-43607394 on Build GRCh38
rs896997093 Chr.19: 43607273-43607273 on Build GRCh38
rs933038659 Chr.19: 43607123-43607123 on Build GRCh38
rs764214599 Chr.19: 43607396-43607396 on Build GRCh38
rs748097548 Chr.19: 43607514-43607514 on Build GRCh38
rs769573679 Chr.19: 43607515-43607515 on Build GRCh38
rs1052829422 Chr.19: 43607353-43607353 on Build GRCh38
rs147172567 Chr.19: 43607424-43607425 on Build GRCh38
rs2005852 Chr.19: 43607411-43607411 on Build GRCh38
rs1049796953 Chr.19: 43607229-43607229 on Build GRCh38
rs201204234 Chr.19: 43607435-43607435 on Build GRCh38
rs1014111361 Chr.19: 43607358-43607358 on Build GRCh38
rs547262966 Chr.19: 43607200-43607200 on Build GRCh38
rs762383467 Chr.19: 43607528-43607528 on Build GRCh38
rs764064605 Chr.19: 43607417-43607420 on Build GRCh38
rs771756693 Chr.19: 43607419-43607420 on Build GRCh38
rs905395215 Chr.19: 43607373-43607384 on Build GRCh38
rs922593304 Chr.19: 43607176-43607176 on Build GRCh38
rs911290862 Chr.19: 43607235-43607235 on Build GRCh38
rs545686674 Chr.19: 43607455-43607455 on Build GRCh38
rs1011868022 Chr.19: 43607355-43607355 on Build GRCh38
rs1038861849 Chr.19: 43607519-43607519 on Build GRCh38

View Full Product Details