Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119468420-119468875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A5
Entrez Gene ID
292
Gene Name
solute carrier family 25 member 5
Gene Aliases
2F1, AAC2, ANT2, T2, T3
UniGene
Hs.632282
Gene Chromosome Location
Chr.X: 119468400-119471396 on Build GRCh38
Transcripts
NM_001152.4 NR_028443.1 NR_134914.1

Gene Symbol
SLC25A5-AS1
Entrez Gene ID
100303728
Gene Name
SLC25A5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.714405
Gene Chromosome Location
Chr.X: 119466033-119469120 on Build GRCh38
Transcripts
NM_001152.4 NR_028443.1 NR_134914.1

SNP Detail

SNP ID Chromosome Location
rs759855460 Chr.X: 119468519-119468519 on Build GRCh38
rs753474355 Chr.X: 119468479-119468479 on Build GRCh38
rs962834075 Chr.X: 119468689-119468689 on Build GRCh38
rs200356671 Chr.X: 119468468-119468468 on Build GRCh38
rs760888044 Chr.X: 119468467-119468467 on Build GRCh38
rs754654611 Chr.X: 119468638-119468638 on Build GRCh38
rs755824654 Chr.X: 119468455-119468455 on Build GRCh38
rs770907853 Chr.X: 119468489-119468489 on Build GRCh38
rs758656296 Chr.X: 119468671-119468671 on Build GRCh38
rs771229485 Chr.X: 119468471-119468471 on Build GRCh38
rs772653432 Chr.X: 119468484-119468484 on Build GRCh38
rs986954695 Chr.X: 119468841-119468841 on Build GRCh38
rs1024649989 Chr.X: 119468726-119468726 on Build GRCh38
rs777616981 Chr.X: 119468724-119468724 on Build GRCh38
rs761468080 Chr.X: 119468533-119468533 on Build GRCh38
rs764819800 Chr.X: 119468640-119468640 on Build GRCh38
rs1020481199 Chr.X: 119468809-119468809 on Build GRCh38
rs767319955 Chr.X: 119468476-119468476 on Build GRCh38
rs770077050 Chr.X: 119468527-119468527 on Build GRCh38
rs750889118 Chr.X: 119468500-119468500 on Build GRCh38
rs779454023 Chr.X: 119468466-119468466 on Build GRCh38
rs776683368 Chr.X: 119468472-119468472 on Build GRCh38
rs761817456 Chr.X: 119468474-119468474 on Build GRCh38
rs751935289 Chr.X: 119468664-119468664 on Build GRCh38
rs772285357 Chr.X: 119468465-119468466 on Build GRCh38
rs370450611 Chr.X: 119468620-119468620 on Build GRCh38
rs1047166757 Chr.X: 119468535-119468535 on Build GRCh38
rs774321900 Chr.X: 119468530-119468530 on Build GRCh38
rs911479328 Chr.X: 119468844-119468844 on Build GRCh38
rs773789290 Chr.X: 119468754-119468754 on Build GRCh38
rs773581260 Chr.X: 119468502-119468504 on Build GRCh38
rs761933349 Chr.X: 119468478-119468478 on Build GRCh38
rs747336191 Chr.X: 119468764-119468764 on Build GRCh38
rs912978623 Chr.X: 119468797-119468797 on Build GRCh38
rs975531661 Chr.X: 119468704-119468704 on Build GRCh38
rs763380534 Chr.X: 119468622-119468622 on Build GRCh38
rs11552294 Chr.X: 119468543-119468543 on Build GRCh38
rs753367168 Chr.X: 119468732-119468732 on Build GRCh38
rs756788147 Chr.X: 119468762-119468762 on Build GRCh38
rs779252086 Chr.X: 119468481-119468481 on Build GRCh38
rs754566791 Chr.X: 119468480-119468480 on Build GRCh38
rs755557887 Chr.X: 119468477-119468477 on Build GRCh38
rs957019311 Chr.X: 119468768-119468768 on Build GRCh38
rs778309600 Chr.X: 119468838-119468838 on Build GRCh38
rs969971767 Chr.X: 119468757-119468757 on Build GRCh38
rs763972216 Chr.X: 119468473-119468473 on Build GRCh38
rs760300531 Chr.X: 119468592-119468592 on Build GRCh38
rs766696402 Chr.X: 119468637-119468637 on Build GRCh38
rs749703747 Chr.X: 119468461-119468461 on Build GRCh38
rs765328027 Chr.X: 119468485-119468485 on Build GRCh38
rs955780186 Chr.X: 119468761-119468761 on Build GRCh38
rs767542233 Chr.X: 119468564-119468564 on Build GRCh38
rs979493153 Chr.X: 119468813-119468813 on Build GRCh38
rs988552385 Chr.X: 119468787-119468787 on Build GRCh38
rs1005920056 Chr.X: 119468502-119468502 on Build GRCh38
rs750296457 Chr.X: 119468575-119468575 on Build GRCh38
rs748470956 Chr.X: 119468483-119468483 on Build GRCh38
rs1013510074 Chr.X: 119468469-119468469 on Build GRCh38
rs755813427 Chr.X: 119468675-119468675 on Build GRCh38
rs1028503856 Chr.X: 119468760-119468760 on Build GRCh38

View Full Product Details