Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153782850-153783405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRPK3
Entrez Gene ID
26576
Gene Name
SRSF protein kinase 3
Gene Aliases
MSSK-1, MSSK1, STK23
UniGene
Hs.104865
Gene Chromosome Location
Chr.X: 153781001-153785732 on Build GRCh38
Transcripts
NM_001170760.1 NM_001170761.1 NM_014370.3

SNP Detail

SNP ID Chromosome Location
rs999559599 Chr.X: 153783305-153783305 on Build GRCh38
rs375593729 Chr.X: 153783327-153783327 on Build GRCh38
rs782698225 Chr.X: 153783183-153783183 on Build GRCh38
rs142432448 Chr.X: 153783037-153783037 on Build GRCh38
rs782343428 Chr.X: 153783143-153783143 on Build GRCh38
rs782261773 Chr.X: 153783195-153783195 on Build GRCh38
rs782267049 Chr.X: 153783207-153783207 on Build GRCh38
rs375627608 Chr.X: 153783110-153783110 on Build GRCh38
rs782379799 Chr.X: 153783238-153783238 on Build GRCh38
rs148799822 Chr.X: 153783033-153783033 on Build GRCh38
rs893585285 Chr.X: 153783348-153783348 on Build GRCh38
rs782541167 Chr.X: 153783212-153783212 on Build GRCh38
rs782043348 Chr.X: 153783104-153783104 on Build GRCh38
rs782144135 Chr.X: 153783095-153783095 on Build GRCh38
rs782431998 Chr.X: 153783069-153783069 on Build GRCh38
rs781801643 Chr.X: 153783028-153783028 on Build GRCh38
rs372400669 Chr.X: 153783200-153783200 on Build GRCh38
rs781987227 Chr.X: 153782941-153782941 on Build GRCh38
rs782754940 Chr.X: 153783030-153783030 on Build GRCh38
rs782184192 Chr.X: 153783043-153783043 on Build GRCh38
rs782012170 Chr.X: 153783199-153783200 on Build GRCh38
rs111731456 Chr.X: 153782897-153782897 on Build GRCh38
rs782268437 Chr.X: 153783121-153783121 on Build GRCh38
rs782437305 Chr.X: 153782945-153782946 on Build GRCh38
rs782586952 Chr.X: 153783215-153783215 on Build GRCh38
rs782314180 Chr.X: 153783129-153783129 on Build GRCh38
rs909924547 Chr.X: 153783154-153783154 on Build GRCh38
rs781928586 Chr.X: 153783066-153783066 on Build GRCh38
rs371637307 Chr.X: 153783029-153783029 on Build GRCh38
rs150937454 Chr.X: 153783079-153783079 on Build GRCh38
rs781926477 Chr.X: 153783197-153783197 on Build GRCh38
rs782668508 Chr.X: 153783114-153783114 on Build GRCh38
rs782176533 Chr.X: 153783216-153783216 on Build GRCh38
rs782809092 Chr.X: 153783096-153783096 on Build GRCh38
rs191542808 Chr.X: 153783302-153783302 on Build GRCh38
rs782784289 Chr.X: 153783188-153783188 on Build GRCh38
rs34497419 Chr.X: 153783067-153783067 on Build GRCh38
rs782182661 Chr.X: 153782926-153782926 on Build GRCh38
rs781835413 Chr.X: 153783193-153783194 on Build GRCh38
rs367934508 Chr.X: 153783075-153783075 on Build GRCh38
rs781982564 Chr.X: 153782997-153782997 on Build GRCh38
rs1051403667 Chr.X: 153783053-153783053 on Build GRCh38
rs781851056 Chr.X: 153783192-153783193 on Build GRCh38
rs782166144 Chr.X: 153783003-153783003 on Build GRCh38
rs782304485 Chr.X: 153783263-153783263 on Build GRCh38
rs190774414 Chr.X: 153782870-153782870 on Build GRCh38
rs1010637294 Chr.X: 153783371-153783371 on Build GRCh38
rs782013944 Chr.X: 153783244-153783244 on Build GRCh38
rs782600233 Chr.X: 153783218-153783218 on Build GRCh38
rs782085059 Chr.X: 153782953-153782953 on Build GRCh38
rs782333088 Chr.X: 153782940-153782940 on Build GRCh38
rs941448763 Chr.X: 153783229-153783229 on Build GRCh38
rs951433728 Chr.X: 153783134-153783134 on Build GRCh38
rs781858574 Chr.X: 153783092-153783092 on Build GRCh38
rs782774387 Chr.X: 153783090-153783090 on Build GRCh38
rs781895348 Chr.X: 153783201-153783201 on Build GRCh38
rs916001 Chr.X: 153782948-153782948 on Build GRCh38
rs782799190 Chr.X: 153783283-153783283 on Build GRCh38
rs782754230 Chr.X: 153783203-153783203 on Build GRCh38
rs201877372 Chr.X: 153783202-153783202 on Build GRCh38
rs781836442 Chr.X: 153783259-153783259 on Build GRCh38
rs910067337 Chr.X: 153782968-153782968 on Build GRCh38
rs782362977 Chr.X: 153783056-153783056 on Build GRCh38
rs782185180 Chr.X: 153783349-153783349 on Build GRCh38
rs782264909 Chr.X: 153783220-153783220 on Build GRCh38
rs782084667 Chr.X: 153783078-153783078 on Build GRCh38
rs782157672 Chr.X: 153783280-153783280 on Build GRCh38
rs782381724 Chr.X: 153782930-153782930 on Build GRCh38
rs782007485 Chr.X: 153783036-153783036 on Build GRCh38
rs367826190 Chr.X: 153783006-153783006 on Build GRCh38
rs781981394 Chr.X: 153783279-153783279 on Build GRCh38
rs782309018 Chr.X: 153783196-153783196 on Build GRCh38
rs146121291 Chr.X: 153782985-153782985 on Build GRCh38
rs375284895 Chr.X: 153783055-153783055 on Build GRCh38
rs35757683 Chr.X: 153783193-153783193 on Build GRCh38
rs782088949 Chr.X: 153783202-153783203 on Build GRCh38
rs370996947 Chr.X: 153783199-153783199 on Build GRCh38
rs782016592 Chr.X: 153783182-153783182 on Build GRCh38
rs782475051 Chr.X: 153782917-153782917 on Build GRCh38
rs782649043 Chr.X: 153783205-153783205 on Build GRCh38
rs782192075 Chr.X: 153783261-153783261 on Build GRCh38
rs782598828 Chr.X: 153783194-153783194 on Build GRCh38
rs782791740 Chr.X: 153783268-153783268 on Build GRCh38
rs781830949 Chr.X: 153783192-153783192 on Build GRCh38
rs201302475 Chr.X: 153783193-153783193 on Build GRCh38
rs782436215 Chr.X: 153783180-153783180 on Build GRCh38
rs782586390 Chr.X: 153783039-153783039 on Build GRCh38
rs782231600 Chr.X: 153783141-153783141 on Build GRCh38
rs111574154 Chr.X: 153783127-153783127 on Build GRCh38
rs782101711 Chr.X: 153782961-153782961 on Build GRCh38
rs781938511 Chr.X: 153783087-153783087 on Build GRCh38
rs2314343 Chr.X: 153783202-153783202 on Build GRCh38
rs782569380 Chr.X: 153783138-153783138 on Build GRCh38
rs781811544 Chr.X: 153783291-153783291 on Build GRCh38
rs782523802 Chr.X: 153783190-153783190 on Build GRCh38
rs186688595 Chr.X: 153783198-153783198 on Build GRCh38
rs371188111 Chr.X: 153782886-153782886 on Build GRCh38
rs782042839 Chr.X: 153782937-153782937 on Build GRCh38
rs782613591 Chr.X: 153782918-153782918 on Build GRCh38
rs781903088 Chr.X: 153783189-153783189 on Build GRCh38
rs782737721 Chr.X: 153783204-153783204 on Build GRCh38
rs782626428 Chr.X: 153783109-153783109 on Build GRCh38
rs781849828 Chr.X: 153783187-153783187 on Build GRCh38
rs782534269 Chr.X: 153783035-153783035 on Build GRCh38
rs941485247 Chr.X: 153782982-153782982 on Build GRCh38
rs782798674 Chr.X: 153783277-153783277 on Build GRCh38

View Full Product Details