Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35910057-35910638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPSL
Entrez Gene ID
133690
Gene Name
calcyphosine like
Gene Aliases
-
UniGene
Hs.55150
Gene Chromosome Location
Chr.5: 35904295-35938779 on Build GRCh38
Transcripts
NM_001042625.1 NM_144647.3 XM_006714444.3 XM_006714445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4537709 c.244G>A p.E82K Chr.5: 35910437-35910437 on Build GRCh38
4895750 c.195G>A p.M65I Chr.5: 35910486-35910486 on Build GRCh38
2687305 c.226G>A p.E76K Chr.5: 35910455-35910455 on Build GRCh38
3615798 c.200G>A p.G67E Chr.5: 35910481-35910481 on Build GRCh38
6103789 c.178G>T p.D60Y Chr.5: 35910503-35910503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759379022 Chr.5: 35910503-35910503 on Build GRCh38
rs116630600 Chr.5: 35910206-35910206 on Build GRCh38
rs781449697 Chr.5: 35910545-35910546 on Build GRCh38
rs552359343 Chr.5: 35910253-35910253 on Build GRCh38
rs748338288 Chr.5: 35910092-35910092 on Build GRCh38
rs772992939 Chr.5: 35910418-35910418 on Build GRCh38
rs868067758 Chr.5: 35910410-35910410 on Build GRCh38
rs769934304 Chr.5: 35910566-35910566 on Build GRCh38
rs760433308 Chr.5: 35910425-35910425 on Build GRCh38
rs747326182 Chr.5: 35910366-35910366 on Build GRCh38
rs200428487 Chr.5: 35910467-35910467 on Build GRCh38
rs201880660 Chr.5: 35910388-35910388 on Build GRCh38
rs1043311808 Chr.5: 35910182-35910182 on Build GRCh38
rs746325313 Chr.5: 35910385-35910385 on Build GRCh38
rs758559698 Chr.5: 35910363-35910363 on Build GRCh38
rs199837654 Chr.5: 35910511-35910511 on Build GRCh38
rs560300537 Chr.5: 35910230-35910230 on Build GRCh38
rs1041527693 Chr.5: 35910310-35910310 on Build GRCh38
rs771781791 Chr.5: 35910408-35910408 on Build GRCh38
rs754395303 Chr.5: 35910196-35910196 on Build GRCh38
rs149545789 Chr.5: 35910296-35910296 on Build GRCh38
rs767296914 Chr.5: 35910316-35910316 on Build GRCh38
rs1044848933 Chr.5: 35910138-35910138 on Build GRCh38
rs768033968 Chr.5: 35910122-35910122 on Build GRCh38
rs746611359 Chr.5: 35910493-35910493 on Build GRCh38
rs773445907 Chr.5: 35910593-35910594 on Build GRCh38
rs1043050074 Chr.5: 35910413-35910413 on Build GRCh38
rs748521578 Chr.5: 35910556-35910556 on Build GRCh38
rs773472513 Chr.5: 35910583-35910583 on Build GRCh38
rs779107309 Chr.5: 35910085-35910085 on Build GRCh38
rs111414827 Chr.5: 35910489-35910489 on Build GRCh38
rs904752205 Chr.5: 35910194-35910194 on Build GRCh38
rs767464889 Chr.5: 35910189-35910189 on Build GRCh38
rs1445898 Chr.5: 35910427-35910427 on Build GRCh38
rs761513771 Chr.5: 35910315-35910315 on Build GRCh38
rs372265672 Chr.5: 35910152-35910152 on Build GRCh38
rs374905244 Chr.5: 35910492-35910492 on Build GRCh38
rs756634622 Chr.5: 35910472-35910472 on Build GRCh38
rs751705913 Chr.5: 35910462-35910462 on Build GRCh38
rs754992263 Chr.5: 35910337-35910337 on Build GRCh38
rs1036906202 Chr.5: 35910607-35910608 on Build GRCh38
rs563280961 Chr.5: 35910251-35910251 on Build GRCh38
rs112546964 Chr.5: 35910608-35910608 on Build GRCh38
rs568162790 Chr.5: 35910319-35910319 on Build GRCh38
rs757552496 Chr.5: 35910372-35910372 on Build GRCh38
rs778514974 Chr.5: 35910393-35910393 on Build GRCh38
rs75736388 Chr.5: 35910522-35910522 on Build GRCh38
rs769552588 Chr.5: 35910560-35910560 on Build GRCh38
rs747567733 Chr.5: 35910512-35910513 on Build GRCh38
rs372030340 Chr.5: 35910350-35910350 on Build GRCh38
rs778063988 Chr.5: 35910097-35910097 on Build GRCh38
rs529029610 Chr.5: 35910200-35910200 on Build GRCh38
rs780714295 Chr.5: 35910480-35910480 on Build GRCh38
rs766095343 Chr.5: 35910110-35910110 on Build GRCh38
rs575166416 Chr.5: 35910186-35910186 on Build GRCh38
rs898000622 Chr.5: 35910608-35910608 on Build GRCh38
rs1008644150 Chr.5: 35910244-35910244 on Build GRCh38
rs991533594 Chr.5: 35910287-35910287 on Build GRCh38
rs765172267 Chr.5: 35910442-35910442 on Build GRCh38
rs1007193277 Chr.5: 35910397-35910397 on Build GRCh38
rs769642485 Chr.5: 35910506-35910506 on Build GRCh38
rs774665279 Chr.5: 35910086-35910088 on Build GRCh38
rs904529862 Chr.5: 35910415-35910415 on Build GRCh38
rs753317475 Chr.5: 35910317-35910318 on Build GRCh38
rs369979397 Chr.5: 35910428-35910428 on Build GRCh38
rs752797164 Chr.5: 35910354-35910354 on Build GRCh38
rs76988839 Chr.5: 35910166-35910166 on Build GRCh38
rs114583055 Chr.5: 35910500-35910500 on Build GRCh38
rs201588644 Chr.5: 35910502-35910502 on Build GRCh38
rs761895563 Chr.5: 35910513-35910513 on Build GRCh38
rs946057717 Chr.5: 35910169-35910169 on Build GRCh38
rs761029049 Chr.5: 35910587-35910587 on Build GRCh38
rs374400961 Chr.5: 35910514-35910514 on Build GRCh38
rs368384088 Chr.5: 35910320-35910320 on Build GRCh38
rs749878381 Chr.5: 35910482-35910482 on Build GRCh38
rs138096862 Chr.5: 35910167-35910167 on Build GRCh38
rs1026329917 Chr.5: 35910170-35910170 on Build GRCh38
rs539642491 Chr.5: 35910488-35910488 on Build GRCh38
rs139601439 Chr.5: 35910507-35910507 on Build GRCh38
rs951639923 Chr.5: 35910190-35910190 on Build GRCh38
rs750611783 Chr.5: 35910301-35910301 on Build GRCh38
rs935963087 Chr.5: 35910336-35910336 on Build GRCh38
rs867406716 Chr.5: 35910212-35910212 on Build GRCh38
rs375119792 Chr.5: 35910399-35910399 on Build GRCh38
rs944993624 Chr.5: 35910590-35910590 on Build GRCh38
rs145854210 Chr.5: 35910403-35910403 on Build GRCh38
rs938742108 Chr.5: 35910164-35910164 on Build GRCh38
rs376886601 Chr.5: 35910545-35910545 on Build GRCh38
rs781376073 Chr.5: 35910469-35910469 on Build GRCh38
rs780552356 Chr.5: 35910470-35910470 on Build GRCh38
rs758650769 Chr.5: 35910096-35910096 on Build GRCh38
rs557706097 Chr.5: 35910573-35910573 on Build GRCh38
rs777462789 Chr.5: 35910316-35910317 on Build GRCh38
rs748659530 Chr.5: 35910527-35910527 on Build GRCh38
rs751398372 Chr.5: 35910123-35910123 on Build GRCh38
rs769146942 Chr.5: 35910519-35910521 on Build GRCh38
rs759342773 Chr.5: 35910431-35910431 on Build GRCh38
rs747389173 Chr.5: 35910098-35910098 on Build GRCh38
rs202156612 Chr.5: 35910512-35910512 on Build GRCh38
rs920537681 Chr.5: 35910103-35910103 on Build GRCh38
rs200956929 Chr.5: 35910114-35910114 on Build GRCh38
rs750689295 Chr.5: 35910519-35910519 on Build GRCh38
rs750825605 Chr.5: 35910471-35910471 on Build GRCh38
rs372581109 Chr.5: 35910161-35910161 on Build GRCh38
rs756689125 Chr.5: 35910450-35910450 on Build GRCh38
rs575353255 Chr.5: 35910521-35910521 on Build GRCh38
rs1445899 Chr.5: 35910317-35910317 on Build GRCh38
rs774240756 Chr.5: 35910577-35910577 on Build GRCh38
rs771405172 Chr.5: 35910101-35910101 on Build GRCh38
rs868106190 Chr.5: 35910380-35910380 on Build GRCh38
rs746541152 Chr.5: 35910542-35910542 on Build GRCh38
rs777099097 Chr.5: 35910531-35910531 on Build GRCh38
rs370102888 Chr.5: 35910449-35910449 on Build GRCh38
rs148259091 Chr.5: 35910581-35910581 on Build GRCh38
rs373561910 Chr.5: 35910591-35910591 on Build GRCh38
rs890617341 Chr.5: 35910443-35910443 on Build GRCh38
rs764212974 Chr.5: 35910460-35910460 on Build GRCh38
rs961673031 Chr.5: 35910256-35910256 on Build GRCh38
rs765262166 Chr.5: 35910342-35910342 on Build GRCh38
rs370482916 Chr.5: 35910568-35910568 on Build GRCh38
rs781660425 Chr.5: 35910378-35910378 on Build GRCh38
rs539362676 Chr.5: 35910357-35910357 on Build GRCh38
rs889001675 Chr.5: 35910579-35910579 on Build GRCh38
rs144015948 Chr.5: 35910298-35910298 on Build GRCh38
rs763014353 Chr.5: 35910459-35910459 on Build GRCh38
rs753705142 Chr.5: 35910332-35910332 on Build GRCh38
rs2287899 Chr.5: 35910235-35910235 on Build GRCh38
rs745575321 Chr.5: 35910549-35910549 on Build GRCh38

View Full Product Details