Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30562869-30563468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR3
Entrez Gene ID
80742
Gene Name
proline rich 3
Gene Aliases
CAT56
UniGene
Hs.118354
Gene Chromosome Location
Chr.6: 30556709-30564696 on Build GRCh38
Transcripts
NM_001077497.2 NM_025263.3

SNP Detail

SNP ID Chromosome Location
rs1026931996 Chr.6: 30563105-30563105 on Build GRCh38
rs913733946 Chr.6: 30562972-30562972 on Build GRCh38
rs878974835 Chr.6: 30563264-30563264 on Build GRCh38
rs1051620894 Chr.6: 30563030-30563030 on Build GRCh38
rs1019512158 Chr.6: 30563315-30563315 on Build GRCh38
rs544637061 Chr.6: 30562935-30562935 on Build GRCh38
rs1023241519 Chr.6: 30563163-30563163 on Build GRCh38
rs188502980 Chr.6: 30563172-30563172 on Build GRCh38
rs377388910 Chr.6: 30563404-30563404 on Build GRCh38
rs777161256 Chr.6: 30563120-30563121 on Build GRCh38
rs750879446 Chr.6: 30563125-30563125 on Build GRCh38
rs550414958 Chr.6: 30563131-30563131 on Build GRCh38
rs912505851 Chr.6: 30563016-30563016 on Build GRCh38
rs1044611199 Chr.6: 30563130-30563130 on Build GRCh38
rs77480457 Chr.6: 30562930-30562930 on Build GRCh38
rs754580655 Chr.6: 30563191-30563191 on Build GRCh38
rs375637350 Chr.6: 30563210-30563210 on Build GRCh38
rs978732155 Chr.6: 30562928-30562928 on Build GRCh38
rs530015695 Chr.6: 30563380-30563380 on Build GRCh38
rs549308499 Chr.6: 30563211-30563211 on Build GRCh38
rs943843636 Chr.6: 30563036-30563036 on Build GRCh38
rs933152113 Chr.6: 30563333-30563333 on Build GRCh38
rs184215395 Chr.6: 30563152-30563152 on Build GRCh38
rs766144755 Chr.6: 30563095-30563095 on Build GRCh38
rs778069971 Chr.6: 30563408-30563408 on Build GRCh38
rs753293201 Chr.6: 30563314-30563314 on Build GRCh38
rs551829035 Chr.6: 30563381-30563381 on Build GRCh38
rs965329630 Chr.6: 30563316-30563316 on Build GRCh38
rs1052700110 Chr.6: 30562937-30562937 on Build GRCh38
rs898918557 Chr.6: 30563087-30563087 on Build GRCh38
rs1021753801 Chr.6: 30563236-30563236 on Build GRCh38
rs767897020 Chr.6: 30563244-30563244 on Build GRCh38
rs894804709 Chr.6: 30563110-30563110 on Build GRCh38
rs1048171214 Chr.6: 30563108-30563108 on Build GRCh38
rs1032149603 Chr.6: 30562910-30562910 on Build GRCh38
rs993222798 Chr.6: 30563097-30563097 on Build GRCh38
rs895971181 Chr.6: 30563151-30563151 on Build GRCh38
rs1037300981 Chr.6: 30563083-30563083 on Build GRCh38
rs983822023 Chr.6: 30563429-30563429 on Build GRCh38
rs954067758 Chr.6: 30562996-30562996 on Build GRCh38
rs541127794 Chr.6: 30562914-30562914 on Build GRCh38
rs933269409 Chr.6: 30563000-30563000 on Build GRCh38
rs761494717 Chr.6: 30562890-30562890 on Build GRCh38
rs533477333 Chr.6: 30563125-30563125 on Build GRCh38
rs564516400 Chr.6: 30563039-30563039 on Build GRCh38
rs140180437 Chr.6: 30563307-30563307 on Build GRCh38
rs972938646 Chr.6: 30563355-30563355 on Build GRCh38
rs1003068344 Chr.6: 30562901-30562905 on Build GRCh38
rs1031849681 Chr.6: 30563205-30563205 on Build GRCh38
rs888807522 Chr.6: 30563051-30563051 on Build GRCh38
rs972603522 Chr.6: 30563042-30563042 on Build GRCh38
rs943256340 Chr.6: 30563412-30563412 on Build GRCh38
rs762647598 Chr.6: 30563074-30563074 on Build GRCh38
rs913756178 Chr.6: 30563326-30563326 on Build GRCh38
rs931078876 Chr.6: 30563090-30563093 on Build GRCh38
rs773172660 Chr.6: 30562948-30562948 on Build GRCh38

View Full Product Details