Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169710090-169710639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF10
Entrez Gene ID
55274
Gene Name
PHD finger protein 10
Gene Aliases
BAF45A, XAP135
UniGene
Hs.435933
Gene Chromosome Location
Chr.6: 169703902-169724049 on Build GRCh38
Transcripts
NM_018288.3 NM_133325.2 XM_017010998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379056 c.1072C>G p.P358A Chr.6: 169710277-169710277 on Build GRCh38
1076154 c.752C>A p.S251Y Chr.6: 169710333-169710333 on Build GRCh38
7383717 c.809C>A p.P270H Chr.6: 169710276-169710276 on Build GRCh38
6453633 c.1057C>G p.P353A Chr.6: 169710292-169710292 on Build GRCh38
3430348 c.797G>T p.R266I Chr.6: 169710288-169710288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17860601 Chr.6: 169710308-169710308 on Build GRCh38
rs769470895 Chr.6: 169710408-169710408 on Build GRCh38
rs763902057 Chr.6: 169710368-169710368 on Build GRCh38
rs752491406 Chr.6: 169710229-169710229 on Build GRCh38
rs1021067361 Chr.6: 169710509-169710509 on Build GRCh38
rs758209211 Chr.6: 169710237-169710237 on Build GRCh38
rs200565755 Chr.6: 169710407-169710407 on Build GRCh38
rs775760779 Chr.6: 169710195-169710195 on Build GRCh38
rs1043491834 Chr.6: 169710172-169710172 on Build GRCh38
rs763357370 Chr.6: 169710198-169710198 on Build GRCh38
rs139332707 Chr.6: 169710383-169710383 on Build GRCh38
rs372584680 Chr.6: 169710284-169710284 on Build GRCh38
rs770318155 Chr.6: 169710194-169710194 on Build GRCh38
rs918057390 Chr.6: 169710570-169710570 on Build GRCh38
rs749617420 Chr.6: 169710292-169710292 on Build GRCh38
rs754027134 Chr.6: 169710333-169710333 on Build GRCh38
rs763787552 Chr.6: 169710585-169710585 on Build GRCh38
rs966154205 Chr.6: 169710139-169710139 on Build GRCh38
rs776437450 Chr.6: 169710181-169710181 on Build GRCh38
rs561110831 Chr.6: 169710135-169710135 on Build GRCh38
rs560089180 Chr.6: 169710562-169710562 on Build GRCh38
rs542935874 Chr.6: 169710126-169710126 on Build GRCh38
rs976060892 Chr.6: 169710140-169710140 on Build GRCh38
rs928590051 Chr.6: 169710465-169710465 on Build GRCh38
rs370388298 Chr.6: 169710203-169710203 on Build GRCh38
rs770499945 Chr.6: 169710178-169710178 on Build GRCh38
rs773327899 Chr.6: 169710319-169710319 on Build GRCh38
rs1055735341 Chr.6: 169710214-169710214 on Build GRCh38
rs140841254 Chr.6: 169710608-169710608 on Build GRCh38
rs768929404 Chr.6: 169710424-169710424 on Build GRCh38
rs377113287 Chr.6: 169710431-169710431 on Build GRCh38
rs949573842 Chr.6: 169710579-169710579 on Build GRCh38
rs766656353 Chr.6: 169710332-169710332 on Build GRCh38
rs762854990 Chr.6: 169710502-169710502 on Build GRCh38
rs41265397 Chr.6: 169710474-169710474 on Build GRCh38
rs761807692 Chr.6: 169710208-169710208 on Build GRCh38
rs777578804 Chr.6: 169710262-169710262 on Build GRCh38
rs913811548 Chr.6: 169710540-169710540 on Build GRCh38
rs759219351 Chr.6: 169710186-169710186 on Build GRCh38
rs545910471 Chr.6: 169710405-169710405 on Build GRCh38
rs550938421 Chr.6: 169710561-169710565 on Build GRCh38
rs541608244 Chr.6: 169710543-169710543 on Build GRCh38
rs750673985 Chr.6: 169710215-169710215 on Build GRCh38
rs564009326 Chr.6: 169710429-169710429 on Build GRCh38
rs767981274 Chr.6: 169710317-169710317 on Build GRCh38
rs775201631 Chr.6: 169710286-169710286 on Build GRCh38
rs34227712 Chr.6: 169710265-169710265 on Build GRCh38
rs773172683 Chr.6: 169710146-169710146 on Build GRCh38
rs767533165 Chr.6: 169710212-169710212 on Build GRCh38
rs988055758 Chr.6: 169710514-169710514 on Build GRCh38
rs889737423 Chr.6: 169710335-169710336 on Build GRCh38
rs1017696399 Chr.6: 169710138-169710138 on Build GRCh38
rs757304668 Chr.6: 169710375-169710375 on Build GRCh38
rs780812700 Chr.6: 169710270-169710270 on Build GRCh38
rs766520439 Chr.6: 169710268-169710268 on Build GRCh38
rs774738568 Chr.6: 169710302-169710302 on Build GRCh38
rs12663375 Chr.6: 169710147-169710147 on Build GRCh38
rs528670519 Chr.6: 169710160-169710160 on Build GRCh38
rs780225800 Chr.6: 169710226-169710226 on Build GRCh38
rs917648001 Chr.6: 169710247-169710247 on Build GRCh38
rs528686786 Chr.6: 169710161-169710161 on Build GRCh38
rs953866447 Chr.6: 169710137-169710137 on Build GRCh38
rs764572248 Chr.6: 169710199-169710199 on Build GRCh38
rs966695982 Chr.6: 169710527-169710527 on Build GRCh38
rs769002632 Chr.6: 169710301-169710301 on Build GRCh38
rs962911351 Chr.6: 169710337-169710337 on Build GRCh38
rs760729295 Chr.6: 169710329-169710329 on Build GRCh38
rs756368426 Chr.6: 169710222-169710222 on Build GRCh38
rs774817715 Chr.6: 169710427-169710427 on Build GRCh38
rs932367319 Chr.6: 169710601-169710601 on Build GRCh38
rs1050791691 Chr.6: 169710610-169710610 on Build GRCh38
rs758141321 Chr.6: 169710340-169710340 on Build GRCh38
rs981478328 Chr.6: 169710449-169710449 on Build GRCh38
rs879040257 Chr.6: 169710462-169710462 on Build GRCh38
rs565377149 Chr.6: 169710400-169710400 on Build GRCh38
rs141680830 Chr.6: 169710276-169710276 on Build GRCh38
rs995408607 Chr.6: 169710116-169710116 on Build GRCh38
rs751486851 Chr.6: 169710371-169710371 on Build GRCh38
rs927920193 Chr.6: 169710130-169710130 on Build GRCh38
rs749511745 Chr.6: 169710113-169710113 on Build GRCh38
rs184410055 Chr.6: 169710542-169710542 on Build GRCh38
rs979848816 Chr.6: 169710530-169710530 on Build GRCh38

View Full Product Details