Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143495315-143495702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA2
Entrez Gene ID
2519
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
dJ20N2.5
UniGene
Hs.17680
Gene Chromosome Location
Chr.6: 143494812-143511883 on Build GRCh38
Transcripts
NM_032020.4

SNP Detail

SNP ID Chromosome Location
rs78619052 Chr.6: 143495349-143495349 on Build GRCh38
rs77488485 Chr.6: 143495365-143495365 on Build GRCh38
rs968061445 Chr.6: 143495616-143495616 on Build GRCh38
rs764222848 Chr.6: 143495657-143495657 on Build GRCh38
rs867988460 Chr.6: 143495541-143495541 on Build GRCh38
rs937082665 Chr.6: 143495368-143495368 on Build GRCh38
rs899858602 Chr.6: 143495652-143495652 on Build GRCh38
rs998037121 Chr.6: 143495499-143495499 on Build GRCh38
rs888116107 Chr.6: 143495377-143495377 on Build GRCh38
rs761751116 Chr.6: 143495676-143495676 on Build GRCh38
rs748194104 Chr.6: 143495554-143495554 on Build GRCh38
rs903061440 Chr.6: 143495617-143495617 on Build GRCh38
rs1022759356 Chr.6: 143495560-143495560 on Build GRCh38
rs993843508 Chr.6: 143495350-143495350 on Build GRCh38
rs532708504 Chr.6: 143495632-143495632 on Build GRCh38
rs565235391 Chr.6: 143495580-143495580 on Build GRCh38
rs779068640 Chr.6: 143495411-143495411 on Build GRCh38
rs899687262 Chr.6: 143495483-143495483 on Build GRCh38
rs1032990441 Chr.6: 143495633-143495633 on Build GRCh38
rs1041631267 Chr.6: 143495570-143495570 on Build GRCh38
rs561448948 Chr.6: 143495442-143495442 on Build GRCh38
rs1029474839 Chr.6: 143495501-143495501 on Build GRCh38
rs1046716728 Chr.6: 143495370-143495370 on Build GRCh38
rs1005917415 Chr.6: 143495449-143495450 on Build GRCh38
rs753845716 Chr.6: 143495659-143495659 on Build GRCh38
rs895637141 Chr.6: 143495412-143495412 on Build GRCh38
rs559545985 Chr.6: 143495348-143495348 on Build GRCh38
rs1018179676 Chr.6: 143495480-143495480 on Build GRCh38
rs550176586 Chr.6: 143495517-143495517 on Build GRCh38
rs531589921 Chr.6: 143495506-143495506 on Build GRCh38
rs956787113 Chr.6: 143495502-143495503 on Build GRCh38

View Full Product Details