Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26369893-26370435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R2A
Entrez Gene ID
5520
Gene Name
protein phosphatase 2 regulatory subunit Balpha
Gene Aliases
B55A, B55ALPHA, PR52A, PR55A
UniGene
Hs.146339
Gene Chromosome Location
Chr.8: 26291491-26372680 on Build GRCh38
Transcripts
NM_001177591.1 NM_002717.3 XM_005273559.1 XM_005273560.1 XM_011544565.1 XM_011544566.1 XM_011544567.2 XM_011544568.1 XM_011544569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919809 c.1129A>G p.I377V Chr.8: 26370198-26370198 on Build GRCh38
3647673 c.1078G>A p.G360R Chr.8: 26370147-26370147 on Build GRCh38
4996939 c.1135C>G p.L379V Chr.8: 26370204-26370204 on Build GRCh38
2787440 c.1166G>A p.R389H Chr.8: 26370235-26370235 on Build GRCh38
6837221 c.1277C>G p.P426R Chr.8: 26370346-26370346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748115278 Chr.8: 26370106-26370109 on Build GRCh38
rs1039033551 Chr.8: 26370323-26370323 on Build GRCh38
rs746820127 Chr.8: 26370314-26370314 on Build GRCh38
rs746785939 Chr.8: 26370204-26370205 on Build GRCh38
rs1050617560 Chr.8: 26370278-26370278 on Build GRCh38
rs369862992 Chr.8: 26370235-26370235 on Build GRCh38
rs991343051 Chr.8: 26370081-26370081 on Build GRCh38
rs567610910 Chr.8: 26370230-26370230 on Build GRCh38
rs149766175 Chr.8: 26370226-26370226 on Build GRCh38
rs372679014 Chr.8: 26370302-26370302 on Build GRCh38
rs754780649 Chr.8: 26370316-26370316 on Build GRCh38
rs979673760 Chr.8: 26370059-26370059 on Build GRCh38
rs908142106 Chr.8: 26370114-26370114 on Build GRCh38
rs763469557 Chr.8: 26370087-26370087 on Build GRCh38
rs755743652 Chr.8: 26370098-26370098 on Build GRCh38
rs771945967 Chr.8: 26370118-26370121 on Build GRCh38
rs3808565 Chr.8: 26370124-26370124 on Build GRCh38
rs1017734131 Chr.8: 26370014-26370014 on Build GRCh38
rs554122676 Chr.8: 26369977-26369977 on Build GRCh38
rs776881640 Chr.8: 26370157-26370157 on Build GRCh38
rs773446850 Chr.8: 26370365-26370365 on Build GRCh38
rs984728546 Chr.8: 26370113-26370113 on Build GRCh38
rs746773831 Chr.8: 26370146-26370146 on Build GRCh38
rs1045558100 Chr.8: 26369918-26369918 on Build GRCh38
rs777592803 Chr.8: 26370129-26370130 on Build GRCh38
rs1011664356 Chr.8: 26370291-26370291 on Build GRCh38
rs751891574 Chr.8: 26370091-26370091 on Build GRCh38
rs545155735 Chr.8: 26369964-26369964 on Build GRCh38
rs544366603 Chr.8: 26369969-26369969 on Build GRCh38
rs376876904 Chr.8: 26370270-26370270 on Build GRCh38
rs773357357 Chr.8: 26370212-26370212 on Build GRCh38
rs1019920773 Chr.8: 26370332-26370332 on Build GRCh38
rs764735324 Chr.8: 26370266-26370266 on Build GRCh38
rs748377977 Chr.8: 26370358-26370358 on Build GRCh38
rs753473897 Chr.8: 26370241-26370241 on Build GRCh38
rs190065785 Chr.8: 26369967-26369967 on Build GRCh38
rs3824232 Chr.8: 26370083-26370083 on Build GRCh38
rs184143190 Chr.8: 26370071-26370071 on Build GRCh38
rs1021097745 Chr.8: 26370049-26370049 on Build GRCh38
rs372782343 Chr.8: 26370252-26370252 on Build GRCh38
rs368687710 Chr.8: 26370403-26370403 on Build GRCh38
rs3200031 Chr.8: 26369968-26369968 on Build GRCh38
rs902850129 Chr.8: 26370366-26370366 on Build GRCh38
rs750742970 Chr.8: 26369988-26369988 on Build GRCh38
rs142234242 Chr.8: 26370072-26370072 on Build GRCh38
rs897761642 Chr.8: 26369924-26369924 on Build GRCh38
rs756735860 Chr.8: 26370108-26370108 on Build GRCh38
rs768792514 Chr.8: 26370155-26370155 on Build GRCh38
rs745804140 Chr.8: 26370120-26370120 on Build GRCh38
rs139723084 Chr.8: 26370203-26370203 on Build GRCh38
rs780005278 Chr.8: 26370308-26370308 on Build GRCh38
rs201037086 Chr.8: 26369925-26369925 on Build GRCh38
rs573326271 Chr.8: 26369944-26369944 on Build GRCh38
rs112876693 Chr.8: 26370179-26370179 on Build GRCh38
rs112640053 Chr.8: 26369997-26369997 on Build GRCh38
rs766656944 Chr.8: 26370090-26370090 on Build GRCh38
rs528225931 Chr.8: 26370066-26370066 on Build GRCh38
rs779731722 Chr.8: 26370139-26370139 on Build GRCh38
rs777438635 Chr.8: 26370100-26370100 on Build GRCh38
rs553365859 Chr.8: 26370355-26370355 on Build GRCh38
rs966102806 Chr.8: 26370023-26370023 on Build GRCh38
rs147228155 Chr.8: 26370284-26370284 on Build GRCh38
rs192044729 Chr.8: 26370058-26370058 on Build GRCh38
rs549012087 Chr.8: 26370052-26370052 on Build GRCh38
rs144437201 Chr.8: 26370360-26370360 on Build GRCh38
rs748289874 Chr.8: 26370193-26370193 on Build GRCh38
rs763565585 Chr.8: 26370222-26370222 on Build GRCh38
rs967251772 Chr.8: 26370038-26370038 on Build GRCh38
rs868669914 Chr.8: 26370144-26370144 on Build GRCh38
rs749340495 Chr.8: 26370380-26370380 on Build GRCh38
rs759910208 Chr.8: 26370234-26370234 on Build GRCh38
rs199800792 Chr.8: 26370187-26370187 on Build GRCh38
rs561256378 Chr.8: 26369971-26369971 on Build GRCh38
rs753325304 Chr.8: 26370101-26370101 on Build GRCh38
rs1011216059 Chr.8: 26369990-26369990 on Build GRCh38
rs778877382 Chr.8: 26370119-26370119 on Build GRCh38
rs1028727937 Chr.8: 26369985-26369985 on Build GRCh38
rs376465686 Chr.8: 26370029-26370029 on Build GRCh38

View Full Product Details