Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13962255-13962846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPON1
Entrez Gene ID
10418
Gene Name
spondin 1
Gene Aliases
VSGP/F-spondin, f-spondin
UniGene
Hs.643864
Gene Chromosome Location
Chr.11: 13962637-14268133 on Build GRCh38
Transcripts
NM_006108.3

SNP Detail

SNP ID Chromosome Location
rs964798568 Chr.11: 13962581-13962581 on Build GRCh38
rs1046553524 Chr.11: 13962671-13962671 on Build GRCh38
rs907389657 Chr.11: 13962699-13962699 on Build GRCh38
rs1026576294 Chr.11: 13962726-13962726 on Build GRCh38
rs2697836 Chr.11: 13962393-13962393 on Build GRCh38
rs570921196 Chr.11: 13962658-13962658 on Build GRCh38
rs11281564 Chr.11: 13962708-13962709 on Build GRCh38
rs902194422 Chr.11: 13962356-13962356 on Build GRCh38
rs28413184 Chr.11: 13962730-13962731 on Build GRCh38
rs1006418646 Chr.11: 13962750-13962750 on Build GRCh38
rs971960179 Chr.11: 13962491-13962491 on Build GRCh38
rs781899106 Chr.11: 13962389-13962389 on Build GRCh38
rs933125337 Chr.11: 13962591-13962591 on Build GRCh38
rs536303056 Chr.11: 13962822-13962822 on Build GRCh38
rs950995092 Chr.11: 13962744-13962744 on Build GRCh38
rs374213188 Chr.11: 13962410-13962410 on Build GRCh38
rs538746539 Chr.11: 13962784-13962784 on Build GRCh38
rs1016246384 Chr.11: 13962775-13962775 on Build GRCh38
rs984723736 Chr.11: 13962552-13962552 on Build GRCh38
rs1053489887 Chr.11: 13962649-13962649 on Build GRCh38
rs1038880949 Chr.11: 13962285-13962285 on Build GRCh38
rs556732732 Chr.11: 13962793-13962793 on Build GRCh38
rs145605891 Chr.11: 13962294-13962294 on Build GRCh38
rs112733575 Chr.11: 13962549-13962549 on Build GRCh38
rs552771436 Chr.11: 13962618-13962618 on Build GRCh38
rs138127114 Chr.11: 13962349-13962349 on Build GRCh38
rs1012001753 Chr.11: 13962458-13962458 on Build GRCh38
rs1026961387 Chr.11: 13962511-13962511 on Build GRCh38
rs923151343 Chr.11: 13962589-13962589 on Build GRCh38
rs964661049 Chr.11: 13962781-13962781 on Build GRCh38
rs1003427474 Chr.11: 13962716-13962716 on Build GRCh38
rs974783497 Chr.11: 13962582-13962582 on Build GRCh38
rs1003150331 Chr.11: 13962499-13962499 on Build GRCh38
rs936323828 Chr.11: 13962635-13962635 on Build GRCh38
rs7122836 Chr.11: 13962817-13962817 on Build GRCh38
rs1030802799 Chr.11: 13962825-13962825 on Build GRCh38
rs566869706 Chr.11: 13962493-13962493 on Build GRCh38
rs370807259 Chr.11: 13962370-13962370 on Build GRCh38
rs1042677586 Chr.11: 13962351-13962351 on Build GRCh38
rs923818958 Chr.11: 13962626-13962626 on Build GRCh38
rs149564039 Chr.11: 13962382-13962382 on Build GRCh38
rs548168158 Chr.11: 13962465-13962465 on Build GRCh38
rs909212492 Chr.11: 13962561-13962561 on Build GRCh38
rs1012032725 Chr.11: 13962664-13962664 on Build GRCh38
rs1023174384 Chr.11: 13962462-13962462 on Build GRCh38
rs538162007 Chr.11: 13962470-13962470 on Build GRCh38
rs1000504486 Chr.11: 13962363-13962363 on Build GRCh38

View Full Product Details