Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56468375-56468921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRYD4
Entrez Gene ID
283377
Gene Name
SPRY domain containing 4
Gene Aliases
-
UniGene
Hs.128676
Gene Chromosome Location
Chr.12: 56468517-56470985 on Build GRCh38
Transcripts
NM_207344.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368890 c.36C>T p.C12C Chr.12: 56468627-56468627 on Build GRCh38
6574918 c.81G>C p.Q27H Chr.12: 56468672-56468672 on Build GRCh38
6582840 c.19C>G p.R7G Chr.12: 56468610-56468610 on Build GRCh38
1636785 c.46G>A p.A16T Chr.12: 56468637-56468637 on Build GRCh38
5762306 c.28C>T p.R10C Chr.12: 56468619-56468619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941705747 Chr.12: 56468527-56468527 on Build GRCh38
rs545187770 Chr.12: 56468427-56468427 on Build GRCh38
rs990106767 Chr.12: 56468399-56468399 on Build GRCh38
rs368147997 Chr.12: 56468452-56468452 on Build GRCh38
rs1037977108 Chr.12: 56468429-56468429 on Build GRCh38
rs749206736 Chr.12: 56468571-56468571 on Build GRCh38
rs371212068 Chr.12: 56468548-56468548 on Build GRCh38
rs61937735 Chr.12: 56468453-56468453 on Build GRCh38
rs958408310 Chr.12: 56468502-56468502 on Build GRCh38
rs1034639250 Chr.12: 56468456-56468456 on Build GRCh38
rs772888157 Chr.12: 56468608-56468608 on Build GRCh38
rs770791312 Chr.12: 56468419-56468419 on Build GRCh38
rs575653389 Chr.12: 56468715-56468715 on Build GRCh38
rs771945119 Chr.12: 56468674-56468674 on Build GRCh38
rs754928887 Chr.12: 56468652-56468652 on Build GRCh38
rs570364057 Chr.12: 56468499-56468499 on Build GRCh38
rs991671489 Chr.12: 56468521-56468521 on Build GRCh38
rs890628623 Chr.12: 56468459-56468459 on Build GRCh38
rs994687149 Chr.12: 56468454-56468454 on Build GRCh38
rs530252305 Chr.12: 56468396-56468396 on Build GRCh38
rs528742747 Chr.12: 56468826-56468826 on Build GRCh38
rs369088791 Chr.12: 56468626-56468626 on Build GRCh38
rs373329154 Chr.12: 56468655-56468655 on Build GRCh38
rs890148584 Chr.12: 56468440-56468440 on Build GRCh38
rs768421186 Chr.12: 56468628-56468628 on Build GRCh38
rs750475871 Chr.12: 56468566-56468566 on Build GRCh38
rs949874218 Chr.12: 56468850-56468850 on Build GRCh38
rs774749872 Chr.12: 56468713-56468713 on Build GRCh38
rs1014211693 Chr.12: 56468479-56468479 on Build GRCh38
rs770909661 Chr.12: 56468663-56468664 on Build GRCh38
rs575448834 Chr.12: 56468762-56468762 on Build GRCh38
rs906500413 Chr.12: 56468634-56468634 on Build GRCh38
rs148775301 Chr.12: 56468614-56468614 on Build GRCh38
rs1055622477 Chr.12: 56468578-56468578 on Build GRCh38
rs781320399 Chr.12: 56468560-56468560 on Build GRCh38
rs770436149 Chr.12: 56468618-56468618 on Build GRCh38
rs188310075 Chr.12: 56468444-56468444 on Build GRCh38
rs541484926 Chr.12: 56468752-56468752 on Build GRCh38
rs1010836375 Chr.12: 56468481-56468481 on Build GRCh38
rs912153317 Chr.12: 56468760-56468760 on Build GRCh38
rs745404567 Chr.12: 56468508-56468508 on Build GRCh38
rs773724857 Chr.12: 56468646-56468646 on Build GRCh38
rs778917895 Chr.12: 56468665-56468665 on Build GRCh38
rs764503222 Chr.12: 56468624-56468624 on Build GRCh38
rs930426048 Chr.12: 56468572-56468572 on Build GRCh38
rs148180043 Chr.12: 56468597-56468597 on Build GRCh38
rs768338495 Chr.12: 56468585-56468585 on Build GRCh38
rs779874850 Chr.12: 56468569-56468569 on Build GRCh38
rs757674013 Chr.12: 56468513-56468513 on Build GRCh38
rs1024964029 Chr.12: 56468485-56468485 on Build GRCh38
rs929585908 Chr.12: 56468550-56468550 on Build GRCh38
rs565238115 Chr.12: 56468737-56468737 on Build GRCh38
rs141941790 Chr.12: 56468759-56468759 on Build GRCh38
rs182275613 Chr.12: 56468754-56468754 on Build GRCh38
rs757460769 Chr.12: 56468559-56468559 on Build GRCh38
rs534682398 Chr.12: 56468500-56468500 on Build GRCh38
rs116824150 Chr.12: 56468865-56468865 on Build GRCh38
rs970737901 Chr.12: 56468491-56468491 on Build GRCh38
rs940373993 Chr.12: 56468553-56468553 on Build GRCh38
rs879427362 Chr.12: 56468883-56468883 on Build GRCh38
rs367615918 Chr.12: 56468536-56468536 on Build GRCh38
rs930395142 Chr.12: 56468811-56468811 on Build GRCh38
rs142431089 Chr.12: 56468629-56468629 on Build GRCh38
rs763439721 Chr.12: 56468698-56468698 on Build GRCh38
rs912121982 Chr.12: 56468869-56468869 on Build GRCh38
rs60020215 Chr.12: 56468822-56468822 on Build GRCh38
rs371299225 Chr.12: 56468725-56468725 on Build GRCh38
rs894387734 Chr.12: 56468469-56468469 on Build GRCh38
rs1039167358 Chr.12: 56468899-56468899 on Build GRCh38
rs767825644 Chr.12: 56468720-56468720 on Build GRCh38
rs943545024 Chr.12: 56468885-56468885 on Build GRCh38
rs879144728 Chr.12: 56468471-56468471 on Build GRCh38
rs911534227 Chr.12: 56468549-56468549 on Build GRCh38
rs920287172 Chr.12: 56468786-56468786 on Build GRCh38
rs531072108 Chr.12: 56468824-56468824 on Build GRCh38
rs922212046 Chr.12: 56468407-56468407 on Build GRCh38
rs770362327 Chr.12: 56468689-56468689 on Build GRCh38
rs936869638 Chr.12: 56468394-56468394 on Build GRCh38
rs548459625 Chr.12: 56468424-56468424 on Build GRCh38
rs747866281 Chr.12: 56468592-56468592 on Build GRCh38
rs190452047 Chr.12: 56468744-56468744 on Build GRCh38
rs767700811 Chr.12: 56468627-56468627 on Build GRCh38
rs776193925 Chr.12: 56468692-56468692 on Build GRCh38
rs777600604 Chr.12: 56468675-56468675 on Build GRCh38
rs374992865 Chr.12: 56468619-56468619 on Build GRCh38
rs989662237 Chr.12: 56468505-56468505 on Build GRCh38
rs183489751 Chr.12: 56468523-56468523 on Build GRCh38
rs766425492 Chr.12: 56468727-56468727 on Build GRCh38
rs971700854 Chr.12: 56468767-56468767 on Build GRCh38
rs1008088929 Chr.12: 56468478-56468478 on Build GRCh38
rs928510195 Chr.12: 56468495-56468495 on Build GRCh38
rs535322155 Chr.12: 56468539-56468539 on Build GRCh38
rs763461563 Chr.12: 56468620-56468620 on Build GRCh38
rs1020872100 Chr.12: 56468669-56468669 on Build GRCh38
rs746608124 Chr.12: 56468687-56468687 on Build GRCh38
rs371448979 Chr.12: 56468541-56468541 on Build GRCh38
rs928940822 Chr.12: 56468844-56468844 on Build GRCh38
rs756122115 Chr.12: 56468567-56468567 on Build GRCh38

View Full Product Details