Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 30734938-30735523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAPTM5
Entrez Gene ID
7805
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
CLAST6
UniGene
Hs.371021
Gene Chromosome Location
Chr.1: 30732468-30757840 on Build GRCh38
Transcripts
NM_006762.2 XM_011542098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5001925 c.677G>A p.R226K Chr.1: 30735195-30735195 on Build GRCh38
3487930 c.657G>A p.M219I Chr.1: 30735215-30735215 on Build GRCh38
6699550 c.670_672delGAG p.E224delE Chr.1: 30735200-30735202 on Build GRCh38
6699549 c.632G>A p.R211Q Chr.1: 30735240-30735240 on Build GRCh38
370979 c.649A>G p.K217E Chr.1: 30735223-30735223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188111001 Chr.1: 30735288-30735288 on Build GRCh38
rs765898053 Chr.1: 30735330-30735330 on Build GRCh38
rs768836998 Chr.1: 30735248-30735248 on Build GRCh38
rs746263342 Chr.1: 30735229-30735229 on Build GRCh38
rs772252143 Chr.1: 30735144-30735144 on Build GRCh38
rs766081076 Chr.1: 30735146-30735146 on Build GRCh38
rs751366621 Chr.1: 30735184-30735184 on Build GRCh38
rs1027667920 Chr.1: 30734998-30734998 on Build GRCh38
rs375164990 Chr.1: 30735303-30735303 on Build GRCh38
rs780267330 Chr.1: 30735317-30735317 on Build GRCh38
rs767217599 Chr.1: 30735188-30735188 on Build GRCh38
rs1034557146 Chr.1: 30735469-30735469 on Build GRCh38
rs538720201 Chr.1: 30735319-30735319 on Build GRCh38
rs1007202738 Chr.1: 30735057-30735057 on Build GRCh38
rs766323570 Chr.1: 30735182-30735182 on Build GRCh38
rs987344783 Chr.1: 30735205-30735205 on Build GRCh38
rs764594879 Chr.1: 30735258-30735258 on Build GRCh38
rs755659020 Chr.1: 30735207-30735207 on Build GRCh38
rs1046816286 Chr.1: 30735445-30735445 on Build GRCh38
rs368099878 Chr.1: 30735236-30735236 on Build GRCh38
rs1041820263 Chr.1: 30734982-30734982 on Build GRCh38
rs923769236 Chr.1: 30734984-30734984 on Build GRCh38
rs575482265 Chr.1: 30735406-30735406 on Build GRCh38
rs911779750 Chr.1: 30735462-30735462 on Build GRCh38
rs774686280 Chr.1: 30735127-30735127 on Build GRCh38
rs532243663 Chr.1: 30735497-30735497 on Build GRCh38
rs138163595 Chr.1: 30735240-30735240 on Build GRCh38
rs771019420 Chr.1: 30735280-30735280 on Build GRCh38
rs367632339 Chr.1: 30735166-30735166 on Build GRCh38
rs1535637 Chr.1: 30735055-30735055 on Build GRCh38
rs537360651 Chr.1: 30735107-30735107 on Build GRCh38
rs760820990 Chr.1: 30735161-30735161 on Build GRCh38
rs1047420218 Chr.1: 30735289-30735289 on Build GRCh38
rs993761590 Chr.1: 30735433-30735433 on Build GRCh38
rs1023534740 Chr.1: 30735337-30735337 on Build GRCh38
rs776789935 Chr.1: 30735304-30735304 on Build GRCh38
rs773264858 Chr.1: 30735176-30735176 on Build GRCh38
rs993918662 Chr.1: 30735128-30735128 on Build GRCh38
rs754429654 Chr.1: 30735209-30735209 on Build GRCh38
rs1025309414 Chr.1: 30735138-30735138 on Build GRCh38
rs540268973 Chr.1: 30735421-30735421 on Build GRCh38
rs761225896 Chr.1: 30735210-30735210 on Build GRCh38
rs191970330 Chr.1: 30735034-30735034 on Build GRCh38
rs894630438 Chr.1: 30735481-30735481 on Build GRCh38
rs374608139 Chr.1: 30735216-30735216 on Build GRCh38
rs72876490 Chr.1: 30735148-30735148 on Build GRCh38
rs759360466 Chr.1: 30735187-30735187 on Build GRCh38
rs574774147 Chr.1: 30735415-30735415 on Build GRCh38
rs147484958 Chr.1: 30735020-30735020 on Build GRCh38
rs371790554 Chr.1: 30735251-30735251 on Build GRCh38
rs759421669 Chr.1: 30735287-30735287 on Build GRCh38
rs752463551 Chr.1: 30735290-30735290 on Build GRCh38
rs1038070279 Chr.1: 30735069-30735069 on Build GRCh38
rs544398625 Chr.1: 30735408-30735408 on Build GRCh38
rs571708777 Chr.1: 30735064-30735064 on Build GRCh38
rs774378981 Chr.1: 30735284-30735284 on Build GRCh38
rs368441175 Chr.1: 30735314-30735314 on Build GRCh38
rs116352948 Chr.1: 30735315-30735315 on Build GRCh38
rs112626544 Chr.1: 30735153-30735153 on Build GRCh38
rs751354799 Chr.1: 30735200-30735202 on Build GRCh38
rs770995931 Chr.1: 30734972-30734972 on Build GRCh38
rs1051023077 Chr.1: 30735045-30735045 on Build GRCh38
rs1038691373 Chr.1: 30735414-30735414 on Build GRCh38
rs1023016777 Chr.1: 30735145-30735145 on Build GRCh38
rs35351292 Chr.1: 30735195-30735195 on Build GRCh38
rs762969146 Chr.1: 30735181-30735181 on Build GRCh38
rs559846654 Chr.1: 30735463-30735463 on Build GRCh38
rs34923956 Chr.1: 30735131-30735132 on Build GRCh38
rs759488183 Chr.1: 30735026-30735026 on Build GRCh38
rs376284297 Chr.1: 30735162-30735162 on Build GRCh38
rs754504521 Chr.1: 30735450-30735450 on Build GRCh38
rs751273668 Chr.1: 30735380-30735380 on Build GRCh38
rs561203417 Chr.1: 30735412-30735412 on Build GRCh38
rs139827360 Chr.1: 30735392-30735392 on Build GRCh38
rs772379976 Chr.1: 30735234-30735234 on Build GRCh38
rs774668460 Chr.1: 30735018-30735018 on Build GRCh38
rs553091100 Chr.1: 30735292-30735292 on Build GRCh38
rs370275111 Chr.1: 30735208-30735208 on Build GRCh38
rs187822247 Chr.1: 30734979-30734979 on Build GRCh38
rs746140587 Chr.1: 30735143-30735143 on Build GRCh38

View Full Product Details