Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32330455-32331052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC1
Entrez Gene ID
3065
Gene Name
histone deacetylase 1
Gene Aliases
GON-10, HD1, RPD3, RPD3L1
UniGene
Hs.88556
Gene Chromosome Location
Chr.1: 32292103-32333628 on Build GRCh38
Transcripts
NM_004964.2 XM_011541309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4589639 c.839-2_839-1insG p.? Chr.1: 32330766-32330767 on Build GRCh38
7166510 c.826C>A p.L276I Chr.1: 32330674-32330674 on Build GRCh38
6062982 c.895G>T p.G299* Chr.1: 32330824-32330824 on Build GRCh38
2077934 c.844G>A p.A282T Chr.1: 32330773-32330773 on Build GRCh38
7222977 c.901G>A p.G301S Chr.1: 32330830-32330830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037525551 Chr.1: 32330998-32330998 on Build GRCh38
rs766486993 Chr.1: 32330936-32330936 on Build GRCh38
rs971613278 Chr.1: 32330789-32330789 on Build GRCh38
rs1026004931 Chr.1: 32330720-32330720 on Build GRCh38
rs770299349 Chr.1: 32330871-32330871 on Build GRCh38
rs781657568 Chr.1: 32330858-32330858 on Build GRCh38
rs150636639 Chr.1: 32330772-32330772 on Build GRCh38
rs915326776 Chr.1: 32330494-32330494 on Build GRCh38
rs761216909 Chr.1: 32330590-32330590 on Build GRCh38
rs112156956 Chr.1: 32330920-32330920 on Build GRCh38
rs866943523 Chr.1: 32330488-32330488 on Build GRCh38
rs901651382 Chr.1: 32330703-32330703 on Build GRCh38
rs748487694 Chr.1: 32330868-32330868 on Build GRCh38
rs569851180 Chr.1: 32331007-32331007 on Build GRCh38
rs778088376 Chr.1: 32330981-32330981 on Build GRCh38
rs144913235 Chr.1: 32330739-32330739 on Build GRCh38
rs771053554 Chr.1: 32330531-32330531 on Build GRCh38
rs927587234 Chr.1: 32330846-32330846 on Build GRCh38
rs149903418 Chr.1: 32330554-32330554 on Build GRCh38
rs889041428 Chr.1: 32330995-32330995 on Build GRCh38
rs190374705 Chr.1: 32330643-32330643 on Build GRCh38
rs1007360168 Chr.1: 32330568-32330568 on Build GRCh38
rs889028942 Chr.1: 32330989-32330989 on Build GRCh38
rs937376262 Chr.1: 32330508-32330508 on Build GRCh38
rs373980046 Chr.1: 32330914-32330914 on Build GRCh38
rs927536348 Chr.1: 32330502-32330502 on Build GRCh38
rs559431042 Chr.1: 32330519-32330519 on Build GRCh38
rs756126049 Chr.1: 32330806-32330806 on Build GRCh38
rs763960381 Chr.1: 32330829-32330829 on Build GRCh38
rs906201378 Chr.1: 32331031-32331031 on Build GRCh38
rs139364145 Chr.1: 32330883-32330883 on Build GRCh38
rs897467188 Chr.1: 32331013-32331013 on Build GRCh38
rs780754516 Chr.1: 32330845-32330845 on Build GRCh38
rs1006109887 Chr.1: 32331004-32331004 on Build GRCh38
rs565534358 Chr.1: 32330894-32330894 on Build GRCh38
rs547964584 Chr.1: 32330987-32330987 on Build GRCh38
rs781304829 Chr.1: 32330956-32330956 on Build GRCh38
rs898976591 Chr.1: 32330999-32330999 on Build GRCh38
rs370783702 Chr.1: 32330551-32330551 on Build GRCh38
rs780284831 Chr.1: 32330725-32330725 on Build GRCh38
rs1041952475 Chr.1: 32330681-32330681 on Build GRCh38
rs755148127 Chr.1: 32330953-32330953 on Build GRCh38
rs769069282 Chr.1: 32330737-32330737 on Build GRCh38
rs922639096 Chr.1: 32330928-32330928 on Build GRCh38
rs751266844 Chr.1: 32330790-32330790 on Build GRCh38
rs547455476 Chr.1: 32330767-32330767 on Build GRCh38
rs769168564 Chr.1: 32330764-32330764 on Build GRCh38
rs201632929 Chr.1: 32330932-32330932 on Build GRCh38
rs1015951069 Chr.1: 32331010-32331010 on Build GRCh38
rs781215783 Chr.1: 32330706-32330706 on Build GRCh38
rs759389151 Chr.1: 32330793-32330793 on Build GRCh38
rs377238510 Chr.1: 32330548-32330548 on Build GRCh38
rs750474499 Chr.1: 32330838-32330838 on Build GRCh38
rs767482750 Chr.1: 32330799-32330799 on Build GRCh38
rs755305485 Chr.1: 32330853-32330853 on Build GRCh38
rs368390726 Chr.1: 32331003-32331003 on Build GRCh38
rs777137616 Chr.1: 32330566-32330566 on Build GRCh38
rs370274596 Chr.1: 32330913-32330913 on Build GRCh38
rs367710446 Chr.1: 32330895-32330895 on Build GRCh38
rs747305440 Chr.1: 32330730-32330730 on Build GRCh38
rs536372070 Chr.1: 32330931-32330931 on Build GRCh38
rs766065383 Chr.1: 32330777-32330777 on Build GRCh38
rs750012758 Chr.1: 32330597-32330597 on Build GRCh38
rs993552744 Chr.1: 32331019-32331019 on Build GRCh38
rs35678923 Chr.1: 32330496-32330497 on Build GRCh38
rs751065178 Chr.1: 32330616-32330616 on Build GRCh38
rs990117314 Chr.1: 32330890-32330890 on Build GRCh38
rs749691706 Chr.1: 32330988-32330988 on Build GRCh38
rs778986200 Chr.1: 32330532-32330532 on Build GRCh38
rs761048323 Chr.1: 32330561-32330561 on Build GRCh38
rs769110096 Chr.1: 32330563-32330563 on Build GRCh38
rs751758361 Chr.1: 32330951-32330951 on Build GRCh38
rs763738370 Chr.1: 32330716-32330717 on Build GRCh38
rs933204239 Chr.1: 32330960-32330960 on Build GRCh38
rs1042358622 Chr.1: 32330509-32330509 on Build GRCh38
rs751362760 Chr.1: 32330647-32330647 on Build GRCh38
rs758469745 Chr.1: 32330935-32330935 on Build GRCh38
rs199676425 Chr.1: 32330614-32330614 on Build GRCh38
rs747301084 Chr.1: 32330851-32330851 on Build GRCh38
rs202175581 Chr.1: 32330752-32330752 on Build GRCh38
rs185231354 Chr.1: 32330483-32330483 on Build GRCh38
rs34386633 Chr.1: 32330988-32330989 on Build GRCh38
rs201672032 Chr.1: 32330882-32330882 on Build GRCh38
rs993260556 Chr.1: 32331002-32331002 on Build GRCh38
rs530977896 Chr.1: 32330707-32330707 on Build GRCh38
rs376404835 Chr.1: 32330719-32330719 on Build GRCh38
rs764870283 Chr.1: 32330596-32330596 on Build GRCh38
rs1015900348 Chr.1: 32330722-32330722 on Build GRCh38
rs933949409 Chr.1: 32330984-32330984 on Build GRCh38
rs1053804424 Chr.1: 32330494-32330495 on Build GRCh38
rs757276885 Chr.1: 32330709-32330709 on Build GRCh38
rs535464617 Chr.1: 32330926-32330926 on Build GRCh38
rs200411094 Chr.1: 32330718-32330718 on Build GRCh38
rs762720122 Chr.1: 32330620-32330620 on Build GRCh38
rs754827385 Chr.1: 32330704-32330704 on Build GRCh38
rs138410521 Chr.1: 32330615-32330615 on Build GRCh38
rs780243739 Chr.1: 32330850-32330850 on Build GRCh38

View Full Product Details