Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96088435-96089016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf132
Entrez Gene ID
56967
Gene Name
chromosome 14 open reading frame 132
Gene Aliases
C14orf88
UniGene
Hs.6434
Gene Chromosome Location
Chr.14: 96039324-96093971 on Build GRCh38
Transcripts
NM_001252507.2 NM_001282463.1 NM_001282464.1 NM_001289139.1 XM_017021469.1

SNP Detail

SNP ID Chromosome Location
rs560961467 Chr.14: 96088478-96088478 on Build GRCh38
rs566872111 Chr.14: 96088742-96088742 on Build GRCh38
rs1034196002 Chr.14: 96088512-96088512 on Build GRCh38
rs888507977 Chr.14: 96088711-96088711 on Build GRCh38
rs930223761 Chr.14: 96088931-96088931 on Build GRCh38
rs1034747595 Chr.14: 96088641-96088641 on Build GRCh38
rs1010015122 Chr.14: 96088505-96088505 on Build GRCh38
rs965309799 Chr.14: 96088523-96088523 on Build GRCh38
rs1006496599 Chr.14: 96088750-96088750 on Build GRCh38
rs538873945 Chr.14: 96088807-96088807 on Build GRCh38
rs189990458 Chr.14: 96088516-96088516 on Build GRCh38
rs958714788 Chr.14: 96088617-96088617 on Build GRCh38
rs575318529 Chr.14: 96088824-96088824 on Build GRCh38
rs547925587 Chr.14: 96088781-96088781 on Build GRCh38
rs971928574 Chr.14: 96088792-96088792 on Build GRCh38
rs918850356 Chr.14: 96088673-96088673 on Build GRCh38
rs1039687477 Chr.14: 96088749-96088749 on Build GRCh38
rs79262207 Chr.14: 96088682-96088682 on Build GRCh38
rs534137550 Chr.14: 96088752-96088752 on Build GRCh38
rs75041031 Chr.14: 96088945-96088945 on Build GRCh38
rs577106760 Chr.14: 96088969-96088969 on Build GRCh38
rs971725205 Chr.14: 96088908-96088908 on Build GRCh38
rs147787196 Chr.14: 96088880-96088880 on Build GRCh38
rs540533975 Chr.14: 96088514-96088514 on Build GRCh38
rs749942173 Chr.14: 96088839-96088839 on Build GRCh38
rs60338535 Chr.14: 96088595-96088595 on Build GRCh38
rs200363717 Chr.14: 96088485-96088485 on Build GRCh38
rs142510994 Chr.14: 96088504-96088504 on Build GRCh38
rs916925951 Chr.14: 96088646-96088646 on Build GRCh38
rs544504645 Chr.14: 96088982-96088982 on Build GRCh38
rs557096900 Chr.14: 96088819-96088819 on Build GRCh38
rs45528638 Chr.14: 96088882-96088882 on Build GRCh38
rs763919040 Chr.14: 96088650-96088650 on Build GRCh38
rs1007413510 Chr.14: 96088722-96088722 on Build GRCh38
rs1049055777 Chr.14: 96088705-96088705 on Build GRCh38
rs916913906 Chr.14: 96088881-96088881 on Build GRCh38
rs1055117419 Chr.14: 96088891-96088891 on Build GRCh38
rs967152509 Chr.14: 96088584-96088584 on Build GRCh38
rs1029963694 Chr.14: 96088808-96088808 on Build GRCh38
rs566626914 Chr.14: 96088782-96088782 on Build GRCh38
rs759879935 Chr.14: 96088513-96088513 on Build GRCh38
rs1048480612 Chr.14: 96088507-96088507 on Build GRCh38
rs754067952 Chr.14: 96088986-96088986 on Build GRCh38
rs144309743 Chr.14: 96088865-96088865 on Build GRCh38
rs897625386 Chr.14: 96088947-96088947 on Build GRCh38
rs530185738 Chr.14: 96088649-96088649 on Build GRCh38
rs914049221 Chr.14: 96088874-96088874 on Build GRCh38
rs888533146 Chr.14: 96088735-96088735 on Build GRCh38
rs1046715581 Chr.14: 96088657-96088657 on Build GRCh38
rs985756115 Chr.14: 96088854-96088854 on Build GRCh38
rs75295665 Chr.14: 96088566-96088566 on Build GRCh38
rs967287142 Chr.14: 96088830-96088830 on Build GRCh38
rs534011729 Chr.14: 96088533-96088533 on Build GRCh38
rs922918946 Chr.14: 96088578-96088578 on Build GRCh38
rs867599485 Chr.14: 96088602-96088602 on Build GRCh38
rs776750231 Chr.14: 96088506-96088506 on Build GRCh38
rs71415023 Chr.14: 96088915-96088915 on Build GRCh38
rs368400665 Chr.14: 96088877-96088877 on Build GRCh38
rs1030243963 Chr.14: 96088989-96088989 on Build GRCh38
rs1005613043 Chr.14: 96088929-96088929 on Build GRCh38
rs909156057 Chr.14: 96088515-96088515 on Build GRCh38
rs1033804539 Chr.14: 96088712-96088712 on Build GRCh38
rs991159024 Chr.14: 96088636-96088636 on Build GRCh38
rs999958086 Chr.14: 96088860-96088860 on Build GRCh38
rs1002296213 Chr.14: 96088703-96088703 on Build GRCh38
rs978570443 Chr.14: 96088585-96088585 on Build GRCh38
rs76557394 Chr.14: 96088683-96088683 on Build GRCh38
rs770138156 Chr.14: 96088553-96088553 on Build GRCh38
rs79737394 Chr.14: 96088864-96088864 on Build GRCh38
rs918817804 Chr.14: 96088924-96088924 on Build GRCh38
rs910062960 Chr.14: 96088946-96088946 on Build GRCh38
rs938029130 Chr.14: 96088884-96088884 on Build GRCh38
rs193028827 Chr.14: 96088938-96088938 on Build GRCh38
rs188965286 Chr.14: 96088616-96088616 on Build GRCh38

View Full Product Details