Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40090756-40091320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMF
Entrez Gene ID
90427
Gene Name
Bcl2 modifying factor
Gene Aliases
-
UniGene
Hs.591104
Gene Chromosome Location
Chr.15: 40087890-40108884 on Build GRCh38
Transcripts
NM_001003940.1 NM_001003942.1 NM_001003943.2 NM_033503.3

SNP Detail

SNP ID Chromosome Location
rs146020024 Chr.15: 40090887-40090887 on Build GRCh38
rs1007437427 Chr.15: 40090854-40090854 on Build GRCh38
rs780255669 Chr.15: 40090866-40090866 on Build GRCh38
rs545017505 Chr.15: 40090949-40090949 on Build GRCh38
rs924297748 Chr.15: 40090959-40090959 on Build GRCh38
rs1047606668 Chr.15: 40091266-40091266 on Build GRCh38
rs1026304090 Chr.15: 40090872-40090872 on Build GRCh38
rs1026325935 Chr.15: 40090979-40090979 on Build GRCh38
rs890277647 Chr.15: 40090832-40090832 on Build GRCh38
rs886434405 Chr.15: 40091096-40091096 on Build GRCh38
rs547997483 Chr.15: 40091134-40091134 on Build GRCh38
rs915455486 Chr.15: 40090913-40090913 on Build GRCh38
rs911611196 Chr.15: 40091216-40091216 on Build GRCh38
rs1038213256 Chr.15: 40091203-40091203 on Build GRCh38
rs776543661 Chr.15: 40091189-40091189 on Build GRCh38
rs1003522580 Chr.15: 40091295-40091295 on Build GRCh38
rs902960040 Chr.15: 40090791-40090791 on Build GRCh38
rs955664496 Chr.15: 40091177-40091177 on Build GRCh38
rs1003638495 Chr.15: 40091098-40091098 on Build GRCh38
rs911727419 Chr.15: 40091005-40091005 on Build GRCh38
rs150650651 Chr.15: 40091263-40091263 on Build GRCh38
rs943110872 Chr.15: 40091222-40091222 on Build GRCh38
rs934391378 Chr.15: 40090806-40090806 on Build GRCh38
rs1039303981 Chr.15: 40091234-40091234 on Build GRCh38
rs936494 Chr.15: 40091146-40091146 on Build GRCh38
rs1035063977 Chr.15: 40091099-40091099 on Build GRCh38
rs899158121 Chr.15: 40090909-40090909 on Build GRCh38
rs890473690 Chr.15: 40091249-40091249 on Build GRCh38
rs987198745 Chr.15: 40091192-40091192 on Build GRCh38
rs982192503 Chr.15: 40090882-40090882 on Build GRCh38
rs564995796 Chr.15: 40090905-40090905 on Build GRCh38
rs1042824914 Chr.15: 40090785-40090785 on Build GRCh38
rs371844346 Chr.15: 40090989-40090989 on Build GRCh38
rs1048232785 Chr.15: 40091050-40091050 on Build GRCh38
rs78537933 Chr.15: 40090807-40090807 on Build GRCh38
rs376823616 Chr.15: 40091016-40091016 on Build GRCh38
rs1012449549 Chr.15: 40091117-40091117 on Build GRCh38
rs781506615 Chr.15: 40090901-40090901 on Build GRCh38
rs188357199 Chr.15: 40091149-40091149 on Build GRCh38
rs772266864 Chr.15: 40091197-40091197 on Build GRCh38
rs968315008 Chr.15: 40090922-40090922 on Build GRCh38
rs543601333 Chr.15: 40090867-40090867 on Build GRCh38
rs1022443457 Chr.15: 40091129-40091129 on Build GRCh38
rs959546135 Chr.15: 40091107-40091107 on Build GRCh38
rs899108502 Chr.15: 40091036-40091036 on Build GRCh38
rs141829942 Chr.15: 40091141-40091141 on Build GRCh38
rs934445048 Chr.15: 40090984-40090984 on Build GRCh38
rs934360040 Chr.15: 40090828-40090828 on Build GRCh38
rs899214994 Chr.15: 40091213-40091213 on Build GRCh38
rs2131975 Chr.15: 40090927-40090927 on Build GRCh38
rs148093475 Chr.15: 40091017-40091017 on Build GRCh38
rs562805633 Chr.15: 40091074-40091074 on Build GRCh38
rs771639919 Chr.15: 40091118-40091118 on Build GRCh38
rs112193244 Chr.15: 40091075-40091075 on Build GRCh38
rs768609129 Chr.15: 40091158-40091158 on Build GRCh38
rs554674107 Chr.15: 40090831-40090831 on Build GRCh38
rs1035221350 Chr.15: 40090891-40090891 on Build GRCh38
rs943107919 Chr.15: 40090784-40090784 on Build GRCh38
rs745320364 Chr.15: 40091041-40091041 on Build GRCh38
rs941226637 Chr.15: 40091172-40091172 on Build GRCh38
rs978858789 Chr.15: 40090937-40090937 on Build GRCh38
rs576817811 Chr.15: 40090988-40090988 on Build GRCh38
rs995219742 Chr.15: 40090869-40090869 on Build GRCh38
rs78789343 Chr.15: 40090883-40090883 on Build GRCh38
rs531375512 Chr.15: 40091112-40091112 on Build GRCh38
rs761863193 Chr.15: 40091256-40091256 on Build GRCh38
rs978525523 Chr.15: 40091022-40091022 on Build GRCh38
rs377186408 Chr.15: 40091257-40091257 on Build GRCh38
rs559871001 Chr.15: 40090980-40090980 on Build GRCh38
rs950890084 Chr.15: 40090878-40090878 on Build GRCh38
rs1017433664 Chr.15: 40090865-40090865 on Build GRCh38
rs886442815 Chr.15: 40091286-40091286 on Build GRCh38

View Full Product Details