Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74671779-74672364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLKL
Entrez Gene ID
197259
Gene Name
mixed lineage kinase domain-like
Gene Aliases
hMLKL
UniGene
Hs.119878
Gene Chromosome Location
Chr.16: 74671855-74701187 on Build GRCh38
Transcripts
NM_001142497.2 NM_152649.3 XM_005255834.3

SNP Detail

SNP ID Chromosome Location
rs1006145784 Chr.16: 74672015-74672015 on Build GRCh38
rs550160717 Chr.16: 74671834-74671834 on Build GRCh38
rs1008103756 Chr.16: 74671832-74671832 on Build GRCh38
rs975803785 Chr.16: 74672066-74672066 on Build GRCh38
rs541018794 Chr.16: 74672245-74672245 on Build GRCh38
rs933346633 Chr.16: 74672303-74672303 on Build GRCh38
rs1007741040 Chr.16: 74672127-74672127 on Build GRCh38
rs961179263 Chr.16: 74672261-74672261 on Build GRCh38
rs925455216 Chr.16: 74671831-74671831 on Build GRCh38
rs959236867 Chr.16: 74671984-74671984 on Build GRCh38
rs1003580943 Chr.16: 74672183-74672183 on Build GRCh38
rs548849655 Chr.16: 74671873-74671873 on Build GRCh38
rs756342574 Chr.16: 74672247-74672247 on Build GRCh38
rs930078519 Chr.16: 74672226-74672226 on Build GRCh38
rs537101191 Chr.16: 74671997-74671997 on Build GRCh38
rs187552316 Chr.16: 74671911-74671911 on Build GRCh38
rs764413680 Chr.16: 74672331-74672331 on Build GRCh38
rs949654381 Chr.16: 74672208-74672208 on Build GRCh38
rs772644020 Chr.16: 74671952-74671952 on Build GRCh38
rs551498452 Chr.16: 74671923-74671923 on Build GRCh38
rs1020656643 Chr.16: 74671885-74671885 on Build GRCh38
rs572118788 Chr.16: 74672202-74672202 on Build GRCh38
rs893961872 Chr.16: 74672132-74672132 on Build GRCh38
rs565764201 Chr.16: 74671905-74671905 on Build GRCh38
rs753130880 Chr.16: 74672107-74672107 on Build GRCh38
rs909811267 Chr.16: 74672000-74672000 on Build GRCh38
rs146875457 Chr.16: 74672099-74672099 on Build GRCh38
rs895139350 Chr.16: 74671849-74671849 on Build GRCh38
rs75884420 Chr.16: 74671870-74671870 on Build GRCh38
rs970420451 Chr.16: 74672181-74672181 on Build GRCh38
rs991495106 Chr.16: 74672158-74672158 on Build GRCh38
rs966474503 Chr.16: 74672089-74672089 on Build GRCh38
rs183429081 Chr.16: 74671837-74671837 on Build GRCh38
rs776446474 Chr.16: 74671805-74671805 on Build GRCh38
rs1052265915 Chr.16: 74671808-74671808 on Build GRCh38
rs769449764 Chr.16: 74671912-74671912 on Build GRCh38
rs1017844036 Chr.16: 74672282-74672282 on Build GRCh38
rs139694013 Chr.16: 74671981-74671981 on Build GRCh38
rs889166933 Chr.16: 74672116-74672116 on Build GRCh38
rs970369642 Chr.16: 74671925-74671925 on Build GRCh38
rs151025923 Chr.16: 74671893-74671893 on Build GRCh38
rs972657101 Chr.16: 74672266-74672266 on Build GRCh38
rs191146204 Chr.16: 74672163-74672163 on Build GRCh38
rs1046416283 Chr.16: 74672215-74672215 on Build GRCh38
rs939916021 Chr.16: 74672027-74672027 on Build GRCh38
rs981918144 Chr.16: 74671957-74671957 on Build GRCh38
rs8998 Chr.16: 74672063-74672063 on Build GRCh38
rs1009852347 Chr.16: 74672157-74672157 on Build GRCh38
rs529204081 Chr.16: 74671872-74671872 on Build GRCh38
rs765971434 Chr.16: 74672037-74672037 on Build GRCh38
rs887628761 Chr.16: 74672262-74672262 on Build GRCh38
rs994234789 Chr.16: 74671903-74671903 on Build GRCh38
rs984326434 Chr.16: 74671991-74671991 on Build GRCh38
rs571178091 Chr.16: 74672128-74672128 on Build GRCh38
rs959352520 Chr.16: 74672220-74672220 on Build GRCh38
rs910657730 Chr.16: 74672334-74672334 on Build GRCh38
rs997842037 Chr.16: 74672316-74672316 on Build GRCh38
rs866567331 Chr.16: 74672131-74672131 on Build GRCh38
rs552408440 Chr.16: 74672338-74672338 on Build GRCh38
rs900442424 Chr.16: 74672306-74672306 on Build GRCh38
rs774697305 Chr.16: 74672051-74672051 on Build GRCh38
rs1006031213 Chr.16: 74672271-74672271 on Build GRCh38
rs922913135 Chr.16: 74672078-74672078 on Build GRCh38
rs897144467 Chr.16: 74672221-74672221 on Build GRCh38
rs11861223 Chr.16: 74672065-74672065 on Build GRCh38
rs543612685 Chr.16: 74671839-74671839 on Build GRCh38
rs142879114 Chr.16: 74671828-74671828 on Build GRCh38
rs557746653 Chr.16: 74672255-74672255 on Build GRCh38

View Full Product Details