Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165041403-165041957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM60
Entrez Gene ID
166655
Gene Name
tripartite motif containing 60
Gene Aliases
RNF129, RNF33
UniGene
Hs.368004
Gene Chromosome Location
Chr.4: 165031953-165041821 on Build GRCh38
Transcripts
NM_001258025.1 NM_152620.2 XM_011531683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584895 c.1362T>G p.T454T Chr.4: 165041434-165041434 on Build GRCh38
4599427 c.1346G>A p.W449* Chr.4: 165041418-165041418 on Build GRCh38
6808500 c.1362T>A p.T454T Chr.4: 165041434-165041434 on Build GRCh38
4600940 c.1357T>C p.Y453H Chr.4: 165041429-165041429 on Build GRCh38
1230378 c.1375G>A p.E459K Chr.4: 165041447-165041447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752664959 Chr.4: 165041445-165041445 on Build GRCh38
rs751866324 Chr.4: 165041526-165041526 on Build GRCh38
rs552303477 Chr.4: 165041476-165041476 on Build GRCh38
rs771599222 Chr.4: 165041466-165041466 on Build GRCh38
rs572987977 Chr.4: 165041562-165041563 on Build GRCh38
rs150779555 Chr.4: 165041909-165041909 on Build GRCh38
rs59349426 Chr.4: 165041760-165041761 on Build GRCh38
rs967883943 Chr.4: 165041732-165041732 on Build GRCh38
rs139140339 Chr.4: 165041924-165041924 on Build GRCh38
rs866436244 Chr.4: 165041591-165041591 on Build GRCh38
rs753644939 Chr.4: 165041450-165041450 on Build GRCh38
rs1043626499 Chr.4: 165041717-165041717 on Build GRCh38
rs974662178 Chr.4: 165041882-165041882 on Build GRCh38
rs1056006720 Chr.4: 165041787-165041787 on Build GRCh38
rs780526481 Chr.4: 165041520-165041520 on Build GRCh38
rs140063171 Chr.4: 165041760-165041760 on Build GRCh38
rs201584034 Chr.4: 165041465-165041465 on Build GRCh38
rs73007605 Chr.4: 165041666-165041666 on Build GRCh38
rs762146450 Chr.4: 165041518-165041518 on Build GRCh38
rs780234367 Chr.4: 165041587-165041587 on Build GRCh38
rs766489991 Chr.4: 165041526-165041526 on Build GRCh38
rs937593492 Chr.4: 165041781-165041781 on Build GRCh38
rs894965242 Chr.4: 165041670-165041670 on Build GRCh38
rs570660301 Chr.4: 165041604-165041604 on Build GRCh38
rs954725322 Chr.4: 165041646-165041646 on Build GRCh38
rs756882262 Chr.4: 165041457-165041457 on Build GRCh38
rs755642848 Chr.4: 165041772-165041775 on Build GRCh38
rs769114256 Chr.4: 165041485-165041485 on Build GRCh38
rs1007920073 Chr.4: 165041767-165041767 on Build GRCh38
rs975328391 Chr.4: 165041590-165041590 on Build GRCh38
rs903792721 Chr.4: 165041738-165041738 on Build GRCh38
rs759952728 Chr.4: 165041530-165041531 on Build GRCh38
rs890827511 Chr.4: 165041763-165041763 on Build GRCh38
rs202216539 Chr.4: 165041748-165041748 on Build GRCh38
rs964222938 Chr.4: 165041862-165041862 on Build GRCh38
rs778604867 Chr.4: 165041459-165041459 on Build GRCh38
rs188499338 Chr.4: 165041828-165041828 on Build GRCh38
rs963921699 Chr.4: 165041564-165041564 on Build GRCh38
rs73007606 Chr.4: 165041749-165041749 on Build GRCh38
rs371098223 Chr.4: 165041514-165041514 on Build GRCh38
rs779482704 Chr.4: 165041470-165041470 on Build GRCh38
rs947903838 Chr.4: 165041710-165041710 on Build GRCh38
rs916900492 Chr.4: 165041814-165041814 on Build GRCh38
rs547101809 Chr.4: 165041739-165041739 on Build GRCh38
rs535513720 Chr.4: 165041749-165041750 on Build GRCh38
rs375612994 Chr.4: 165041528-165041528 on Build GRCh38
rs913339473 Chr.4: 165041711-165041711 on Build GRCh38
rs763053812 Chr.4: 165041434-165041434 on Build GRCh38
rs756055313 Chr.4: 165041446-165041446 on Build GRCh38
rs563565592 Chr.4: 165041460-165041460 on Build GRCh38
rs777713015 Chr.4: 165041447-165041447 on Build GRCh38
rs766879039 Chr.4: 165041481-165041481 on Build GRCh38
rs777004830 Chr.4: 165041504-165041504 on Build GRCh38
rs879851745 Chr.4: 165041558-165041565 on Build GRCh38
rs773474719 Chr.4: 165041519-165041519 on Build GRCh38
rs1018466480 Chr.4: 165041835-165041835 on Build GRCh38

View Full Product Details