Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 89333363-89333951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF804B
Entrez Gene ID
219578
Gene Name
zinc finger protein 804B
Gene Aliases
-
UniGene
Hs.684410
Gene Chromosome Location
Chr.7: 88759368-89337057 on Build GRCh38
Transcripts
NM_181646.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7399707 c.794G>A p.C265Y Chr.7: 89333776-89333776 on Build GRCh38
6833315 c.617C>T p.T206I Chr.7: 89333599-89333599 on Build GRCh38
4514068 c.961C>T p.H321Y Chr.7: 89333943-89333943 on Build GRCh38
6110849 c.448A>T p.I150F Chr.7: 89333430-89333430 on Build GRCh38
4987559 c.743G>A p.C248Y Chr.7: 89333725-89333725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750948643 Chr.7: 89333516-89333516 on Build GRCh38
rs761078937 Chr.7: 89333916-89333916 on Build GRCh38
rs1041562958 Chr.7: 89333505-89333505 on Build GRCh38
rs145877596 Chr.7: 89333764-89333764 on Build GRCh38
rs761587371 Chr.7: 89333827-89333827 on Build GRCh38
rs755291781 Chr.7: 89333596-89333596 on Build GRCh38
rs368157318 Chr.7: 89333791-89333791 on Build GRCh38
rs773501845 Chr.7: 89333568-89333568 on Build GRCh38
rs543127229 Chr.7: 89333578-89333578 on Build GRCh38
rs777906157 Chr.7: 89333706-89333706 on Build GRCh38
rs977415085 Chr.7: 89333597-89333597 on Build GRCh38
rs756343735 Chr.7: 89333683-89333683 on Build GRCh38
rs759384329 Chr.7: 89333424-89333424 on Build GRCh38
rs775191182 Chr.7: 89333408-89333408 on Build GRCh38
rs954348307 Chr.7: 89333924-89333924 on Build GRCh38
rs781202005 Chr.7: 89333526-89333526 on Build GRCh38
rs771609302 Chr.7: 89333738-89333738 on Build GRCh38
rs985856022 Chr.7: 89333558-89333558 on Build GRCh38
rs376211299 Chr.7: 89333637-89333637 on Build GRCh38
rs760322171 Chr.7: 89333552-89333552 on Build GRCh38
rs1015735068 Chr.7: 89333867-89333867 on Build GRCh38
rs138598777 Chr.7: 89333790-89333790 on Build GRCh38
rs777615058 Chr.7: 89333878-89333878 on Build GRCh38
rs772523537 Chr.7: 89333908-89333908 on Build GRCh38
rs753057937 Chr.7: 89333681-89333681 on Build GRCh38
rs764779090 Chr.7: 89333772-89333772 on Build GRCh38
rs753765369 Chr.7: 89333756-89333758 on Build GRCh38
rs150010780 Chr.7: 89333554-89333554 on Build GRCh38
rs749438461 Chr.7: 89333710-89333710 on Build GRCh38
rs151146081 Chr.7: 89333803-89333803 on Build GRCh38
rs776309098 Chr.7: 89333748-89333748 on Build GRCh38
rs759020919 Chr.7: 89333904-89333904 on Build GRCh38
rs1003965867 Chr.7: 89333833-89333833 on Build GRCh38
rs1028974235 Chr.7: 89333906-89333906 on Build GRCh38
rs767277662 Chr.7: 89333434-89333434 on Build GRCh38
rs761439054 Chr.7: 89333565-89333565 on Build GRCh38
rs772556687 Chr.7: 89333386-89333386 on Build GRCh38
rs772003003 Chr.7: 89333617-89333617 on Build GRCh38
rs768316286 Chr.7: 89333622-89333622 on Build GRCh38
rs761631428 Chr.7: 89333853-89333853 on Build GRCh38
rs184558452 Chr.7: 89333829-89333829 on Build GRCh38
rs989331135 Chr.7: 89333879-89333879 on Build GRCh38
rs774817274 Chr.7: 89333692-89333692 on Build GRCh38
rs952466840 Chr.7: 89333531-89333531 on Build GRCh38
rs953968601 Chr.7: 89333792-89333792 on Build GRCh38
rs770858340 Chr.7: 89333420-89333420 on Build GRCh38
rs775370152 Chr.7: 89333545-89333545 on Build GRCh38
rs139327261 Chr.7: 89333527-89333527 on Build GRCh38
rs888184927 Chr.7: 89333458-89333458 on Build GRCh38
rs774321079 Chr.7: 89333544-89333544 on Build GRCh38
rs937726880 Chr.7: 89333724-89333724 on Build GRCh38
rs965526921 Chr.7: 89333892-89333892 on Build GRCh38
rs555838660 Chr.7: 89333591-89333591 on Build GRCh38
rs774960884 Chr.7: 89333774-89333774 on Build GRCh38
rs114789722 Chr.7: 89333727-89333727 on Build GRCh38
rs374226876 Chr.7: 89333783-89333783 on Build GRCh38
rs747219977 Chr.7: 89333399-89333399 on Build GRCh38
rs775964468 Chr.7: 89333910-89333910 on Build GRCh38
rs766227200 Chr.7: 89333654-89333655 on Build GRCh38
rs915366636 Chr.7: 89333457-89333457 on Build GRCh38
rs139681818 Chr.7: 89333610-89333610 on Build GRCh38
rs754165461 Chr.7: 89333922-89333922 on Build GRCh38
rs772992744 Chr.7: 89333755-89333755 on Build GRCh38
rs369408292 Chr.7: 89333603-89333603 on Build GRCh38
rs751955064 Chr.7: 89333594-89333594 on Build GRCh38
rs752384427 Chr.7: 89333854-89333868 on Build GRCh38
rs781114062 Chr.7: 89333868-89333868 on Build GRCh38
rs145233331 Chr.7: 89333574-89333574 on Build GRCh38
rs765783436 Chr.7: 89333769-89333769 on Build GRCh38
rs754880145 Chr.7: 89333866-89333866 on Build GRCh38
rs753423387 Chr.7: 89333564-89333564 on Build GRCh38
rs988890423 Chr.7: 89333601-89333601 on Build GRCh38
rs370263764 Chr.7: 89333753-89333753 on Build GRCh38
rs544119286 Chr.7: 89333642-89333642 on Build GRCh38
rs947203435 Chr.7: 89333467-89333467 on Build GRCh38
rs778894435 Chr.7: 89333863-89333863 on Build GRCh38
rs201970697 Chr.7: 89333626-89333626 on Build GRCh38
rs769850704 Chr.7: 89333538-89333538 on Build GRCh38
rs571833966 Chr.7: 89333875-89333875 on Build GRCh38
rs761738065 Chr.7: 89333417-89333417 on Build GRCh38
rs199767132 Chr.7: 89333598-89333598 on Build GRCh38
rs769696807 Chr.7: 89333876-89333876 on Build GRCh38
rs761593394 Chr.7: 89333629-89333629 on Build GRCh38
rs147649508 Chr.7: 89333659-89333659 on Build GRCh38
rs989859642 Chr.7: 89333927-89333927 on Build GRCh38
rs34688143 Chr.7: 89333738-89333739 on Build GRCh38
rs1005726305 Chr.7: 89333760-89333760 on Build GRCh38
rs758040012 Chr.7: 89333906-89333907 on Build GRCh38
rs758986998 Chr.7: 89333760-89333783 on Build GRCh38
rs1916830 Chr.7: 89333725-89333725 on Build GRCh38
rs1041508328 Chr.7: 89333653-89333653 on Build GRCh38
rs762482573 Chr.7: 89333652-89333652 on Build GRCh38
rs372640521 Chr.7: 89333397-89333397 on Build GRCh38
rs767741505 Chr.7: 89333679-89333679 on Build GRCh38
rs187269414 Chr.7: 89333513-89333513 on Build GRCh38
rs773989027 Chr.7: 89333797-89333797 on Build GRCh38
rs760398588 Chr.7: 89333446-89333446 on Build GRCh38
rs771383465 Chr.7: 89333635-89333635 on Build GRCh38
rs370604248 Chr.7: 89333674-89333674 on Build GRCh38
rs749309800 Chr.7: 89333616-89333616 on Build GRCh38
rs749687808 Chr.7: 89333614-89333614 on Build GRCh38
rs1012981644 Chr.7: 89333800-89333800 on Build GRCh38
rs780647075 Chr.7: 89333584-89333584 on Build GRCh38
rs768716684 Chr.7: 89333406-89333406 on Build GRCh38
rs768602003 Chr.7: 89333747-89333747 on Build GRCh38
rs764360005 Chr.7: 89333813-89333813 on Build GRCh38
rs768934748 Chr.7: 89333524-89333524 on Build GRCh38
rs752449749 Chr.7: 89333439-89333439 on Build GRCh38
rs372995954 Chr.7: 89333515-89333515 on Build GRCh38
rs746303492 Chr.7: 89333384-89333384 on Build GRCh38
rs137963601 Chr.7: 89333903-89333903 on Build GRCh38
rs764413366 Chr.7: 89333921-89333921 on Build GRCh38
rs913302881 Chr.7: 89333891-89333891 on Build GRCh38
rs747890840 Chr.7: 89333750-89333750 on Build GRCh38
rs111719576 Chr.7: 89333896-89333896 on Build GRCh38
rs915298289 Chr.7: 89333632-89333632 on Build GRCh38
rs145219401 Chr.7: 89333621-89333621 on Build GRCh38
rs773131730 Chr.7: 89333542-89333543 on Build GRCh38
rs745915610 Chr.7: 89333864-89333864 on Build GRCh38
rs372282374 Chr.7: 89333897-89333897 on Build GRCh38
rs150328762 Chr.7: 89333884-89333884 on Build GRCh38
rs755417691 Chr.7: 89333662-89333662 on Build GRCh38
rs760493101 Chr.7: 89333655-89333655 on Build GRCh38
rs957367931 Chr.7: 89333852-89333852 on Build GRCh38
rs370064148 Chr.7: 89333501-89333501 on Build GRCh38
rs150823294 Chr.7: 89333517-89333517 on Build GRCh38
rs140043558 Chr.7: 89333809-89333809 on Build GRCh38
rs540004551 Chr.7: 89333618-89333618 on Build GRCh38
rs572989562 Chr.7: 89333608-89333608 on Build GRCh38
rs746882193 Chr.7: 89333620-89333620 on Build GRCh38

View Full Product Details