Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 56359114-56359707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZWINT
Entrez Gene ID
11130
Gene Name
ZW10 interacting kinetochore protein
Gene Aliases
HZwint-1, KNTC2AP, SIP30, ZWINT1
UniGene
Hs.591363
Gene Chromosome Location
Chr.10: 56357257-56361302 on Build GRCh38
Transcripts
NM_001005413.1 NM_007057.3 NM_032997.2 XM_017015605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603570 c.474A>G p.L158L Chr.10: 56359482-56359482 on Build GRCh38
1348337 c.433G>A p.A145T Chr.10: 56359523-56359523 on Build GRCh38
4014815 c.455T>C p.V152A Chr.10: 56359501-56359501 on Build GRCh38
4495588 c.456C>T p.V152V Chr.10: 56359500-56359500 on Build GRCh38
7152570 c.424-2A>G p.? Chr.10: 56359534-56359534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750116427 Chr.10: 56359500-56359500 on Build GRCh38
rs957857669 Chr.10: 56359142-56359142 on Build GRCh38
rs779842036 Chr.10: 56359510-56359510 on Build GRCh38
rs1042377658 Chr.10: 56359338-56359338 on Build GRCh38
rs757464937 Chr.10: 56359658-56359658 on Build GRCh38
rs770167450 Chr.10: 56359460-56359460 on Build GRCh38
rs541979989 Chr.10: 56359188-56359188 on Build GRCh38
rs767896120 Chr.10: 56359479-56359479 on Build GRCh38
rs373707810 Chr.10: 56359527-56359527 on Build GRCh38
rs561862977 Chr.10: 56359195-56359195 on Build GRCh38
rs944930259 Chr.10: 56359328-56359328 on Build GRCh38
rs1021422331 Chr.10: 56359147-56359147 on Build GRCh38
rs1017505173 Chr.10: 56359208-56359208 on Build GRCh38
rs746575900 Chr.10: 56359511-56359511 on Build GRCh38
rs557511244 Chr.10: 56359593-56359593 on Build GRCh38
rs761150337 Chr.10: 56359242-56359242 on Build GRCh38
rs768353445 Chr.10: 56359453-56359453 on Build GRCh38
rs760084464 Chr.10: 56359678-56359678 on Build GRCh38
rs1027522102 Chr.10: 56359597-56359597 on Build GRCh38
rs758424425 Chr.10: 56359506-56359506 on Build GRCh38
rs572257534 Chr.10: 56359344-56359344 on Build GRCh38
rs902153530 Chr.10: 56359340-56359340 on Build GRCh38
rs929108777 Chr.10: 56359305-56359305 on Build GRCh38
rs527759088 Chr.10: 56359281-56359281 on Build GRCh38
rs758885193 Chr.10: 56359674-56359674 on Build GRCh38
rs920433430 Chr.10: 56359290-56359290 on Build GRCh38
rs775650850 Chr.10: 56359681-56359681 on Build GRCh38
rs371404579 Chr.10: 56359467-56359467 on Build GRCh38
rs1044437674 Chr.10: 56359397-56359397 on Build GRCh38
rs570469710 Chr.10: 56359231-56359233 on Build GRCh38
rs769369963 Chr.10: 56359547-56359547 on Build GRCh38
rs577565804 Chr.10: 56359615-56359615 on Build GRCh38
rs1017577791 Chr.10: 56359507-56359507 on Build GRCh38
rs202029427 Chr.10: 56359434-56359434 on Build GRCh38
rs747814067 Chr.10: 56359542-56359542 on Build GRCh38
rs76978115 Chr.10: 56359431-56359431 on Build GRCh38
rs776243327 Chr.10: 56359456-56359456 on Build GRCh38
rs576233791 Chr.10: 56359148-56359148 on Build GRCh38
rs890652475 Chr.10: 56359310-56359310 on Build GRCh38
rs982023658 Chr.10: 56359165-56359165 on Build GRCh38
rs753861072 Chr.10: 56359421-56359421 on Build GRCh38
rs760980079 Chr.10: 56359682-56359682 on Build GRCh38
rs749604801 Chr.10: 56359576-56359576 on Build GRCh38
rs765133248 Chr.10: 56359488-56359488 on Build GRCh38
rs746495051 Chr.10: 56359444-56359444 on Build GRCh38
rs191476918 Chr.10: 56359468-56359468 on Build GRCh38
rs775872463 Chr.10: 56359216-56359216 on Build GRCh38
rs983256920 Chr.10: 56359339-56359339 on Build GRCh38
rs547502633 Chr.10: 56359351-56359351 on Build GRCh38
rs953425390 Chr.10: 56359520-56359520 on Build GRCh38
rs562790549 Chr.10: 56359145-56359145 on Build GRCh38
rs534704030 Chr.10: 56359552-56359552 on Build GRCh38
rs983670214 Chr.10: 56359621-56359621 on Build GRCh38
rs1021063189 Chr.10: 56359448-56359448 on Build GRCh38
rs373790438 Chr.10: 56359418-56359418 on Build GRCh38
rs550095575 Chr.10: 56359435-56359435 on Build GRCh38
rs916981934 Chr.10: 56359354-56359354 on Build GRCh38
rs2393069 Chr.10: 56359144-56359144 on Build GRCh38
rs763844460 Chr.10: 56359269-56359269 on Build GRCh38
rs973272497 Chr.10: 56359253-56359253 on Build GRCh38
rs748550854 Chr.10: 56359459-56359459 on Build GRCh38
rs865985201 Chr.10: 56359215-56359215 on Build GRCh38
rs565656259 Chr.10: 56359518-56359518 on Build GRCh38
rs778807461 Chr.10: 56359430-56359430 on Build GRCh38
rs949769574 Chr.10: 56359382-56359382 on Build GRCh38
rs376513163 Chr.10: 56359470-56359470 on Build GRCh38
rs753003775 Chr.10: 56359480-56359480 on Build GRCh38
rs1011974232 Chr.10: 56359414-56359414 on Build GRCh38
rs893542951 Chr.10: 56359410-56359411 on Build GRCh38
rs567157398 Chr.10: 56359172-56359172 on Build GRCh38
rs771521511 Chr.10: 56359583-56359583 on Build GRCh38
rs994723590 Chr.10: 56359546-56359546 on Build GRCh38
rs773392178 Chr.10: 56359463-56359463 on Build GRCh38
rs912037697 Chr.10: 56359608-56359608 on Build GRCh38
rs375548084 Chr.10: 56359662-56359662 on Build GRCh38
rs973802310 Chr.10: 56359491-56359491 on Build GRCh38
rs58152081 Chr.10: 56359679-56359679 on Build GRCh38
rs74876352 Chr.10: 56359657-56359657 on Build GRCh38
rs999602103 Chr.10: 56359350-56359350 on Build GRCh38
rs756352555 Chr.10: 56359487-56359487 on Build GRCh38
rs879268061 Chr.10: 56359402-56359402 on Build GRCh38
rs964612379 Chr.10: 56359225-56359225 on Build GRCh38

View Full Product Details