Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57456403-57456967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBE
Entrez Gene ID
83729
Gene Name
inhibin beta E subunit
Gene Aliases
-
UniGene
Hs.632713
Gene Chromosome Location
Chr.12: 57455313-57458008 on Build GRCh38
Transcripts
NM_031479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1988936 c.725C>A p.P242H Chr.12: 57456520-57456520 on Build GRCh38
4381988 c.920C>G p.P307R Chr.12: 57456715-57456715 on Build GRCh38
1988934 c.641A>G p.H214R Chr.12: 57456436-57456436 on Build GRCh38
42719 c.649C>T p.P217S Chr.12: 57456444-57456444 on Build GRCh38
240231 c.742T>C p.C248R Chr.12: 57456537-57456537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754703774 Chr.12: 57456483-57456483 on Build GRCh38
rs774834771 Chr.12: 57456433-57456433 on Build GRCh38
rs759206435 Chr.12: 57456555-57456555 on Build GRCh38
rs148453718 Chr.12: 57456758-57456758 on Build GRCh38
rs765376541 Chr.12: 57456719-57456719 on Build GRCh38
rs372615182 Chr.12: 57456580-57456580 on Build GRCh38
rs772700168 Chr.12: 57456436-57456436 on Build GRCh38
rs763025380 Chr.12: 57456887-57456887 on Build GRCh38
rs745438889 Chr.12: 57456788-57456788 on Build GRCh38
rs554478218 Chr.12: 57456870-57456870 on Build GRCh38
rs902691880 Chr.12: 57456666-57456666 on Build GRCh38
rs780392794 Chr.12: 57456734-57456734 on Build GRCh38
rs751561858 Chr.12: 57456719-57456720 on Build GRCh38
rs760033547 Chr.12: 57456435-57456435 on Build GRCh38
rs761389641 Chr.12: 57456495-57456495 on Build GRCh38
rs553356284 Chr.12: 57456572-57456572 on Build GRCh38
rs753856716 Chr.12: 57456591-57456591 on Build GRCh38
rs776145331 Chr.12: 57456461-57456461 on Build GRCh38
rs763800579 Chr.12: 57456710-57456710 on Build GRCh38
rs146517777 Chr.12: 57456554-57456554 on Build GRCh38
rs997771765 Chr.12: 57456606-57456606 on Build GRCh38
rs760258476 Chr.12: 57456805-57456805 on Build GRCh38
rs747468719 Chr.12: 57456634-57456634 on Build GRCh38
rs765325146 Chr.12: 57456878-57456878 on Build GRCh38
rs538464070 Chr.12: 57456515-57456515 on Build GRCh38
rs770320971 Chr.12: 57456762-57456762 on Build GRCh38
rs867532642 Chr.12: 57456718-57456719 on Build GRCh38
rs201477879 Chr.12: 57456466-57456466 on Build GRCh38
rs578050663 Chr.12: 57456932-57456932 on Build GRCh38
rs1005647635 Chr.12: 57456736-57456736 on Build GRCh38
rs772587242 Chr.12: 57456544-57456544 on Build GRCh38
rs373832184 Chr.12: 57456737-57456737 on Build GRCh38
rs1014734661 Chr.12: 57456886-57456886 on Build GRCh38
rs775132929 Chr.12: 57456690-57456690 on Build GRCh38
rs963243359 Chr.12: 57456921-57456921 on Build GRCh38
rs776084211 Chr.12: 57456437-57456437 on Build GRCh38
rs998111101 Chr.12: 57456778-57456778 on Build GRCh38
rs543618544 Chr.12: 57456912-57456912 on Build GRCh38
rs576228282 Chr.12: 57456871-57456871 on Build GRCh38
rs375663495 Chr.12: 57456677-57456677 on Build GRCh38
rs953889231 Chr.12: 57456895-57456895 on Build GRCh38
rs772819150 Chr.12: 57456459-57456459 on Build GRCh38
rs144527638 Chr.12: 57456754-57456754 on Build GRCh38
rs755981328 Chr.12: 57456579-57456579 on Build GRCh38
rs536111730 Chr.12: 57456523-57456523 on Build GRCh38
rs777681034 Chr.12: 57456495-57456495 on Build GRCh38
rs766194668 Chr.12: 57456644-57456655 on Build GRCh38
rs143404510 Chr.12: 57456516-57456516 on Build GRCh38
rs761873601 Chr.12: 57456715-57456715 on Build GRCh38
rs764881360 Chr.12: 57456518-57456518 on Build GRCh38
rs746091653 Chr.12: 57456522-57456522 on Build GRCh38
rs989918842 Chr.12: 57456941-57456941 on Build GRCh38
rs749434469 Chr.12: 57456500-57456500 on Build GRCh38
rs571755879 Chr.12: 57456600-57456600 on Build GRCh38
rs183394512 Chr.12: 57456490-57456490 on Build GRCh38
rs755315461 Chr.12: 57456741-57456741 on Build GRCh38
rs377350183 Chr.12: 57456806-57456806 on Build GRCh38
rs750530215 Chr.12: 57456883-57456883 on Build GRCh38
rs370691355 Chr.12: 57456854-57456854 on Build GRCh38
rs769043643 Chr.12: 57456641-57456641 on Build GRCh38
rs1032265309 Chr.12: 57456477-57456477 on Build GRCh38
rs747443276 Chr.12: 57456543-57456543 on Build GRCh38
rs746048975 Chr.12: 57456608-57456608 on Build GRCh38
rs1005381126 Chr.12: 57456869-57456869 on Build GRCh38
rs777131104 Chr.12: 57456659-57456659 on Build GRCh38
rs775142337 Chr.12: 57456798-57456798 on Build GRCh38
rs549735432 Chr.12: 57456489-57456489 on Build GRCh38
rs753750949 Chr.12: 57456714-57456714 on Build GRCh38
rs1034292154 Chr.12: 57456935-57456935 on Build GRCh38
rs1030452932 Chr.12: 57456497-57456497 on Build GRCh38
rs992970086 Chr.12: 57456573-57456573 on Build GRCh38
rs533795674 Chr.12: 57456640-57456640 on Build GRCh38
rs762481508 Chr.12: 57456596-57456596 on Build GRCh38
rs192076894 Chr.12: 57456829-57456829 on Build GRCh38
rs779157178 Chr.12: 57456602-57456602 on Build GRCh38
rs748327531 Chr.12: 57456768-57456768 on Build GRCh38
rs758681685 Chr.12: 57456731-57456731 on Build GRCh38
rs571336896 Chr.12: 57456548-57456548 on Build GRCh38
rs140813283 Chr.12: 57456928-57456928 on Build GRCh38
rs201023614 Chr.12: 57456484-57456484 on Build GRCh38
rs758734692 Chr.12: 57456616-57456616 on Build GRCh38
rs772658213 Chr.12: 57456545-57456545 on Build GRCh38
rs910604131 Chr.12: 57456524-57456524 on Build GRCh38
rs778794069 Chr.12: 57456695-57456695 on Build GRCh38
rs937029805 Chr.12: 57456558-57456558 on Build GRCh38
rs771617277 Chr.12: 57456795-57456795 on Build GRCh38
rs751353200 Chr.12: 57456472-57456472 on Build GRCh38
rs370912843 Chr.12: 57456687-57456687 on Build GRCh38
rs775956626 Chr.12: 57456535-57456535 on Build GRCh38
rs954680971 Chr.12: 57456506-57456506 on Build GRCh38
rs138708295 Chr.12: 57456686-57456686 on Build GRCh38
rs762494353 Chr.12: 57456465-57456465 on Build GRCh38
rs142754901 Chr.12: 57456720-57456720 on Build GRCh38
rs761269326 Chr.12: 57456447-57456447 on Build GRCh38
rs977019815 Chr.12: 57456475-57456475 on Build GRCh38
rs138960179 Chr.12: 57456455-57456455 on Build GRCh38
rs778370434 Chr.12: 57456764-57456764 on Build GRCh38
rs780261059 Chr.12: 57456617-57456617 on Build GRCh38

View Full Product Details