Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55966942-55967361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK2
Entrez Gene ID
1017
Gene Name
cyclin dependent kinase 2
Gene Aliases
CDKN2, p33(CDK2)
UniGene
Hs.19192
Gene Chromosome Location
Chr.12: 55966769-55972789 on Build GRCh38
Transcripts
NM_001290230.1 NM_001798.4 NM_052827.3 XM_011537732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019698 c.84G>A p.E28E Chr.12: 55967092-55967092 on Build GRCh38
5017284 c.35delA p.E12fs*14 Chr.12: 55967043-55967043 on Build GRCh38
4671130 c.37G>T p.G13C Chr.12: 55967045-55967045 on Build GRCh38
3359837 c.58A>G p.K20E Chr.12: 55967066-55967066 on Build GRCh38
941454 c.101A>C p.K34T Chr.12: 55967109-55967109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757007587 Chr.12: 55967132-55967132 on Build GRCh38
rs746850184 Chr.12: 55966976-55966976 on Build GRCh38
rs959791678 Chr.12: 55967222-55967222 on Build GRCh38
rs2069397 Chr.12: 55966971-55966971 on Build GRCh38
rs770551803 Chr.12: 55966977-55966977 on Build GRCh38
rs996801189 Chr.12: 55967157-55967157 on Build GRCh38
rs571053619 Chr.12: 55967050-55967050 on Build GRCh38
rs1015177588 Chr.12: 55967187-55967187 on Build GRCh38
rs953108225 Chr.12: 55967271-55967271 on Build GRCh38
rs759688421 Chr.12: 55967083-55967083 on Build GRCh38
rs772494885 Chr.12: 55967016-55967016 on Build GRCh38
rs766396000 Chr.12: 55967175-55967175 on Build GRCh38
rs113816950 Chr.12: 55967054-55967054 on Build GRCh38
rs961143148 Chr.12: 55967194-55967194 on Build GRCh38
rs917952705 Chr.12: 55967000-55967000 on Build GRCh38
rs949391273 Chr.12: 55967019-55967019 on Build GRCh38
rs773811074 Chr.12: 55967173-55967173 on Build GRCh38
rs774877336 Chr.12: 55967001-55967001 on Build GRCh38
rs995212755 Chr.12: 55967202-55967202 on Build GRCh38
rs139342756 Chr.12: 55967017-55967017 on Build GRCh38
rs758327721 Chr.12: 55967113-55967113 on Build GRCh38
rs1008329090 Chr.12: 55967180-55967180 on Build GRCh38
rs535976635 Chr.12: 55967213-55967213 on Build GRCh38
rs751524550 Chr.12: 55967131-55967131 on Build GRCh38
rs780775267 Chr.12: 55966980-55966980 on Build GRCh38
rs761072055 Chr.12: 55967038-55967038 on Build GRCh38
rs978095560 Chr.12: 55967275-55967275 on Build GRCh38
rs112682211 Chr.12: 55967117-55967117 on Build GRCh38
rs777622443 Chr.12: 55966972-55966972 on Build GRCh38
rs1014802330 Chr.12: 55967226-55967226 on Build GRCh38
rs568622337 Chr.12: 55967175-55967175 on Build GRCh38
rs936779600 Chr.12: 55967328-55967328 on Build GRCh38
rs769282649 Chr.12: 55966997-55966997 on Build GRCh38
rs3087335 Chr.12: 55967052-55967052 on Build GRCh38
rs11554376 Chr.12: 55967060-55967060 on Build GRCh38
rs1027132217 Chr.12: 55967211-55967211 on Build GRCh38
rs989547278 Chr.12: 55967330-55967330 on Build GRCh38
rs745467761 Chr.12: 55967141-55967141 on Build GRCh38
rs762376662 Chr.12: 55967004-55967004 on Build GRCh38
rs750300173 Chr.12: 55967179-55967179 on Build GRCh38
rs892972897 Chr.12: 55967119-55967119 on Build GRCh38
rs956857141 Chr.12: 55967327-55967328 on Build GRCh38
rs1048878253 Chr.12: 55967171-55967171 on Build GRCh38
rs539412785 Chr.12: 55967320-55967320 on Build GRCh38
rs557458862 Chr.12: 55967221-55967221 on Build GRCh38
rs12823915 Chr.12: 55967258-55967258 on Build GRCh38
rs769391479 Chr.12: 55967144-55967144 on Build GRCh38
rs369499858 Chr.12: 55967150-55967150 on Build GRCh38
rs1032515463 Chr.12: 55967323-55967323 on Build GRCh38
rs376551262 Chr.12: 55966981-55966981 on Build GRCh38
rs751307131 Chr.12: 55967174-55967175 on Build GRCh38
rs2069398 Chr.12: 55967092-55967092 on Build GRCh38
rs752789935 Chr.12: 55967095-55967095 on Build GRCh38
rs569308993 Chr.12: 55967267-55967267 on Build GRCh38
rs777189859 Chr.12: 55967176-55967176 on Build GRCh38
rs200863916 Chr.12: 55967163-55967163 on Build GRCh38
rs755065253 Chr.12: 55966967-55966967 on Build GRCh38
rs780969766 Chr.12: 55967136-55967136 on Build GRCh38
rs754042510 Chr.12: 55967061-55967061 on Build GRCh38
rs1028689326 Chr.12: 55967270-55967270 on Build GRCh38
rs936706017 Chr.12: 55967086-55967086 on Build GRCh38
rs990974243 Chr.12: 55967227-55967227 on Build GRCh38
rs376599347 Chr.12: 55967124-55967124 on Build GRCh38

View Full Product Details