Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95966217-95966781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
UniGene
Hs.424907
Gene Chromosome Location
Chr.12: 95943293-95968592 on Build GRCh38
Transcripts
NM_152435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5986954 c.1141G>C p.G381R Chr.12: 95966456-95966456 on Build GRCh38
944714 c.1117G>A p.A373T Chr.12: 95966432-95966432 on Build GRCh38
6553608 c.1093G>T p.A365S Chr.12: 95966408-95966408 on Build GRCh38
3466638 c.1166G>A p.G389E Chr.12: 95966481-95966481 on Build GRCh38
432023 c.1138C>A p.H380N Chr.12: 95966453-95966453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025607 Chr.12: 95966728-95966728 on Build GRCh38
rs756177512 Chr.12: 95966450-95966450 on Build GRCh38
rs761263883 Chr.12: 95966474-95966474 on Build GRCh38
rs746787781 Chr.12: 95966466-95966466 on Build GRCh38
rs764758506 Chr.12: 95966481-95966481 on Build GRCh38
rs371734841 Chr.12: 95966538-95966538 on Build GRCh38
rs892213540 Chr.12: 95966724-95966724 on Build GRCh38
rs79312289 Chr.12: 95966377-95966377 on Build GRCh38
rs768472516 Chr.12: 95966468-95966468 on Build GRCh38
rs200832722 Chr.12: 95966732-95966732 on Build GRCh38
rs374697669 Chr.12: 95966345-95966345 on Build GRCh38
rs773608825 Chr.12: 95966485-95966485 on Build GRCh38
rs1056739393 Chr.12: 95966268-95966268 on Build GRCh38
rs996899462 Chr.12: 95966534-95966534 on Build GRCh38
rs776173932 Chr.12: 95966550-95966550 on Build GRCh38
rs781449069 Chr.12: 95966434-95966434 on Build GRCh38
rs922044644 Chr.12: 95966269-95966269 on Build GRCh38
rs751700607 Chr.12: 95966328-95966328 on Build GRCh38
rs267603727 Chr.12: 95966503-95966503 on Build GRCh38
rs781312532 Chr.12: 95966351-95966351 on Build GRCh38
rs202031881 Chr.12: 95966346-95966346 on Build GRCh38
rs141724326 Chr.12: 95966508-95966508 on Build GRCh38
rs879023574 Chr.12: 95966290-95966290 on Build GRCh38
rs769986017 Chr.12: 95966444-95966445 on Build GRCh38
rs142306346 Chr.12: 95966464-95966464 on Build GRCh38
rs755248408 Chr.12: 95966430-95966430 on Build GRCh38
rs564079646 Chr.12: 95966256-95966256 on Build GRCh38
rs779462844 Chr.12: 95966309-95966309 on Build GRCh38
rs778472842 Chr.12: 95966727-95966727 on Build GRCh38
rs968042239 Chr.12: 95966666-95966666 on Build GRCh38
rs748292264 Chr.12: 95966383-95966383 on Build GRCh38
rs908682797 Chr.12: 95966754-95966754 on Build GRCh38
rs748216029 Chr.12: 95966447-95966447 on Build GRCh38
rs753347919 Chr.12: 95966390-95966390 on Build GRCh38
rs191163722 Chr.12: 95966404-95966404 on Build GRCh38
rs778433337 Chr.12: 95966446-95966447 on Build GRCh38
rs780341874 Chr.12: 95966411-95966411 on Build GRCh38
rs1022270062 Chr.12: 95966642-95966642 on Build GRCh38
rs745740133 Chr.12: 95966460-95966460 on Build GRCh38
rs182467786 Chr.12: 95966240-95966240 on Build GRCh38
rs17024908 Chr.12: 95966533-95966533 on Build GRCh38
rs572326991 Chr.12: 95966238-95966238 on Build GRCh38
rs763835378 Chr.12: 95966385-95966385 on Build GRCh38
rs936318332 Chr.12: 95966709-95966709 on Build GRCh38
rs35450680 Chr.12: 95966439-95966440 on Build GRCh38
rs996581752 Chr.12: 95966322-95966322 on Build GRCh38
rs1053570918 Chr.12: 95966721-95966721 on Build GRCh38
rs780288885 Chr.12: 95966323-95966323 on Build GRCh38
rs372012329 Chr.12: 95966358-95966358 on Build GRCh38
rs767440686 Chr.12: 95966331-95966335 on Build GRCh38
rs766854784 Chr.12: 95966494-95966494 on Build GRCh38
rs777841138 Chr.12: 95966509-95966509 on Build GRCh38
rs188271815 Chr.12: 95966675-95966675 on Build GRCh38
rs1025606 Chr.12: 95966572-95966572 on Build GRCh38
rs77796595 Chr.12: 95966296-95966296 on Build GRCh38
rs75221805 Chr.12: 95966732-95966732 on Build GRCh38
rs770586555 Chr.12: 95966697-95966697 on Build GRCh38
rs79677920 Chr.12: 95966421-95966421 on Build GRCh38
rs768460847 Chr.12: 95966545-95966545 on Build GRCh38
rs778736935 Chr.12: 95966455-95966455 on Build GRCh38
rs747377365 Chr.12: 95966277-95966277 on Build GRCh38
rs1036266735 Chr.12: 95966348-95966348 on Build GRCh38
rs961880411 Chr.12: 95966548-95966548 on Build GRCh38
rs370171574 Chr.12: 95966624-95966624 on Build GRCh38
rs774067051 Chr.12: 95966631-95966631 on Build GRCh38
rs755301280 Chr.12: 95966499-95966499 on Build GRCh38
rs750905548 Chr.12: 95966407-95966407 on Build GRCh38
rs368214286 Chr.12: 95966357-95966357 on Build GRCh38
rs561476126 Chr.12: 95966299-95966299 on Build GRCh38
rs146251081 Chr.12: 95966354-95966354 on Build GRCh38
rs531222035 Chr.12: 95966289-95966289 on Build GRCh38
rs567800823 Chr.12: 95966604-95966604 on Build GRCh38
rs10507068 Chr.12: 95966695-95966695 on Build GRCh38
rs887352345 Chr.12: 95966281-95966281 on Build GRCh38
rs759394978 Chr.12: 95966663-95966663 on Build GRCh38
rs966281077 Chr.12: 95966453-95966453 on Build GRCh38
rs17024904 Chr.12: 95966394-95966394 on Build GRCh38
rs750805926 Chr.12: 95966310-95966310 on Build GRCh38
rs751915375 Chr.12: 95966415-95966415 on Build GRCh38
rs776320325 Chr.12: 95966470-95966470 on Build GRCh38
rs532564025 Chr.12: 95966623-95966623 on Build GRCh38
rs758734521 Chr.12: 95966314-95966314 on Build GRCh38
rs767663697 Chr.12: 95966505-95966505 on Build GRCh38
rs1057033743 Chr.12: 95966506-95966506 on Build GRCh38
rs374628537 Chr.12: 95966461-95966461 on Build GRCh38
rs747780982 Chr.12: 95966555-95966555 on Build GRCh38
rs771943179 Chr.12: 95966363-95966363 on Build GRCh38
rs749376288 Chr.12: 95966514-95966514 on Build GRCh38
rs145193484 Chr.12: 95966378-95966378 on Build GRCh38
rs151279149 Chr.12: 95966507-95966507 on Build GRCh38
rs763515851 Chr.12: 95966487-95966487 on Build GRCh38
rs139392418 Chr.12: 95966361-95966361 on Build GRCh38
rs1038057038 Chr.12: 95966520-95966520 on Build GRCh38
rs751966597 Chr.12: 95966497-95966497 on Build GRCh38
rs780117745 Chr.12: 95966535-95966535 on Build GRCh38
rs369609960 Chr.12: 95966634-95966634 on Build GRCh38
rs770818203 Chr.12: 95966355-95966355 on Build GRCh38
rs371501532 Chr.12: 95966306-95966306 on Build GRCh38
rs758787672 Chr.12: 95966408-95966408 on Build GRCh38

View Full Product Details