Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53496984-53497568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1D
Entrez Gene ID
776
Gene Name
calcium voltage-gated channel subunit alpha1 D
Gene Aliases
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
UniGene
Hs.476358
Gene Chromosome Location
Chr.3: 53495049-53813151 on Build GRCh38
Transcripts
NM_000720.3 NM_001128839.2 NM_001128840.2 XM_005265448.3 XM_011534094.2 XM_011534096.2 XM_011534100.2 XM_017007137.1 XM_017007138.1 XM_017007139.1 XM_017007140.1 XM_017007141.1 XM_017007142.1 XM_017007143.1 XM_017007144.1 XM_017007145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2780686 c.100C>G p.L34V Chr.3: 53497184-53497184 on Build GRCh38
5128781 c.244C>A p.Q82K Chr.3: 53497328-53497328 on Build GRCh38
2780695 c.201A>G p.Q67Q Chr.3: 53497285-53497285 on Build GRCh38
5983574 c.294C>T p.S98S Chr.3: 53497378-53497378 on Build GRCh38
2154640 c.377+1G>T p.? Chr.3: 53497462-53497462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759586333 Chr.3: 53497112-53497112 on Build GRCh38
rs758400808 Chr.3: 53497298-53497298 on Build GRCh38
rs377363390 Chr.3: 53497216-53497216 on Build GRCh38
rs954756294 Chr.3: 53497060-53497060 on Build GRCh38
rs762132463 Chr.3: 53497253-53497253 on Build GRCh38
rs554379312 Chr.3: 53497046-53497046 on Build GRCh38
rs749334571 Chr.3: 53497502-53497502 on Build GRCh38
rs28365112 Chr.3: 53497348-53497348 on Build GRCh38
rs1007646210 Chr.3: 53497400-53497400 on Build GRCh38
rs775933698 Chr.3: 53497116-53497116 on Build GRCh38
rs541931890 Chr.3: 53497121-53497121 on Build GRCh38
rs764380630 Chr.3: 53497125-53497125 on Build GRCh38
rs150640742 Chr.3: 53497424-53497424 on Build GRCh38
rs777777778 Chr.3: 53497116-53497117 on Build GRCh38
rs190182060 Chr.3: 53497044-53497044 on Build GRCh38
rs772267259 Chr.3: 53497385-53497385 on Build GRCh38
rs28365111 Chr.3: 53497378-53497378 on Build GRCh38
rs781058978 Chr.3: 53497321-53497321 on Build GRCh38
rs764773584 Chr.3: 53497294-53497294 on Build GRCh38
rs780101855 Chr.3: 53497354-53497354 on Build GRCh38
rs752623993 Chr.3: 53497197-53497197 on Build GRCh38
rs147916220 Chr.3: 53497319-53497319 on Build GRCh38
rs766003838 Chr.3: 53497141-53497141 on Build GRCh38
rs759550494 Chr.3: 53497078-53497078 on Build GRCh38
rs888119455 Chr.3: 53497232-53497232 on Build GRCh38
rs774682333 Chr.3: 53497252-53497252 on Build GRCh38
rs564124151 Chr.3: 53497396-53497396 on Build GRCh38
rs900052469 Chr.3: 53497039-53497039 on Build GRCh38
rs751400848 Chr.3: 53497459-53497459 on Build GRCh38
rs185674174 Chr.3: 53497474-53497474 on Build GRCh38
rs140749530 Chr.3: 53497306-53497306 on Build GRCh38
rs762596454 Chr.3: 53497419-53497419 on Build GRCh38
rs575300482 Chr.3: 53497289-53497289 on Build GRCh38
rs917022424 Chr.3: 53497155-53497155 on Build GRCh38
rs543405396 Chr.3: 53497116-53497116 on Build GRCh38
rs1039308287 Chr.3: 53497349-53497349 on Build GRCh38
rs758415159 Chr.3: 53497212-53497212 on Build GRCh38
rs752325493 Chr.3: 53497297-53497297 on Build GRCh38
rs933683401 Chr.3: 53497375-53497375 on Build GRCh38
rs962241848 Chr.3: 53497019-53497019 on Build GRCh38
rs192959416 Chr.3: 53497144-53497144 on Build GRCh38
rs1018065755 Chr.3: 53497418-53497418 on Build GRCh38
rs774878323 Chr.3: 53497377-53497377 on Build GRCh38
rs767957783 Chr.3: 53497267-53497267 on Build GRCh38
rs991468238 Chr.3: 53497139-53497139 on Build GRCh38
rs775119327 Chr.3: 53497414-53497414 on Build GRCh38
rs77610564 Chr.3: 53497127-53497127 on Build GRCh38
rs778510463 Chr.3: 53497174-53497174 on Build GRCh38
rs986157961 Chr.3: 53497066-53497066 on Build GRCh38
rs78820127 Chr.3: 53497126-53497126 on Build GRCh38
rs763722743 Chr.3: 53497384-53497384 on Build GRCh38
rs910544944 Chr.3: 53497110-53497110 on Build GRCh38
rs776373174 Chr.3: 53497446-53497446 on Build GRCh38
rs1016886112 Chr.3: 53497015-53497015 on Build GRCh38
rs746863213 Chr.3: 53497213-53497213 on Build GRCh38
rs751372761 Chr.3: 53497316-53497316 on Build GRCh38
rs765082147 Chr.3: 53497115-53497116 on Build GRCh38
rs769710409 Chr.3: 53497231-53497231 on Build GRCh38
rs1005593744 Chr.3: 53497276-53497276 on Build GRCh38
rs144915770 Chr.3: 53497395-53497395 on Build GRCh38
rs982022592 Chr.3: 53497203-53497203 on Build GRCh38
rs746016693 Chr.3: 53497336-53497336 on Build GRCh38
rs935163999 Chr.3: 53497233-53497233 on Build GRCh38
rs914971602 Chr.3: 53497337-53497337 on Build GRCh38
rs753384864 Chr.3: 53497272-53497272 on Build GRCh38
rs755003019 Chr.3: 53497508-53497508 on Build GRCh38
rs539335706 Chr.3: 53497217-53497217 on Build GRCh38
rs756313438 Chr.3: 53497486-53497486 on Build GRCh38
rs770301253 Chr.3: 53497115-53497115 on Build GRCh38
rs760371924 Chr.3: 53497137-53497137 on Build GRCh38
rs775557387 Chr.3: 53497240-53497240 on Build GRCh38
rs781312928 Chr.3: 53497218-53497218 on Build GRCh38
rs899497986 Chr.3: 53497356-53497356 on Build GRCh38
rs574313334 Chr.3: 53497068-53497068 on Build GRCh38
rs749426004 Chr.3: 53497366-53497366 on Build GRCh38
rs1004362019 Chr.3: 53497437-53497437 on Build GRCh38
rs558061317 Chr.3: 53497478-53497478 on Build GRCh38
rs533046755 Chr.3: 53497397-53497397 on Build GRCh38
rs769218354 Chr.3: 53497246-53497246 on Build GRCh38
rs1039360846 Chr.3: 53497021-53497021 on Build GRCh38
rs757227559 Chr.3: 53497462-53497462 on Build GRCh38
rs763383320 Chr.3: 53497242-53497242 on Build GRCh38
rs368995943 Chr.3: 53497456-53497456 on Build GRCh38
rs371735256 Chr.3: 53497109-53497109 on Build GRCh38
rs780303281 Chr.3: 53497487-53497487 on Build GRCh38
rs754407641 Chr.3: 53497146-53497146 on Build GRCh38
rs746130121 Chr.3: 53497230-53497230 on Build GRCh38
rs955939748 Chr.3: 53497517-53497517 on Build GRCh38
rs769128755 Chr.3: 53497374-53497374 on Build GRCh38
rs751925700 Chr.3: 53497128-53497128 on Build GRCh38
rs759240348 Chr.3: 53497284-53497284 on Build GRCh38
rs150067408 Chr.3: 53497156-53497156 on Build GRCh38

View Full Product Details