Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74483096-74483687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC142
Entrez Gene ID
84865
Gene Name
coiled-coil domain containing 142
Gene Aliases
-
UniGene
Hs.430199
Gene Chromosome Location
Chr.2: 74472832-74483230 on Build GRCh38
Transcripts
NM_032779.3 NM_022492.4 NR_027749.1 XM_005264492.1 XM_011533035.1 XM_011533036.1 XM_011533037.1 XM_011533040.1 XM_017004706.1 XM_017004707.1 XM_017004708.1

Gene Symbol
TTC31
Entrez Gene ID
64427
Gene Name
tetratricopeptide repeat domain 31
Gene Aliases
-
UniGene
Hs.557709
Gene Chromosome Location
Chr.2: 74483032-74494564 on Build GRCh38
Transcripts
NM_032779.3 NM_022492.4 NR_027749.1 XM_005264492.1 XM_011533035.1 XM_011533036.1 XM_011533037.1 XM_011533040.1 XM_017004706.1 XM_017004707.1 XM_017004708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155880 c.41-2A>G p.? Chr.2: 74483320-74483320 on Build GRCh38
7395597 c.107G>T p.G36V Chr.2: 74483388-74483388 on Build GRCh38
2155881 c.41-3T>G p.? Chr.2: 74483319-74483319 on Build GRCh38
5862873 c.29G>A p.R10Q Chr.2: 74483124-74483124 on Build GRCh38
3939169 c.49C>G p.L17V Chr.2: 74483330-74483330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570700130 Chr.2: 74483284-74483284 on Build GRCh38
rs948379043 Chr.2: 74483369-74483369 on Build GRCh38
rs748636073 Chr.2: 74483316-74483317 on Build GRCh38
rs946429506 Chr.2: 74483491-74483491 on Build GRCh38
rs748085480 Chr.2: 74483456-74483456 on Build GRCh38
rs754437644 Chr.2: 74483359-74483359 on Build GRCh38
rs776039947 Chr.2: 74483540-74483540 on Build GRCh38
rs780596274 Chr.2: 74483300-74483301 on Build GRCh38
rs1018892466 Chr.2: 74483276-74483276 on Build GRCh38
rs765794063 Chr.2: 74483352-74483352 on Build GRCh38
rs765682025 Chr.2: 74483367-74483367 on Build GRCh38
rs762200912 Chr.2: 74483574-74483574 on Build GRCh38
rs749888689 Chr.2: 74483150-74483150 on Build GRCh38
rs746761834 Chr.2: 74483510-74483510 on Build GRCh38
rs370951288 Chr.2: 74483141-74483141 on Build GRCh38
rs1042171118 Chr.2: 74483205-74483205 on Build GRCh38
rs554740962 Chr.2: 74483399-74483399 on Build GRCh38
rs766694753 Chr.2: 74483628-74483628 on Build GRCh38
rs776020436 Chr.2: 74483411-74483411 on Build GRCh38
rs747131687 Chr.2: 74483304-74483304 on Build GRCh38
rs756452664 Chr.2: 74483521-74483521 on Build GRCh38
rs778464092 Chr.2: 74483437-74483437 on Build GRCh38
rs770552540 Chr.2: 74483410-74483410 on Build GRCh38
rs373643480 Chr.2: 74483126-74483126 on Build GRCh38
rs781133847 Chr.2: 74483390-74483390 on Build GRCh38
rs773292601 Chr.2: 74483172-74483172 on Build GRCh38
rs779142193 Chr.2: 74483290-74483290 on Build GRCh38
rs761654775 Chr.2: 74483337-74483337 on Build GRCh38
rs761558785 Chr.2: 74483281-74483281 on Build GRCh38
rs779046721 Chr.2: 74483453-74483453 on Build GRCh38
rs117680729 Chr.2: 74483615-74483615 on Build GRCh38
rs767224931 Chr.2: 74483282-74483282 on Build GRCh38
rs192671393 Chr.2: 74483269-74483269 on Build GRCh38
rs768569204 Chr.2: 74483325-74483325 on Build GRCh38
rs751881361 Chr.2: 74483432-74483432 on Build GRCh38
rs902357669 Chr.2: 74483267-74483267 on Build GRCh38
rs753089585 Chr.2: 74483372-74483372 on Build GRCh38
rs774211184 Chr.2: 74483279-74483279 on Build GRCh38
rs576108592 Chr.2: 74483402-74483402 on Build GRCh38
rs750640111 Chr.2: 74483591-74483591 on Build GRCh38
rs772059812 Chr.2: 74483457-74483457 on Build GRCh38
rs948658284 Chr.2: 74483619-74483619 on Build GRCh38
rs756552041 Chr.2: 74483361-74483361 on Build GRCh38
rs767127270 Chr.2: 74483146-74483146 on Build GRCh38
rs775299088 Chr.2: 74483275-74483275 on Build GRCh38
rs763361228 Chr.2: 74483425-74483425 on Build GRCh38
rs755267402 Chr.2: 74483160-74483160 on Build GRCh38
rs756235884 Chr.2: 74483442-74483442 on Build GRCh38
rs763452342 Chr.2: 74483566-74483566 on Build GRCh38
rs952073788 Chr.2: 74483317-74483317 on Build GRCh38
rs377684910 Chr.2: 74483140-74483140 on Build GRCh38
rs1010687386 Chr.2: 74483183-74483183 on Build GRCh38
rs1006773844 Chr.2: 74483140-74483141 on Build GRCh38
rs754278748 Chr.2: 74483288-74483288 on Build GRCh38
rs774303672 Chr.2: 74483334-74483334 on Build GRCh38
rs762851632 Chr.2: 74483272-74483272 on Build GRCh38
rs888227401 Chr.2: 74483132-74483132 on Build GRCh38
rs778956839 Chr.2: 74483401-74483401 on Build GRCh38
rs577023398 Chr.2: 74483498-74483498 on Build GRCh38
rs1017617829 Chr.2: 74483559-74483559 on Build GRCh38
rs373601586 Chr.2: 74483408-74483408 on Build GRCh38
rs370970504 Chr.2: 74483427-74483427 on Build GRCh38
rs887794139 Chr.2: 74483647-74483647 on Build GRCh38
rs200922006 Chr.2: 74483115-74483115 on Build GRCh38
rs373770296 Chr.2: 74483326-74483326 on Build GRCh38
rs986159474 Chr.2: 74483236-74483236 on Build GRCh38
rs771885981 Chr.2: 74483339-74483339 on Build GRCh38
rs777930801 Chr.2: 74483301-74483301 on Build GRCh38
rs534915022 Chr.2: 74483289-74483289 on Build GRCh38
rs996504015 Chr.2: 74483639-74483639 on Build GRCh38
rs775552009 Chr.2: 74483316-74483316 on Build GRCh38
rs771974104 Chr.2: 74483404-74483404 on Build GRCh38
rs556659631 Chr.2: 74483644-74483645 on Build GRCh38
rs1044000730 Chr.2: 74483662-74483662 on Build GRCh38
rs569753223 Chr.2: 74483116-74483116 on Build GRCh38
rs986105439 Chr.2: 74483537-74483537 on Build GRCh38
rs769800794 Chr.2: 74483179-74483179 on Build GRCh38
rs760282084 Chr.2: 74483347-74483347 on Build GRCh38
rs751762496 Chr.2: 74483386-74483386 on Build GRCh38
rs775377929 Chr.2: 74483131-74483131 on Build GRCh38
rs140312972 Chr.2: 74483192-74483192 on Build GRCh38
rs749850269 Chr.2: 74483378-74483378 on Build GRCh38
rs745900934 Chr.2: 74483396-74483396 on Build GRCh38
rs560009541 Chr.2: 74483539-74483539 on Build GRCh38
rs185669877 Chr.2: 74483387-74483387 on Build GRCh38
rs6707475 Chr.2: 74483364-74483364 on Build GRCh38
rs747039843 Chr.2: 74483178-74483178 on Build GRCh38
rs1045997839 Chr.2: 74483118-74483118 on Build GRCh38
rs765772090 Chr.2: 74483419-74483419 on Build GRCh38
rs767675922 Chr.2: 74483584-74483584 on Build GRCh38
rs1042140044 Chr.2: 74483500-74483500 on Build GRCh38
rs914917744 Chr.2: 74483485-74483485 on Build GRCh38
rs374507059 Chr.2: 74483435-74483435 on Build GRCh38
rs1027615753 Chr.2: 74483642-74483642 on Build GRCh38
rs917138579 Chr.2: 74483612-74483612 on Build GRCh38
rs1027732071 Chr.2: 74483310-74483310 on Build GRCh38
rs756704368 Chr.2: 74483153-74483153 on Build GRCh38
rs759045341 Chr.2: 74483412-74483412 on Build GRCh38
rs772207545 Chr.2: 74483164-74483164 on Build GRCh38
rs748446505 Chr.2: 74483291-74483291 on Build GRCh38
rs772117460 Chr.2: 74483121-74483121 on Build GRCh38
rs778018836 Chr.2: 74483385-74483385 on Build GRCh38
rs749697429 Chr.2: 74483157-74483157 on Build GRCh38
rs761922844 Chr.2: 74483117-74483117 on Build GRCh38
rs553236961 Chr.2: 74483312-74483312 on Build GRCh38
rs749174701 Chr.2: 74483324-74483324 on Build GRCh38
rs368872326 Chr.2: 74483421-74483421 on Build GRCh38
rs751145498 Chr.2: 74483142-74483142 on Build GRCh38
rs954954527 Chr.2: 74483527-74483527 on Build GRCh38
rs373929649 Chr.2: 74483161-74483161 on Build GRCh38
rs775259581 Chr.2: 74483182-74483182 on Build GRCh38
rs781430195 Chr.2: 74483437-74483437 on Build GRCh38
rs80284005 Chr.2: 74483650-74483650 on Build GRCh38
rs565132032 Chr.2: 74483441-74483441 on Build GRCh38
rs776609740 Chr.2: 74483123-74483123 on Build GRCh38
rs754441894 Chr.2: 74483116-74483117 on Build GRCh38
rs1008078637 Chr.2: 74483357-74483357 on Build GRCh38
rs770519452 Chr.2: 74483520-74483520 on Build GRCh38
rs758730361 Chr.2: 74483375-74483375 on Build GRCh38
rs780380021 Chr.2: 74483155-74483155 on Build GRCh38
rs529628097 Chr.2: 74483573-74483573 on Build GRCh38
rs541284003 Chr.2: 74483525-74483525 on Build GRCh38
rs202176729 Chr.2: 74483342-74483342 on Build GRCh38
rs934495059 Chr.2: 74483580-74483580 on Build GRCh38
rs764523737 Chr.2: 74483569-74483569 on Build GRCh38

View Full Product Details