Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218737597-218738095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL4
Entrez Gene ID
9654
Gene Name
tubulin tyrosine ligase like 4
Gene Aliases
-
UniGene
Hs.471405
Gene Chromosome Location
Chr.2: 218710835-218757484 on Build GRCh38
Transcripts
NM_014640.4 XM_006712875.2 XM_017005387.1 XM_017005388.1 XM_017005389.1 XM_017005390.1 XM_017005391.1 XM_017005392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631846 c.313C>A p.L105I Chr.2: 218737989-218737989 on Build GRCh38
6298446 c.93G>T p.T31T Chr.2: 218737769-218737769 on Build GRCh38
4091520 c.76G>T p.G26C Chr.2: 218737752-218737752 on Build GRCh38
3391479 c.365G>C p.S122T Chr.2: 218738041-218738041 on Build GRCh38
6298447 c.396G>T p.E132D Chr.2: 218738072-218738072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368505769 Chr.2: 218737995-218737995 on Build GRCh38
rs769117546 Chr.2: 218737844-218737844 on Build GRCh38
rs757483368 Chr.2: 218737699-218737699 on Build GRCh38
rs766751761 Chr.2: 218738035-218738035 on Build GRCh38
rs115468380 Chr.2: 218737895-218737895 on Build GRCh38
rs767775738 Chr.2: 218737931-218737931 on Build GRCh38
rs770593465 Chr.2: 218737809-218737809 on Build GRCh38
rs956396156 Chr.2: 218737877-218737877 on Build GRCh38
rs372440850 Chr.2: 218737734-218737734 on Build GRCh38
rs774166545 Chr.2: 218737629-218737629 on Build GRCh38
rs912127172 Chr.2: 218737902-218737902 on Build GRCh38
rs962219133 Chr.2: 218737760-218737760 on Build GRCh38
rs745376125 Chr.2: 218737829-218737829 on Build GRCh38
rs558021515 Chr.2: 218737672-218737672 on Build GRCh38
rs980791875 Chr.2: 218738066-218738066 on Build GRCh38
rs763454705 Chr.2: 218737772-218737772 on Build GRCh38
rs979339807 Chr.2: 218737860-218737860 on Build GRCh38
rs368175704 Chr.2: 218737695-218737695 on Build GRCh38
rs201497979 Chr.2: 218738048-218738048 on Build GRCh38
rs748695101 Chr.2: 218738026-218738026 on Build GRCh38
rs530693896 Chr.2: 218737652-218737652 on Build GRCh38
rs781044606 Chr.2: 218738000-218738000 on Build GRCh38
rs200689128 Chr.2: 218738029-218738029 on Build GRCh38
rs758377105 Chr.2: 218737962-218737962 on Build GRCh38
rs750503144 Chr.2: 218737786-218737786 on Build GRCh38
rs11542786 Chr.2: 218737726-218737726 on Build GRCh38
rs760969926 Chr.2: 218737942-218737942 on Build GRCh38
rs773070468 Chr.2: 218737653-218737653 on Build GRCh38
rs769140870 Chr.2: 218738017-218738017 on Build GRCh38
rs779498722 Chr.2: 218738013-218738013 on Build GRCh38
rs943392250 Chr.2: 218737953-218737953 on Build GRCh38
rs760046892 Chr.2: 218737670-218737670 on Build GRCh38
rs974687265 Chr.2: 218737975-218737975 on Build GRCh38
rs759013360 Chr.2: 218737757-218737757 on Build GRCh38
rs199689447 Chr.2: 218738020-218738020 on Build GRCh38
rs750373542 Chr.2: 218737645-218737645 on Build GRCh38
rs752627930 Chr.2: 218737788-218737788 on Build GRCh38
rs1018392361 Chr.2: 218737939-218737939 on Build GRCh38
rs781293566 Chr.2: 218737707-218737707 on Build GRCh38
rs760411669 Chr.2: 218737655-218737655 on Build GRCh38
rs955325644 Chr.2: 218737851-218737851 on Build GRCh38
rs778775604 Chr.2: 218737716-218737716 on Build GRCh38
rs769192243 Chr.2: 218737901-218737901 on Build GRCh38
rs139343526 Chr.2: 218737768-218737768 on Build GRCh38
rs528176022 Chr.2: 218737712-218737712 on Build GRCh38
rs919731277 Chr.2: 218737994-218737994 on Build GRCh38
rs770517720 Chr.2: 218737749-218737749 on Build GRCh38
rs894844437 Chr.2: 218737783-218737783 on Build GRCh38
rs114586336 Chr.2: 218738031-218738031 on Build GRCh38
rs575186082 Chr.2: 218738038-218738038 on Build GRCh38
rs764294698 Chr.2: 218737692-218737692 on Build GRCh38
rs140821266 Chr.2: 218737793-218737793 on Build GRCh38
rs773652562 Chr.2: 218738032-218738032 on Build GRCh38
rs911926983 Chr.2: 218737639-218737639 on Build GRCh38
rs374640625 Chr.2: 218738056-218738056 on Build GRCh38
rs771930908 Chr.2: 218737730-218737730 on Build GRCh38
rs372893141 Chr.2: 218737929-218737929 on Build GRCh38
rs61733652 Chr.2: 218737817-218737817 on Build GRCh38
rs774678094 Chr.2: 218737913-218737913 on Build GRCh38
rs934888085 Chr.2: 218737617-218737617 on Build GRCh38
rs758435339 Chr.2: 218737791-218737791 on Build GRCh38
rs776827335 Chr.2: 218738036-218738036 on Build GRCh38
rs3731877 Chr.2: 218737776-218737776 on Build GRCh38
rs199614245 Chr.2: 218737926-218737926 on Build GRCh38
rs745977672 Chr.2: 218737709-218737709 on Build GRCh38
rs920687045 Chr.2: 218737991-218737991 on Build GRCh38
rs558671258 Chr.2: 218737680-218737680 on Build GRCh38
rs763400004 Chr.2: 218737896-218737896 on Build GRCh38
rs973964811 Chr.2: 218737990-218737990 on Build GRCh38
rs757050930 Chr.2: 218737621-218737621 on Build GRCh38
rs370489811 Chr.2: 218737642-218737642 on Build GRCh38
rs776173312 Chr.2: 218737753-218737753 on Build GRCh38
rs148148927 Chr.2: 218738034-218738034 on Build GRCh38
rs761992701 Chr.2: 218737782-218737782 on Build GRCh38
rs926617358 Chr.2: 218738075-218738075 on Build GRCh38
rs758816293 Chr.2: 218737682-218737682 on Build GRCh38
rs765700041 Chr.2: 218737679-218737679 on Build GRCh38
rs144997742 Chr.2: 218737933-218737933 on Build GRCh38
rs114179677 Chr.2: 218738014-218738014 on Build GRCh38
rs764119204 Chr.2: 218738057-218738057 on Build GRCh38
rs754005610 Chr.2: 218737948-218737948 on Build GRCh38
rs79987978 Chr.2: 218737787-218737787 on Build GRCh38
rs375272990 Chr.2: 218737666-218737666 on Build GRCh38
rs774884769 Chr.2: 218738076-218738077 on Build GRCh38
rs537717303 Chr.2: 218737804-218737804 on Build GRCh38
rs757231512 Chr.2: 218738077-218738077 on Build GRCh38
rs144380659 Chr.2: 218737774-218737774 on Build GRCh38
rs747956757 Chr.2: 218737717-218737717 on Build GRCh38
rs568149810 Chr.2: 218737769-218737769 on Build GRCh38
rs115949954 Chr.2: 218738002-218738002 on Build GRCh38
rs767683905 Chr.2: 218737785-218737785 on Build GRCh38
rs745453069 Chr.2: 218738001-218738001 on Build GRCh38
rs771412079 Chr.2: 218737730-218737731 on Build GRCh38
rs1031877599 Chr.2: 218737863-218737863 on Build GRCh38
rs200209261 Chr.2: 218737996-218737996 on Build GRCh38
rs376884357 Chr.2: 218737719-218737719 on Build GRCh38
rs751472856 Chr.2: 218738067-218738067 on Build GRCh38
rs371006988 Chr.2: 218737957-218737957 on Build GRCh38

View Full Product Details