Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24114201-24114794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
UniGene
Hs.525251
Gene Chromosome Location
Chr.14: 24114697-24125242 on Build GRCh38
Transcripts
NM_001163484.1 NM_025230.4 NR_028099.1 XM_005267708.4 XM_005267710.3 XM_017021350.1

Gene Symbol
NRL
Entrez Gene ID
4901
Gene Name
neural retina leucine zipper
Gene Aliases
D14S46E, NRL-MAF, RP27
UniGene
Hs.652297
Gene Chromosome Location
Chr.14: 24078693-24114924 on Build GRCh38
Transcripts
NM_001163484.1 NM_025230.4 NR_028099.1 XM_005267708.4 XM_005267710.3 XM_017021350.1

SNP Detail

SNP ID Chromosome Location
rs1004415175 Chr.14: 24114471-24114471 on Build GRCh38
rs1048506766 Chr.14: 24114741-24114741 on Build GRCh38
rs3825583 Chr.14: 24114637-24114637 on Build GRCh38
rs975803113 Chr.14: 24114236-24114236 on Build GRCh38
rs775851943 Chr.14: 24114647-24114647 on Build GRCh38
rs953227538 Chr.14: 24114606-24114607 on Build GRCh38
rs1006346760 Chr.14: 24114442-24114442 on Build GRCh38
rs1054415488 Chr.14: 24114275-24114275 on Build GRCh38
rs891722625 Chr.14: 24114281-24114281 on Build GRCh38
rs569926197 Chr.14: 24114416-24114416 on Build GRCh38
rs947250301 Chr.14: 24114339-24114339 on Build GRCh38
rs945965467 Chr.14: 24114640-24114640 on Build GRCh38
rs1024996313 Chr.14: 24114536-24114536 on Build GRCh38
rs901541661 Chr.14: 24114678-24114678 on Build GRCh38
rs961721601 Chr.14: 24114230-24114230 on Build GRCh38
rs935910383 Chr.14: 24114256-24114256 on Build GRCh38
rs888629514 Chr.14: 24114447-24114447 on Build GRCh38
rs370604662 Chr.14: 24114254-24114254 on Build GRCh38
rs747507471 Chr.14: 24114653-24114653 on Build GRCh38
rs927118401 Chr.14: 24114589-24114589 on Build GRCh38
rs777869812 Chr.14: 24114263-24114263 on Build GRCh38
rs565325566 Chr.14: 24114572-24114572 on Build GRCh38
rs1029567007 Chr.14: 24114385-24114385 on Build GRCh38
rs887532354 Chr.14: 24114706-24114706 on Build GRCh38
rs535706101 Chr.14: 24114484-24114484 on Build GRCh38
rs901566598 Chr.14: 24114350-24114350 on Build GRCh38
rs980322836 Chr.14: 24114229-24114229 on Build GRCh38
rs931721536 Chr.14: 24114687-24114687 on Build GRCh38
rs1025822979 Chr.14: 24114670-24114670 on Build GRCh38
rs962280234 Chr.14: 24114476-24114476 on Build GRCh38
rs1015838764 Chr.14: 24114473-24114473 on Build GRCh38
rs977331500 Chr.14: 24114645-24114645 on Build GRCh38
rs945661446 Chr.14: 24114295-24114295 on Build GRCh38
rs182875006 Chr.14: 24114491-24114491 on Build GRCh38
rs997169330 Chr.14: 24114501-24114504 on Build GRCh38
rs969863393 Chr.14: 24114549-24114549 on Build GRCh38
rs1030014860 Chr.14: 24114591-24114591 on Build GRCh38
rs980914604 Chr.14: 24114558-24114558 on Build GRCh38
rs557545991 Chr.14: 24114665-24114665 on Build GRCh38
rs992095225 Chr.14: 24114479-24114479 on Build GRCh38
rs187479944 Chr.14: 24114737-24114737 on Build GRCh38
rs550314882 Chr.14: 24114379-24114379 on Build GRCh38
rs1041663251 Chr.14: 24114340-24114340 on Build GRCh38
rs898062450 Chr.14: 24114774-24114774 on Build GRCh38
rs1040045684 Chr.14: 24114661-24114661 on Build GRCh38
rs1057218183 Chr.14: 24114398-24114398 on Build GRCh38
rs749305477 Chr.14: 24114298-24114298 on Build GRCh38
rs1049001387 Chr.14: 24114428-24114428 on Build GRCh38
rs3811175 Chr.14: 24114255-24114255 on Build GRCh38
rs1050142726 Chr.14: 24114691-24114691 on Build GRCh38
rs887990223 Chr.14: 24114409-24114409 on Build GRCh38
rs894579758 Chr.14: 24114426-24114426 on Build GRCh38
rs985898695 Chr.14: 24114616-24114616 on Build GRCh38
rs1015095886 Chr.14: 24114465-24114465 on Build GRCh38
rs746093247 Chr.14: 24114634-24114634 on Build GRCh38
rs888611702 Chr.14: 24114757-24114757 on Build GRCh38
rs879622712 Chr.14: 24114718-24114718 on Build GRCh38
rs779102158 Chr.14: 24114610-24114610 on Build GRCh38
rs914427906 Chr.14: 24114299-24114299 on Build GRCh38
rs533290455 Chr.14: 24114276-24114276 on Build GRCh38
rs986342747 Chr.14: 24114284-24114284 on Build GRCh38
rs924597922 Chr.14: 24114376-24114376 on Build GRCh38

View Full Product Details